Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28136714-28137258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAN3-AS1
Entrez Gene ID
100288730
Gene Name
PAN3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.123191
Gene Chromosome Location
Chr.13: 28136843-28139174 on Build GRCh38
Transcripts
NR_029383.1

SNP Detail

SNP ID Chromosome Location
rs920514368 Chr.13: 28137149-28137149 on Build GRCh38
rs984123767 Chr.13: 28136876-28136876 on Build GRCh38
rs537403156 Chr.13: 28136968-28136968 on Build GRCh38
rs998507036 Chr.13: 28137072-28137072 on Build GRCh38
rs78042704 Chr.13: 28137113-28137113 on Build GRCh38
rs920596929 Chr.13: 28136844-28136844 on Build GRCh38
rs911870470 Chr.13: 28137206-28137206 on Build GRCh38
rs890283048 Chr.13: 28137199-28137199 on Build GRCh38
rs922087650 Chr.13: 28136940-28136940 on Build GRCh38
rs776811917 Chr.13: 28136895-28136895 on Build GRCh38
rs958283194 Chr.13: 28137004-28137004 on Build GRCh38
rs983497620 Chr.13: 28137201-28137201 on Build GRCh38
rs1027709379 Chr.13: 28137156-28137156 on Build GRCh38
rs970148142 Chr.13: 28136755-28136755 on Build GRCh38
rs750452077 Chr.13: 28137053-28137053 on Build GRCh38
rs546072054 Chr.13: 28137026-28137026 on Build GRCh38
rs759367167 Chr.13: 28136752-28136752 on Build GRCh38
rs765330044 Chr.13: 28136950-28136950 on Build GRCh38
rs61260403 Chr.13: 28136879-28136879 on Build GRCh38
rs375813506 Chr.13: 28137139-28137139 on Build GRCh38
rs950711852 Chr.13: 28136872-28136872 on Build GRCh38
rs761920892 Chr.13: 28136906-28136906 on Build GRCh38
rs369184013 Chr.13: 28136763-28136763 on Build GRCh38
rs1037592555 Chr.13: 28136800-28136800 on Build GRCh38
rs542069146 Chr.13: 28137138-28137138 on Build GRCh38
rs866228127 Chr.13: 28136792-28136792 on Build GRCh38
rs991431573 Chr.13: 28137013-28137013 on Build GRCh38
rs569131804 Chr.13: 28137131-28137131 on Build GRCh38
rs960162929 Chr.13: 28136738-28136738 on Build GRCh38
rs967049189 Chr.13: 28136919-28136919 on Build GRCh38
rs566651114 Chr.13: 28136743-28136743 on Build GRCh38
rs966320112 Chr.13: 28137215-28137215 on Build GRCh38
rs189687591 Chr.13: 28136874-28136874 on Build GRCh38
rs556956063 Chr.13: 28136984-28136984 on Build GRCh38
rs141417654 Chr.13: 28137114-28137114 on Build GRCh38
rs529085363 Chr.13: 28136913-28136913 on Build GRCh38
rs1053178925 Chr.13: 28137089-28137089 on Build GRCh38
rs1050235361 Chr.13: 28136847-28136847 on Build GRCh38
rs904195626 Chr.13: 28137038-28137038 on Build GRCh38
rs763466648 Chr.13: 28137103-28137103 on Build GRCh38
rs895761358 Chr.13: 28137109-28137109 on Build GRCh38
rs899058167 Chr.13: 28136814-28136814 on Build GRCh38
rs1022789814 Chr.13: 28137132-28137132 on Build GRCh38
rs996414099 Chr.13: 28136831-28136831 on Build GRCh38
rs568469525 Chr.13: 28136931-28136931 on Build GRCh38
rs950797351 Chr.13: 28137167-28137167 on Build GRCh38
rs181240120 Chr.13: 28137024-28137024 on Build GRCh38
rs969822193 Chr.13: 28137136-28137136 on Build GRCh38
rs916753821 Chr.13: 28137022-28137022 on Build GRCh38
rs1019991309 Chr.13: 28136896-28136896 on Build GRCh38

View Full Product Details