Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28138162-28138663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAN3
Entrez Gene ID
255967
Gene Name
PAN3 poly(A) specific ribonuclease subunit
Gene Aliases
-
UniGene
Hs.645015
Gene Chromosome Location
Chr.13: 28138079-28295338 on Build GRCh38
Transcripts
NM_175854.7 XM_005266332.3 XM_005266333.3 XM_011535032.2 XM_011535033.2 XM_011535034.2 XM_017020528.1 XM_017020530.1 XM_017020533.1 NR_029383.1

Gene Symbol
PAN3-AS1
Entrez Gene ID
100288730
Gene Name
PAN3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.123191
Gene Chromosome Location
Chr.13: 28136843-28139174 on Build GRCh38
Transcripts
NM_175854.7 XM_005266332.3 XM_005266333.3 XM_011535032.2 XM_011535033.2 XM_011535034.2 XM_017020528.1 XM_017020530.1 XM_017020533.1 NR_029383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
401415 c.5A>T p.N2I Chr.13: 28138662-28138662 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745694355 Chr.13: 28138197-28138197 on Build GRCh38
rs191977107 Chr.13: 28138452-28138452 on Build GRCh38
rs907525751 Chr.13: 28138419-28138419 on Build GRCh38
rs902713348 Chr.13: 28138628-28138629 on Build GRCh38
rs770559488 Chr.13: 28138619-28138624 on Build GRCh38
rs531561215 Chr.13: 28138300-28138300 on Build GRCh38
rs892741732 Chr.13: 28138357-28138357 on Build GRCh38
rs919553519 Chr.13: 28138446-28138446 on Build GRCh38
rs1018987406 Chr.13: 28138343-28138343 on Build GRCh38
rs957479428 Chr.13: 28138386-28138386 on Build GRCh38
rs1022825176 Chr.13: 28138596-28138596 on Build GRCh38
rs138028125 Chr.13: 28138552-28138552 on Build GRCh38
rs570586659 Chr.13: 28138600-28138602 on Build GRCh38
rs73445093 Chr.13: 28138252-28138252 on Build GRCh38
rs923107962 Chr.13: 28138448-28138448 on Build GRCh38
rs892464754 Chr.13: 28138576-28138576 on Build GRCh38
rs1040498162 Chr.13: 28138543-28138543 on Build GRCh38
rs758700541 Chr.13: 28138600-28138601 on Build GRCh38
rs550656977 Chr.13: 28138360-28138360 on Build GRCh38
rs139276478 Chr.13: 28138606-28138607 on Build GRCh38
rs1045412909 Chr.13: 28138220-28138220 on Build GRCh38
rs981203327 Chr.13: 28138615-28138615 on Build GRCh38
rs949642052 Chr.13: 28138586-28138586 on Build GRCh38
rs928364789 Chr.13: 28138409-28138409 on Build GRCh38
rs376891155 Chr.13: 28138232-28138232 on Build GRCh38
rs187574197 Chr.13: 28138567-28138567 on Build GRCh38
rs1050516778 Chr.13: 28138533-28138533 on Build GRCh38
rs971156228 Chr.13: 28138200-28138200 on Build GRCh38
rs926966680 Chr.13: 28138209-28138209 on Build GRCh38
rs757526837 Chr.13: 28138398-28138398 on Build GRCh38
rs932056218 Chr.13: 28138242-28138242 on Build GRCh38
rs969856105 Chr.13: 28138396-28138396 on Build GRCh38
rs186148903 Chr.13: 28138376-28138376 on Build GRCh38
rs1002305222 Chr.13: 28138597-28138597 on Build GRCh38
rs1046599751 Chr.13: 28138590-28138590 on Build GRCh38
rs575747785 Chr.13: 28138594-28138594 on Build GRCh38
rs779468176 Chr.13: 28138603-28138604 on Build GRCh38
rs979821655 Chr.13: 28138205-28138205 on Build GRCh38
rs961104371 Chr.13: 28138633-28138633 on Build GRCh38
rs940601759 Chr.13: 28138228-28138228 on Build GRCh38
rs943030200 Chr.13: 28138624-28138624 on Build GRCh38
rs549671670 Chr.13: 28138502-28138502 on Build GRCh38
rs887553638 Chr.13: 28138620-28138620 on Build GRCh38
rs901887868 Chr.13: 28138551-28138551 on Build GRCh38
rs1049689267 Chr.13: 28138257-28138257 on Build GRCh38
rs1025140093 Chr.13: 28138373-28138373 on Build GRCh38
rs931771734 Chr.13: 28138527-28138527 on Build GRCh38
rs1001248535 Chr.13: 28138259-28138259 on Build GRCh38
rs1034034469 Chr.13: 28138381-28138381 on Build GRCh38
rs770264934 Chr.13: 28138589-28138589 on Build GRCh38
rs764258992 Chr.13: 28138628-28138628 on Build GRCh38
rs889249279 Chr.13: 28138260-28138260 on Build GRCh38
rs889271646 Chr.13: 28138483-28138494 on Build GRCh38
rs951764736 Chr.13: 28138394-28138394 on Build GRCh38
rs925380691 Chr.13: 28138549-28138549 on Build GRCh38
rs549125084 Chr.13: 28138251-28138251 on Build GRCh38
rs1011890076 Chr.13: 28138591-28138591 on Build GRCh38
rs898043064 Chr.13: 28138241-28138241 on Build GRCh38
rs1050464870 Chr.13: 28138250-28138250 on Build GRCh38
rs1025087730 Chr.13: 28138185-28138185 on Build GRCh38
rs1008039974 Chr.13: 28138277-28138277 on Build GRCh38
rs962308816 Chr.13: 28138427-28138427 on Build GRCh38
rs371507953 Chr.13: 28138540-28138540 on Build GRCh38
rs567119201 Chr.13: 28138480-28138480 on Build GRCh38
rs1031667591 Chr.13: 28138562-28138562 on Build GRCh38
rs975957746 Chr.13: 28138444-28138444 on Build GRCh38
rs141596979 Chr.13: 28138427-28138430 on Build GRCh38
rs966045758 Chr.13: 28138375-28138375 on Build GRCh38
rs990033490 Chr.13: 28138392-28138392 on Build GRCh38
rs907761556 Chr.13: 28138226-28138226 on Build GRCh38
rs539402062 Chr.13: 28138378-28138378 on Build GRCh38
rs752407696 Chr.13: 28138283-28138283 on Build GRCh38
rs923096592 Chr.13: 28138239-28138239 on Build GRCh38
rs951768386 Chr.13: 28138219-28138219 on Build GRCh38
rs118190974 Chr.13: 28138222-28138222 on Build GRCh38
rs111938639 Chr.13: 28138606-28138606 on Build GRCh38

View Full Product Details