Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25239636-25240136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORD115-38
Entrez Gene ID
100033812
Gene Name
small nucleolar RNA, C/D box 115-38
Gene Aliases
HBII-52-38
UniGene
Gene Chromosome Location
Chr.15: 25239838-25239919 on Build GRCh38
Transcripts
NR_003353.1

SNP Detail

SNP ID Chromosome Location
rs903552770 Chr.15: 25239759-25239759 on Build GRCh38
rs780871483 Chr.15: 25239824-25239824 on Build GRCh38
rs761679776 Chr.15: 25239943-25239943 on Build GRCh38
rs545467621 Chr.15: 25240045-25240045 on Build GRCh38
rs774504032 Chr.15: 25240051-25240051 on Build GRCh38
rs138150496 Chr.15: 25239854-25239854 on Build GRCh38
rs1013134391 Chr.15: 25239753-25239753 on Build GRCh38
rs777850790 Chr.15: 25239946-25239946 on Build GRCh38
rs892213824 Chr.15: 25239974-25239974 on Build GRCh38
rs376807616 Chr.15: 25239877-25239877 on Build GRCh38
rs1052077412 Chr.15: 25239972-25239972 on Build GRCh38
rs549237430 Chr.15: 25239793-25239793 on Build GRCh38
rs773742240 Chr.15: 25239847-25239847 on Build GRCh38
rs755655165 Chr.15: 25239831-25239831 on Build GRCh38
rs72546396 Chr.15: 25239813-25239813 on Build GRCh38
rs956468927 Chr.15: 25239778-25239778 on Build GRCh38
rs758208684 Chr.15: 25239804-25239804 on Build GRCh38
rs377345192 Chr.15: 25240034-25240034 on Build GRCh38
rs753511924 Chr.15: 25239949-25239949 on Build GRCh38
rs759743627 Chr.15: 25239794-25239794 on Build GRCh38
rs906383851 Chr.15: 25239677-25239677 on Build GRCh38
rs555713011 Chr.15: 25239675-25239675 on Build GRCh38
rs142683632 Chr.15: 25239995-25239995 on Build GRCh38
rs997241616 Chr.15: 25239991-25239991 on Build GRCh38
rs369495212 Chr.15: 25239882-25239882 on Build GRCh38
rs777056706 Chr.15: 25239859-25239859 on Build GRCh38
rs888988808 Chr.15: 25240000-25240000 on Build GRCh38
rs560311594 Chr.15: 25240054-25240054 on Build GRCh38
rs899123810 Chr.15: 25239685-25239685 on Build GRCh38
rs181951596 Chr.15: 25239875-25239875 on Build GRCh38
rs1009315874 Chr.15: 25240118-25240118 on Build GRCh38
rs945163457 Chr.15: 25239670-25239670 on Build GRCh38
rs1026350557 Chr.15: 25239704-25239704 on Build GRCh38
rs755675884 Chr.15: 25239892-25239892 on Build GRCh38
rs527310686 Chr.15: 25239963-25239963 on Build GRCh38
rs534273303 Chr.15: 25239917-25239917 on Build GRCh38
rs750090458 Chr.15: 25239883-25239883 on Build GRCh38
rs550953349 Chr.15: 25239676-25239676 on Build GRCh38
rs746259900 Chr.15: 25239966-25239966 on Build GRCh38
rs994792467 Chr.15: 25239894-25239894 on Build GRCh38
rs774275949 Chr.15: 25239933-25239933 on Build GRCh38
rs770926572 Chr.15: 25239948-25239948 on Build GRCh38
rs752681998 Chr.15: 25239799-25239799 on Build GRCh38
rs745515885 Chr.15: 25239829-25239829 on Build GRCh38
rs950773498 Chr.15: 25239709-25239709 on Build GRCh38
rs759762095 Chr.15: 25239868-25239868 on Build GRCh38
rs987982640 Chr.15: 25239730-25239730 on Build GRCh38
rs565650395 Chr.15: 25239909-25239909 on Build GRCh38
rs572399836 Chr.15: 25240055-25240055 on Build GRCh38
rs909183062 Chr.15: 25239690-25239690 on Build GRCh38
rs981862790 Chr.15: 25240027-25240027 on Build GRCh38
rs913559446 Chr.15: 25239990-25239990 on Build GRCh38
rs574176008 Chr.15: 25239678-25239678 on Build GRCh38
rs772086105 Chr.15: 25240068-25240068 on Build GRCh38
rs542616051 Chr.15: 25240089-25240089 on Build GRCh38
rs541019910 Chr.15: 25239772-25239772 on Build GRCh38
rs752129205 Chr.15: 25239952-25239952 on Build GRCh38
rs1034028656 Chr.15: 25239681-25239681 on Build GRCh38
rs577731193 Chr.15: 25240041-25240041 on Build GRCh38
rs745830241 Chr.15: 25240064-25240064 on Build GRCh38
rs540908757 Chr.15: 25239662-25239662 on Build GRCh38
rs777075182 Chr.15: 25239910-25239910 on Build GRCh38
rs751475781 Chr.15: 25239881-25239881 on Build GRCh38
rs1047689606 Chr.15: 25240073-25240073 on Build GRCh38
rs748713768 Chr.15: 25239841-25239841 on Build GRCh38
rs573476012 Chr.15: 25239660-25239660 on Build GRCh38
rs754516026 Chr.15: 25239950-25239950 on Build GRCh38
rs562649605 Chr.15: 25239698-25239698 on Build GRCh38
rs751314160 Chr.15: 25239817-25239817 on Build GRCh38
rs8025993 Chr.15: 25239947-25239947 on Build GRCh38
rs141571065 Chr.15: 25239701-25239701 on Build GRCh38
rs886417883 Chr.15: 25240079-25240079 on Build GRCh38
rs906340598 Chr.15: 25239873-25239873 on Build GRCh38
rs757299496 Chr.15: 25239870-25239871 on Build GRCh38
rs1044757870 Chr.15: 25239755-25239755 on Build GRCh38
rs566165541 Chr.15: 25239876-25239876 on Build GRCh38
rs776709897 Chr.15: 25239739-25239739 on Build GRCh38
rs373265471 Chr.15: 25239903-25239903 on Build GRCh38
rs772421893 Chr.15: 25239844-25239844 on Build GRCh38
rs762993363 Chr.15: 25239927-25239927 on Build GRCh38
rs544338116 Chr.15: 25239691-25239691 on Build GRCh38
rs754492922 Chr.15: 25239904-25239904 on Build GRCh38
rs957616713 Chr.15: 25239953-25239953 on Build GRCh38
rs556307855 Chr.15: 25240026-25240026 on Build GRCh38
rs114244379 Chr.15: 25239738-25239738 on Build GRCh38
rs778503120 Chr.15: 25239951-25239951 on Build GRCh38
rs201728057 Chr.15: 25239900-25239900 on Build GRCh38
rs770174413 Chr.15: 25239967-25239967 on Build GRCh38
rs562026187 Chr.15: 25239659-25239659 on Build GRCh38
rs550122171 Chr.15: 25240061-25240061 on Build GRCh38
rs775752036 Chr.15: 25239925-25239925 on Build GRCh38
rs747431473 Chr.15: 25239849-25239849 on Build GRCh38
rs1016166282 Chr.15: 25240039-25240039 on Build GRCh38
rs953299526 Chr.15: 25239682-25239688 on Build GRCh38
rs748485444 Chr.15: 25239767-25239767 on Build GRCh38
rs573953210 Chr.15: 25239962-25239962 on Build GRCh38
rs767320771 Chr.15: 25239945-25239945 on Build GRCh38
rs965259949 Chr.15: 25239732-25239732 on Build GRCh38
rs866892441 Chr.15: 25239820-25239820 on Build GRCh38
rs779375241 Chr.15: 25239838-25239838 on Build GRCh38
rs937807327 Chr.15: 25240043-25240043 on Build GRCh38
rs531547053 Chr.15: 25239818-25239818 on Build GRCh38
rs146177695 Chr.15: 25239731-25239731 on Build GRCh38
rs764221786 Chr.15: 25239929-25239929 on Build GRCh38

View Full Product Details