Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68424104-68424599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
UniGene
Hs.437249
Gene Chromosome Location
Chr.17: 68259089-68452000 on Build GRCh38
Transcripts
XM_005257170.3 XM_006721777.3 XM_011524538.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_017024361.1 XM_017024364.1 XM_017024365.1 XM_017024366.1 XM_017024367.1 NR_030365.1 NM_001278433.1 NM_001320772.1 NM_017983.6 NR_135470.1 NR_135471.1 XM_017024808.1

Gene Symbol
MIR635
Entrez Gene ID
693220
Gene Name
microRNA 635
Gene Aliases
MIRN635, hsa-mir-635
UniGene
Gene Chromosome Location
Chr.17: 68424451-68424548 on Build GRCh38
Transcripts
XM_005257170.3 XM_006721777.3 XM_011524538.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_017024361.1 XM_017024364.1 XM_017024365.1 XM_017024366.1 XM_017024367.1 NR_030365.1 NM_001278433.1 NM_001320772.1 NM_017983.6 NR_135470.1 NR_135471.1 XM_017024808.1

Gene Symbol
PRKAR1A
Entrez Gene ID
5573
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Gene Aliases
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
UniGene
Hs.280342
Gene Chromosome Location
Chr.17: 68413623-68551316 on Build GRCh38
Transcripts
XM_005257170.3 XM_006721777.3 XM_011524538.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_017024361.1 XM_017024364.1 XM_017024365.1 XM_017024366.1 XM_017024367.1 NR_030365.1 NM_001278433.1 NM_001320772.1 NM_017983.6 NR_135470.1 NR_135471.1 XM_017024808.1

Gene Symbol
WIPI1
Entrez Gene ID
55062
Gene Name
WD repeat domain, phosphoinositide interacting 1
Gene Aliases
ATG18, ATG18A, WIPI49
UniGene
Hs.463964
Gene Chromosome Location
Chr.17: 68421281-68457524 on Build GRCh38
Transcripts
XM_005257170.3 XM_006721777.3 XM_011524538.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_017024361.1 XM_017024364.1 XM_017024365.1 XM_017024366.1 XM_017024367.1 NR_030365.1 NM_001278433.1 NM_001320772.1 NM_017983.6 NR_135470.1 NR_135471.1 XM_017024808.1

SNP Detail

SNP ID Chromosome Location
rs867167673 Chr.17: 68424319-68424319 on Build GRCh38
rs919922579 Chr.17: 68424296-68424296 on Build GRCh38
rs887977285 Chr.17: 68424384-68424384 on Build GRCh38
rs763276694 Chr.17: 68424307-68424307 on Build GRCh38
rs894705937 Chr.17: 68424171-68424171 on Build GRCh38
rs375520837 Chr.17: 68424291-68424291 on Build GRCh38
rs371223760 Chr.17: 68424240-68424240 on Build GRCh38
rs192867879 Chr.17: 68424464-68424464 on Build GRCh38
rs1006800130 Chr.17: 68424429-68424429 on Build GRCh38
rs1015574533 Chr.17: 68424255-68424255 on Build GRCh38
rs747472544 Chr.17: 68424423-68424423 on Build GRCh38
rs369583542 Chr.17: 68424405-68424405 on Build GRCh38
rs770939948 Chr.17: 68424463-68424463 on Build GRCh38
rs8080370 Chr.17: 68424298-68424298 on Build GRCh38
rs780722943 Chr.17: 68424415-68424415 on Build GRCh38
rs773447750 Chr.17: 68424258-68424258 on Build GRCh38
rs188513993 Chr.17: 68424359-68424359 on Build GRCh38
rs902885888 Chr.17: 68424214-68424214 on Build GRCh38
rs369232831 Chr.17: 68424274-68424274 on Build GRCh38
rs938890796 Chr.17: 68424152-68424152 on Build GRCh38
rs761495041 Chr.17: 68424520-68424520 on Build GRCh38
rs550252218 Chr.17: 68424295-68424295 on Build GRCh38
rs916090560 Chr.17: 68424335-68424335 on Build GRCh38
rs749374782 Chr.17: 68424452-68424452 on Build GRCh38
rs768935510 Chr.17: 68424430-68424430 on Build GRCh38
rs917890975 Chr.17: 68424288-68424288 on Build GRCh38
rs765045345 Chr.17: 68424524-68424524 on Build GRCh38
rs560471254 Chr.17: 68424139-68424139 on Build GRCh38
rs372601596 Chr.17: 68424412-68424412 on Build GRCh38
rs542927034 Chr.17: 68424226-68424226 on Build GRCh38
rs751784346 Chr.17: 68424360-68424360 on Build GRCh38
rs776401033 Chr.17: 68424162-68424162 on Build GRCh38
rs1055926915 Chr.17: 68424308-68424308 on Build GRCh38
rs537208891 Chr.17: 68424392-68424392 on Build GRCh38
rs80130707 Chr.17: 68424285-68424285 on Build GRCh38
rs994692593 Chr.17: 68424516-68424516 on Build GRCh38
rs749867708 Chr.17: 68424412-68424412 on Build GRCh38
rs769090841 Chr.17: 68424535-68424536 on Build GRCh38
rs1007725770 Chr.17: 68424387-68424387 on Build GRCh38
rs776535164 Chr.17: 68424511-68424511 on Build GRCh38
rs1029117026 Chr.17: 68424165-68424165 on Build GRCh38
rs1056379879 Chr.17: 68424157-68424157 on Build GRCh38
rs185345320 Chr.17: 68424300-68424300 on Build GRCh38
rs928775176 Chr.17: 68424146-68424146 on Build GRCh38
rs758007938 Chr.17: 68424541-68424541 on Build GRCh38
rs995846581 Chr.17: 68424163-68424163 on Build GRCh38
rs74000709 Chr.17: 68424337-68424337 on Build GRCh38
rs765949725 Chr.17: 68424542-68424542 on Build GRCh38
rs116218906 Chr.17: 68424342-68424342 on Build GRCh38
rs969463770 Chr.17: 68424292-68424292 on Build GRCh38
rs781652035 Chr.17: 68424435-68424435 on Build GRCh38
rs781539636 Chr.17: 68424565-68424565 on Build GRCh38
rs760619413 Chr.17: 68424469-68424469 on Build GRCh38
rs532551557 Chr.17: 68424299-68424299 on Build GRCh38
rs750041849 Chr.17: 68424536-68424536 on Build GRCh38
rs756971319 Chr.17: 68424403-68424403 on Build GRCh38
rs748440771 Chr.17: 68424443-68424443 on Build GRCh38
rs995898999 Chr.17: 68424372-68424372 on Build GRCh38
rs951405251 Chr.17: 68424297-68424297 on Build GRCh38
rs145943696 Chr.17: 68424207-68424207 on Build GRCh38
rs755141098 Chr.17: 68424461-68424461 on Build GRCh38
rs368986755 Chr.17: 68424474-68424474 on Build GRCh38
rs115390549 Chr.17: 68424382-68424382 on Build GRCh38
rs1011836753 Chr.17: 68424192-68424192 on Build GRCh38
rs200337571 Chr.17: 68424557-68424557 on Build GRCh38
rs376493454 Chr.17: 68424558-68424558 on Build GRCh38
rs766487520 Chr.17: 68424356-68424356 on Build GRCh38
rs1007367299 Chr.17: 68424259-68424259 on Build GRCh38
rs77279010 Chr.17: 68424451-68424451 on Build GRCh38
rs757919596 Chr.17: 68424413-68424413 on Build GRCh38

View Full Product Details