Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202276034-202276519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP58
Entrez Gene ID
51602
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
HSPC120, NOP5, NOP5/NOP58
UniGene
Hs.471104
Gene Chromosome Location
Chr.2: 202265716-202303661 on Build GRCh38
Transcripts
NM_015934.4 NR_003058.1

Gene Symbol
SNORD70
Entrez Gene ID
692110
Gene Name
small nucleolar RNA, C/D box 70
Gene Aliases
HBII-234, SNORD70A
UniGene
Gene Chromosome Location
Chr.2: 202276431-202276518 on Build GRCh38
Transcripts
NM_015934.4 NR_003058.1

SNP Detail

SNP ID Chromosome Location
rs764403594 Chr.2: 202276400-202276400 on Build GRCh38
rs199644897 Chr.2: 202276492-202276492 on Build GRCh38
rs185886350 Chr.2: 202276478-202276478 on Build GRCh38
rs375709244 Chr.2: 202276340-202276341 on Build GRCh38
rs368740057 Chr.2: 202276487-202276487 on Build GRCh38
rs12463892 Chr.2: 202276494-202276494 on Build GRCh38
rs140927221 Chr.2: 202276341-202276341 on Build GRCh38
rs746596521 Chr.2: 202276438-202276438 on Build GRCh38
rs756190041 Chr.2: 202276493-202276493 on Build GRCh38
rs201484819 Chr.2: 202276472-202276472 on Build GRCh38
rs533165790 Chr.2: 202276259-202276259 on Build GRCh38
rs955521988 Chr.2: 202276221-202276221 on Build GRCh38
rs902297468 Chr.2: 202276095-202276095 on Build GRCh38
rs779260369 Chr.2: 202276436-202276436 on Build GRCh38
rs757996265 Chr.2: 202276393-202276394 on Build GRCh38
rs781270152 Chr.2: 202276432-202276432 on Build GRCh38
rs1015308981 Chr.2: 202276235-202276235 on Build GRCh38
rs559597033 Chr.2: 202276286-202276286 on Build GRCh38
rs62194069 Chr.2: 202276273-202276273 on Build GRCh38
rs1006537290 Chr.2: 202276137-202276137 on Build GRCh38
rs769736996 Chr.2: 202276458-202276458 on Build GRCh38
rs917734673 Chr.2: 202276164-202276164 on Build GRCh38
rs950210584 Chr.2: 202276169-202276169 on Build GRCh38
rs1056249886 Chr.2: 202276157-202276157 on Build GRCh38
rs1016328236 Chr.2: 202276210-202276210 on Build GRCh38
rs1029870775 Chr.2: 202276211-202276211 on Build GRCh38
rs547599733 Chr.2: 202276130-202276130 on Build GRCh38
rs763553895 Chr.2: 202276355-202276356 on Build GRCh38
rs759013242 Chr.2: 202276451-202276451 on Build GRCh38
rs754664661 Chr.2: 202276355-202276355 on Build GRCh38
rs767893413 Chr.2: 202276181-202276182 on Build GRCh38
rs1000174334 Chr.2: 202276201-202276201 on Build GRCh38
rs868594688 Chr.2: 202276378-202276378 on Build GRCh38
rs936066921 Chr.2: 202276371-202276371 on Build GRCh38
rs377430709 Chr.2: 202276297-202276297 on Build GRCh38
rs762830714 Chr.2: 202276468-202276468 on Build GRCh38
rs757570107 Chr.2: 202276405-202276405 on Build GRCh38
rs974404052 Chr.2: 202276124-202276124 on Build GRCh38
rs977973319 Chr.2: 202276347-202276347 on Build GRCh38
rs761468344 Chr.2: 202276446-202276446 on Build GRCh38
rs936398432 Chr.2: 202276104-202276104 on Build GRCh38
rs765236808 Chr.2: 202276477-202276477 on Build GRCh38
rs962365715 Chr.2: 202276059-202276059 on Build GRCh38
rs75014246 Chr.2: 202276342-202276342 on Build GRCh38
rs751891929 Chr.2: 202276402-202276402 on Build GRCh38
rs770600790 Chr.2: 202276441-202276441 on Build GRCh38
rs900110040 Chr.2: 202276203-202276203 on Build GRCh38
rs772343884 Chr.2: 202276202-202276202 on Build GRCh38
rs926511512 Chr.2: 202276126-202276126 on Build GRCh38
rs12988602 Chr.2: 202276127-202276127 on Build GRCh38
rs903192351 Chr.2: 202276198-202276198 on Build GRCh38
rs945198562 Chr.2: 202276348-202276348 on Build GRCh38
rs992028152 Chr.2: 202276296-202276296 on Build GRCh38
rs376953696 Chr.2: 202276435-202276435 on Build GRCh38
rs371977864 Chr.2: 202276433-202276433 on Build GRCh38
rs539199591 Chr.2: 202276272-202276272 on Build GRCh38
rs182198734 Chr.2: 202276228-202276228 on Build GRCh38
rs373147418 Chr.2: 202276356-202276357 on Build GRCh38
rs762091959 Chr.2: 202276482-202276482 on Build GRCh38
rs80119214 Chr.2: 202276339-202276339 on Build GRCh38
rs139513732 Chr.2: 202276357-202276357 on Build GRCh38
rs762879039 Chr.2: 202276385-202276385 on Build GRCh38
rs1041768353 Chr.2: 202276170-202276170 on Build GRCh38
rs192145727 Chr.2: 202276205-202276205 on Build GRCh38
rs775353980 Chr.2: 202276465-202276465 on Build GRCh38
rs749622358 Chr.2: 202276434-202276434 on Build GRCh38
rs924698136 Chr.2: 202276364-202276364 on Build GRCh38
rs756649993 Chr.2: 202276452-202276457 on Build GRCh38

View Full Product Details