Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 21285404-21285735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM2
Entrez Gene ID
4685
Gene Name
neural cell adhesion molecule 2
Gene Aliases
NCAM21
UniGene
Hs.473450
Gene Chromosome Location
Chr.21: 20998315-21543324 on Build GRCh38
Transcripts
NM_004540.3 XM_011529575.2 XM_011529576.2 XM_011529579.2 XM_011529580.2 XM_011529581.2 XM_011529582.2 XM_017028353.1 XM_017028354.1 XM_017028355.1 XM_017028356.1 XM_017028357.1 XM_017028358.1

SNP Detail

SNP ID Chromosome Location
rs1039199568 Chr.21: 21285463-21285463 on Build GRCh38
rs753295711 Chr.21: 21285442-21285442 on Build GRCh38
rs540591167 Chr.21: 21285519-21285519 on Build GRCh38
rs577354018 Chr.21: 21285595-21285595 on Build GRCh38
rs550737536 Chr.21: 21285534-21285534 on Build GRCh38
rs563228753 Chr.21: 21285667-21285667 on Build GRCh38
rs573389112 Chr.21: 21285503-21285503 on Build GRCh38
rs185585091 Chr.21: 21285675-21285675 on Build GRCh38
rs764727122 Chr.21: 21285566-21285566 on Build GRCh38
rs542432978 Chr.21: 21285689-21285689 on Build GRCh38
rs957457574 Chr.21: 21285652-21285652 on Build GRCh38
rs2826765 Chr.21: 21285441-21285441 on Build GRCh38
rs546362698 Chr.21: 21285476-21285476 on Build GRCh38
rs926153434 Chr.21: 21285621-21285621 on Build GRCh38
rs1017938580 Chr.21: 21285521-21285521 on Build GRCh38
rs762860109 Chr.21: 21285544-21285544 on Build GRCh38
rs1025935140 Chr.21: 21285462-21285462 on Build GRCh38
rs980983 Chr.21: 21285552-21285552 on Build GRCh38
rs933759297 Chr.21: 21285543-21285543 on Build GRCh38
rs977609102 Chr.21: 21285619-21285619 on Build GRCh38
rs544566156 Chr.21: 21285607-21285607 on Build GRCh38
rs1043657849 Chr.21: 21285696-21285696 on Build GRCh38
rs950058277 Chr.21: 21285693-21285693 on Build GRCh38
rs193103420 Chr.21: 21285437-21285437 on Build GRCh38
rs561637853 Chr.21: 21285429-21285429 on Build GRCh38
rs967916545 Chr.21: 21285614-21285614 on Build GRCh38
rs888201715 Chr.21: 21285482-21285482 on Build GRCh38

View Full Product Details