Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112648252-112648762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
UniGene
Hs.713663
Gene Chromosome Location
Chr.4: 112636964-112657592 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029857.1 NR_029858.1

Gene Symbol
MIR302B
Entrez Gene ID
442894
Gene Name
microRNA 302b
Gene Aliases
MIRN302B, mir-302b
UniGene
Gene Chromosome Location
Chr.4: 112648485-112648557 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029857.1 NR_029858.1

Gene Symbol
MIR302C
Entrez Gene ID
442895
Gene Name
microRNA 302c
Gene Aliases
MIRN302C, mir-302c
UniGene
Gene Chromosome Location
Chr.4: 112648363-112648430 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029857.1 NR_029858.1

SNP Detail

SNP ID Chromosome Location
rs936470649 Chr.4: 112648381-112648381 on Build GRCh38
rs190807868 Chr.4: 112648524-112648524 on Build GRCh38
rs375011523 Chr.4: 112648366-112648366 on Build GRCh38
rs940784199 Chr.4: 112648567-112648567 on Build GRCh38
rs899134458 Chr.4: 112648408-112648409 on Build GRCh38
rs550687674 Chr.4: 112648328-112648328 on Build GRCh38
rs948325214 Chr.4: 112648559-112648559 on Build GRCh38
rs754566437 Chr.4: 112648389-112648389 on Build GRCh38
rs552420111 Chr.4: 112648363-112648363 on Build GRCh38
rs540532231 Chr.4: 112648648-112648648 on Build GRCh38
rs537751343 Chr.4: 112648323-112648323 on Build GRCh38
rs991190599 Chr.4: 112648690-112648690 on Build GRCh38
rs372818265 Chr.4: 112648469-112648469 on Build GRCh38
rs200951552 Chr.4: 112648326-112648326 on Build GRCh38
rs375124094 Chr.4: 112648431-112648431 on Build GRCh38
rs764851038 Chr.4: 112648318-112648318 on Build GRCh38
rs759066554 Chr.4: 112648536-112648536 on Build GRCh38
rs749262875 Chr.4: 112648387-112648388 on Build GRCh38
rs185886239 Chr.4: 112648346-112648346 on Build GRCh38
rs756098665 Chr.4: 112648398-112648398 on Build GRCh38
rs750581183 Chr.4: 112648573-112648573 on Build GRCh38
rs781353576 Chr.4: 112648517-112648517 on Build GRCh38
rs1019196807 Chr.4: 112648658-112648658 on Build GRCh38
rs761878636 Chr.4: 112648561-112648561 on Build GRCh38
rs368089282 Chr.4: 112648314-112648314 on Build GRCh38
rs982349559 Chr.4: 112648709-112648709 on Build GRCh38
rs751245335 Chr.4: 112648383-112648383 on Build GRCh38
rs906647149 Chr.4: 112648384-112648384 on Build GRCh38
rs761761909 Chr.4: 112648339-112648339 on Build GRCh38
rs755087386 Chr.4: 112648591-112648591 on Build GRCh38
rs772656527 Chr.4: 112648372-112648372 on Build GRCh38
rs948247059 Chr.4: 112648382-112648382 on Build GRCh38
rs765952981 Chr.4: 112648516-112648516 on Build GRCh38
rs1050369161 Chr.4: 112648624-112648624 on Build GRCh38
rs563599426 Chr.4: 112648666-112648666 on Build GRCh38
rs780618243 Chr.4: 112648318-112648319 on Build GRCh38
rs750528251 Chr.4: 112648494-112648494 on Build GRCh38
rs970054344 Chr.4: 112648703-112648703 on Build GRCh38
rs770624774 Chr.4: 112648512-112648512 on Build GRCh38
rs987734352 Chr.4: 112648490-112648490 on Build GRCh38
rs73841094 Chr.4: 112648454-112648454 on Build GRCh38
rs758806646 Chr.4: 112648317-112648319 on Build GRCh38
rs373885472 Chr.4: 112648277-112648277 on Build GRCh38
rs994733390 Chr.4: 112648429-112648430 on Build GRCh38
rs928142591 Chr.4: 112648711-112648711 on Build GRCh38
rs757087059 Chr.4: 112648472-112648472 on Build GRCh38
rs773129212 Chr.4: 112648385-112648386 on Build GRCh38
rs767302290 Chr.4: 112648392-112648392 on Build GRCh38
rs752533511 Chr.4: 112648466-112648466 on Build GRCh38
rs769092848 Chr.4: 112648434-112648434 on Build GRCh38
rs774597032 Chr.4: 112648349-112648349 on Build GRCh38
rs774640335 Chr.4: 112648276-112648276 on Build GRCh38
rs574179913 Chr.4: 112648696-112648696 on Build GRCh38
rs765073078 Chr.4: 112648572-112648572 on Build GRCh38
rs373002110 Chr.4: 112648560-112648560 on Build GRCh38
rs763048957 Chr.4: 112648515-112648515 on Build GRCh38
rs372333642 Chr.4: 112648377-112648377 on Build GRCh38
rs747366066 Chr.4: 112648432-112648432 on Build GRCh38
rs775698536 Chr.4: 112648509-112648509 on Build GRCh38
rs775941393 Chr.4: 112648365-112648365 on Build GRCh38
rs762503340 Chr.4: 112648373-112648373 on Build GRCh38
rs916839397 Chr.4: 112648552-112648552 on Build GRCh38
rs770487324 Chr.4: 112648453-112648453 on Build GRCh38
rs775158555 Chr.4: 112648556-112648556 on Build GRCh38
rs533254696 Chr.4: 112648358-112648358 on Build GRCh38
rs961760351 Chr.4: 112648288-112648288 on Build GRCh38
rs749880723 Chr.4: 112648319-112648319 on Build GRCh38
rs771747691 Chr.4: 112648547-112648547 on Build GRCh38
rs779056483 Chr.4: 112648491-112648491 on Build GRCh38
rs914031261 Chr.4: 112648700-112648700 on Build GRCh38
rs763761427 Chr.4: 112648470-112648470 on Build GRCh38
rs1006161088 Chr.4: 112648307-112648307 on Build GRCh38
rs556661625 Chr.4: 112648540-112648540 on Build GRCh38
rs774151348 Chr.4: 112648520-112648520 on Build GRCh38
rs758620645 Chr.4: 112648672-112648672 on Build GRCh38
rs575331343 Chr.4: 112648558-112648558 on Build GRCh38
rs759670726 Chr.4: 112648298-112648298 on Build GRCh38
rs780401879 Chr.4: 112648502-112648502 on Build GRCh38
rs775067262 Chr.4: 112648576-112648577 on Build GRCh38
rs755982016 Chr.4: 112648338-112648338 on Build GRCh38
rs558284535 Chr.4: 112648612-112648612 on Build GRCh38
rs753607927 Chr.4: 112648280-112648280 on Build GRCh38
rs573266564 Chr.4: 112648644-112648644 on Build GRCh38
rs752482245 Chr.4: 112648395-112648395 on Build GRCh38
rs767140760 Chr.4: 112648462-112648462 on Build GRCh38
rs569377692 Chr.4: 112648330-112648330 on Build GRCh38
rs987252205 Chr.4: 112648296-112648296 on Build GRCh38
rs781066685 Chr.4: 112648343-112648345 on Build GRCh38
rs940835175 Chr.4: 112648735-112648735 on Build GRCh38
rs926516412 Chr.4: 112648667-112648667 on Build GRCh38
rs769996552 Chr.4: 112648344-112648344 on Build GRCh38
rs570633839 Chr.4: 112648435-112648435 on Build GRCh38
rs936548435 Chr.4: 112648669-112648669 on Build GRCh38
rs778812652 Chr.4: 112648410-112648410 on Build GRCh38
rs755262015 Chr.4: 112648324-112648338 on Build GRCh38
rs759249853 Chr.4: 112648459-112648459 on Build GRCh38
rs758753182 Chr.4: 112648500-112648500 on Build GRCh38
rs767199286 Chr.4: 112648569-112648611 on Build GRCh38
rs994828003 Chr.4: 112648603-112648603 on Build GRCh38
rs749371862 Chr.4: 112648408-112648408 on Build GRCh38
rs199971565 Chr.4: 112648384-112648387 on Build GRCh38
rs772356066 Chr.4: 112648417-112648417 on Build GRCh38
rs114318553 Chr.4: 112648539-112648539 on Build GRCh38
rs781667474 Chr.4: 112648593-112648593 on Build GRCh38
rs183105707 Chr.4: 112648536-112648536 on Build GRCh38
rs545458617 Chr.4: 112648584-112648584 on Build GRCh38
rs777740790 Chr.4: 112648405-112648405 on Build GRCh38
rs758053064 Chr.4: 112648325-112648325 on Build GRCh38
rs748092879 Chr.4: 112648336-112648336 on Build GRCh38
rs751843581 Chr.4: 112648588-112648588 on Build GRCh38
rs967213810 Chr.4: 112648660-112648660 on Build GRCh38
rs748444975 Chr.4: 112648605-112648605 on Build GRCh38
rs970331148 Chr.4: 112648630-112648630 on Build GRCh38
rs762177459 Chr.4: 112648438-112648438 on Build GRCh38
rs770686921 Chr.4: 112648434-112648448 on Build GRCh38
rs998616945 Chr.4: 112648662-112648662 on Build GRCh38
rs780918238 Chr.4: 112648335-112648335 on Build GRCh38
rs573054049 Chr.4: 112648645-112648645 on Build GRCh38
rs753839927 Chr.4: 112648471-112648471 on Build GRCh38
rs769821630 Chr.4: 112648386-112648391 on Build GRCh38
rs369989762 Chr.4: 112648368-112648368 on Build GRCh38
rs557950134 Chr.4: 112648586-112648586 on Build GRCh38
rs746108731 Chr.4: 112648416-112648416 on Build GRCh38
rs936909465 Chr.4: 112648353-112648353 on Build GRCh38

View Full Product Details