Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 71420553-71421088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00472
Entrez Gene ID
79940
Gene Name
long intergenic non-protein coding RNA 472
Gene Aliases
C6orf155
UniGene
Hs.368337
Gene Chromosome Location
Chr.6: 71407864-71420745 on Build GRCh38
Transcripts
NR_026807.2 NR_121612.1 NR_121613.1 NR_121614.1 NR_121615.1

Gene Symbol
LINC01626
Entrez Gene ID
102724000
Gene Name
long intergenic non-protein coding RNA 1626
Gene Aliases
-
UniGene
Hs.145785
Gene Chromosome Location
Chr.6: 71421027-71458872 on Build GRCh38
Transcripts
NR_026807.2 NR_121612.1 NR_121613.1 NR_121614.1 NR_121615.1

SNP Detail

SNP ID Chromosome Location
rs9455461 Chr.6: 71420945-71420945 on Build GRCh38
rs1037393072 Chr.6: 71420595-71420595 on Build GRCh38
rs1011155630 Chr.6: 71420687-71420687 on Build GRCh38
rs573794735 Chr.6: 71421013-71421013 on Build GRCh38
rs557775252 Chr.6: 71420812-71420812 on Build GRCh38
rs954289446 Chr.6: 71420686-71420686 on Build GRCh38
rs927376617 Chr.6: 71420828-71420828 on Build GRCh38
rs896976960 Chr.6: 71420599-71420599 on Build GRCh38
rs1012443065 Chr.6: 71420996-71420996 on Build GRCh38
rs778622006 Chr.6: 71420797-71420797 on Build GRCh38
rs578238916 Chr.6: 71420794-71420794 on Build GRCh38
rs972661073 Chr.6: 71420778-71420778 on Build GRCh38
rs889532510 Chr.6: 71420866-71420866 on Build GRCh38
rs1015380823 Chr.6: 71420635-71420635 on Build GRCh38
rs902514531 Chr.6: 71420964-71420964 on Build GRCh38
rs895301136 Chr.6: 71420992-71420992 on Build GRCh38
rs1017121445 Chr.6: 71420850-71420850 on Build GRCh38
rs576089889 Chr.6: 71420887-71420887 on Build GRCh38
rs1021749910 Chr.6: 71420921-71420921 on Build GRCh38
rs184443215 Chr.6: 71421008-71421008 on Build GRCh38
rs911054206 Chr.6: 71420878-71420878 on Build GRCh38
rs959567119 Chr.6: 71420832-71420832 on Build GRCh38
rs1003795545 Chr.6: 71420986-71420986 on Build GRCh38
rs986684989 Chr.6: 71420877-71420877 on Build GRCh38
rs138131362 Chr.6: 71421033-71421033 on Build GRCh38
rs1019674693 Chr.6: 71420868-71420868 on Build GRCh38
rs889626625 Chr.6: 71420795-71420795 on Build GRCh38
rs1021660594 Chr.6: 71420704-71420704 on Build GRCh38
rs146231298 Chr.6: 71420764-71420764 on Build GRCh38
rs547690374 Chr.6: 71420578-71420578 on Build GRCh38
rs1017528042 Chr.6: 71420662-71420662 on Build GRCh38
rs555712923 Chr.6: 71420739-71420739 on Build GRCh38
rs45487494 Chr.6: 71420719-71420719 on Build GRCh38
rs902734292 Chr.6: 71420809-71420809 on Build GRCh38
rs987035987 Chr.6: 71420690-71420690 on Build GRCh38
rs909556367 Chr.6: 71420587-71420587 on Build GRCh38
rs962385764 Chr.6: 71420851-71420851 on Build GRCh38
rs953655514 Chr.6: 71420803-71420803 on Build GRCh38
rs35318985 Chr.6: 71420801-71420801 on Build GRCh38
rs1035774069 Chr.6: 71420796-71420796 on Build GRCh38
rs1004453632 Chr.6: 71420609-71420609 on Build GRCh38
rs931169388 Chr.6: 71421046-71421046 on Build GRCh38
rs896428731 Chr.6: 71420836-71420836 on Build GRCh38
rs576522913 Chr.6: 71420741-71420742 on Build GRCh38
rs933869383 Chr.6: 71420871-71420871 on Build GRCh38
rs550588010 Chr.6: 71420701-71420701 on Build GRCh38
rs1057437791 Chr.6: 71420834-71420834 on Build GRCh38
rs1051777006 Chr.6: 71420616-71420616 on Build GRCh38
rs993900650 Chr.6: 71420950-71420950 on Build GRCh38
rs960532608 Chr.6: 71420608-71420608 on Build GRCh38
rs959076484 Chr.6: 71420753-71420753 on Build GRCh38
rs537537423 Chr.6: 71420730-71420730 on Build GRCh38
rs920906164 Chr.6: 71420671-71420671 on Build GRCh38
rs998455167 Chr.6: 71420601-71420601 on Build GRCh38
rs1042746015 Chr.6: 71420963-71420963 on Build GRCh38
rs1035198318 Chr.6: 71420744-71420744 on Build GRCh38
rs1035147380 Chr.6: 71420825-71420825 on Build GRCh38
rs942571283 Chr.6: 71420922-71420922 on Build GRCh38
rs912848175 Chr.6: 71420808-71420808 on Build GRCh38
rs934884752 Chr.6: 71420723-71420723 on Build GRCh38
rs947452480 Chr.6: 71420742-71420742 on Build GRCh38
rs1003805328 Chr.6: 71420811-71420811 on Build GRCh38
rs1056765754 Chr.6: 71420991-71420991 on Build GRCh38
rs941248391 Chr.6: 71420594-71420594 on Build GRCh38
rs1017119317 Chr.6: 71421029-71421029 on Build GRCh38
rs1004515839 Chr.6: 71420844-71420844 on Build GRCh38
rs115268963 Chr.6: 71420645-71420645 on Build GRCh38
rs1044418774 Chr.6: 71420824-71420824 on Build GRCh38
rs776618186 Chr.6: 71421043-71421044 on Build GRCh38
rs567153487 Chr.6: 71420776-71420776 on Build GRCh38
rs1030212021 Chr.6: 71420777-71420786 on Build GRCh38
rs966892001 Chr.6: 71420716-71420716 on Build GRCh38
rs972785548 Chr.6: 71420865-71420865 on Build GRCh38
rs115221307 Chr.6: 71420763-71420763 on Build GRCh38
rs1039082184 Chr.6: 71420923-71420923 on Build GRCh38
rs933944082 Chr.6: 71420789-71420789 on Build GRCh38
rs990559018 Chr.6: 71420833-71420833 on Build GRCh38
rs995259353 Chr.6: 71420857-71420857 on Build GRCh38
rs918521304 Chr.6: 71420781-71420781 on Build GRCh38
rs565603069 Chr.6: 71420636-71420636 on Build GRCh38
rs536014705 Chr.6: 71420806-71420806 on Build GRCh38
rs907840254 Chr.6: 71421015-71421015 on Build GRCh38
rs947481539 Chr.6: 71420818-71420818 on Build GRCh38
rs893800022 Chr.6: 71420798-71420798 on Build GRCh38
rs914713358 Chr.6: 71420815-71420815 on Build GRCh38
rs1035275010 Chr.6: 71420591-71420591 on Build GRCh38
rs538021845 Chr.6: 71420969-71420969 on Build GRCh38
rs918273734 Chr.6: 71420867-71420867 on Build GRCh38
rs1002081104 Chr.6: 71420767-71420767 on Build GRCh38
rs992863160 Chr.6: 71421009-71421009 on Build GRCh38
rs1049576968 Chr.6: 71421067-71421067 on Build GRCh38
rs1011354131 Chr.6: 71420799-71420799 on Build GRCh38
rs566586064 Chr.6: 71420937-71420937 on Build GRCh38
rs980230561 Chr.6: 71420984-71420984 on Build GRCh38
rs893907534 Chr.6: 71420944-71420944 on Build GRCh38
rs989433937 Chr.6: 71420894-71420894 on Build GRCh38
rs1028116769 Chr.6: 71420861-71420861 on Build GRCh38
rs893804373 Chr.6: 71420672-71420672 on Build GRCh38
rs559269567 Chr.6: 71421039-71421039 on Build GRCh38

View Full Product Details