Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 163309881-163310388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACRG
Entrez Gene ID
135138
Gene Name
PARK2 coregulated
Gene Aliases
GLUP, HAK005771, PARK2CRG
UniGene
Hs.25791
Gene Chromosome Location
Chr.6: 162726870-163315492 on Build GRCh38
Transcripts
NM_001080378.1 NM_001080379.1 NM_152410.2 NR_028390.1

Gene Symbol
PACRG-AS1
Entrez Gene ID
285796
Gene Name
PACRG antisense RNA 1
Gene Aliases
-
UniGene
Hs.668693
Gene Chromosome Location
Chr.6: 163309985-163324473 on Build GRCh38
Transcripts
NM_001080378.1 NM_001080379.1 NM_152410.2 NR_028390.1

SNP Detail

SNP ID Chromosome Location
rs980895705 Chr.6: 163310015-163310015 on Build GRCh38
rs748641773 Chr.6: 163310310-163310310 on Build GRCh38
rs568443253 Chr.6: 163309930-163309930 on Build GRCh38
rs1050433099 Chr.6: 163310225-163310225 on Build GRCh38
rs572044292 Chr.6: 163310304-163310304 on Build GRCh38
rs538158253 Chr.6: 163310322-163310322 on Build GRCh38
rs1014080799 Chr.6: 163310307-163310307 on Build GRCh38
rs940174210 Chr.6: 163310026-163310026 on Build GRCh38
rs558820135 Chr.6: 163309976-163309976 on Build GRCh38
rs780927838 Chr.6: 163309991-163309991 on Build GRCh38
rs540146171 Chr.6: 163310030-163310030 on Build GRCh38
rs541076282 Chr.6: 163310353-163310353 on Build GRCh38
rs189900159 Chr.6: 163310051-163310051 on Build GRCh38
rs1028870448 Chr.6: 163309919-163309919 on Build GRCh38
rs548694238 Chr.6: 163309927-163309927 on Build GRCh38
rs778571772 Chr.6: 163309948-163309948 on Build GRCh38
rs555480772 Chr.6: 163310299-163310299 on Build GRCh38
rs777025415 Chr.6: 163310191-163310191 on Build GRCh38
rs988386061 Chr.6: 163310226-163310226 on Build GRCh38
rs995970775 Chr.6: 163309918-163309918 on Build GRCh38
rs992248474 Chr.6: 163309978-163309978 on Build GRCh38
rs980438724 Chr.6: 163309985-163309985 on Build GRCh38
rs185199594 Chr.6: 163309936-163309936 on Build GRCh38
rs951935384 Chr.6: 163309921-163309921 on Build GRCh38
rs2143069 Chr.6: 163310005-163310005 on Build GRCh38
rs891572702 Chr.6: 163310301-163310302 on Build GRCh38
rs769768813 Chr.6: 163310150-163310150 on Build GRCh38
rs1024695417 Chr.6: 163309996-163309996 on Build GRCh38
rs535650254 Chr.6: 163310249-163310249 on Build GRCh38
rs778930477 Chr.6: 163310316-163310316 on Build GRCh38
rs570810587 Chr.6: 163310024-163310024 on Build GRCh38
rs970312751 Chr.6: 163310345-163310345 on Build GRCh38
rs1041245451 Chr.6: 163310069-163310069 on Build GRCh38
rs1024622293 Chr.6: 163309973-163309973 on Build GRCh38
rs944018004 Chr.6: 163310317-163310317 on Build GRCh38
rs999897052 Chr.6: 163310275-163310275 on Build GRCh38
rs914152197 Chr.6: 163310248-163310248 on Build GRCh38
rs922775936 Chr.6: 163310103-163310103 on Build GRCh38
rs150029998 Chr.6: 163310224-163310224 on Build GRCh38
rs572098745 Chr.6: 163310124-163310124 on Build GRCh38

View Full Product Details