Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143697837-143698333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BREA2
Entrez Gene ID
286076
Gene Name
breast cancer estrogen-induced apoptosis 2
Gene Aliases
-
UniGene
Hs.178095
Gene Chromosome Location
Chr.8: 143697115-143698413 on Build GRCh38
Transcripts
NR_015445.1

SNP Detail

SNP ID Chromosome Location
rs547163587 Chr.8: 143697863-143697863 on Build GRCh38
rs529076604 Chr.8: 143697913-143697913 on Build GRCh38
rs1005665695 Chr.8: 143698188-143698188 on Build GRCh38
rs114516700 Chr.8: 143697910-143697910 on Build GRCh38
rs1016601225 Chr.8: 143698214-143698214 on Build GRCh38
rs990602016 Chr.8: 143698291-143698291 on Build GRCh38
rs901019471 Chr.8: 143698132-143698132 on Build GRCh38
rs1017645974 Chr.8: 143698027-143698027 on Build GRCh38
rs181388853 Chr.8: 143697945-143697945 on Build GRCh38
rs998008999 Chr.8: 143698133-143698133 on Build GRCh38
rs540475425 Chr.8: 143697865-143697865 on Build GRCh38
rs983771207 Chr.8: 143698018-143698018 on Build GRCh38
rs557103625 Chr.8: 143698163-143698163 on Build GRCh38
rs552621412 Chr.8: 143698031-143698031 on Build GRCh38
rs917441329 Chr.8: 143698054-143698054 on Build GRCh38
rs927363212 Chr.8: 143698078-143698078 on Build GRCh38
rs556642131 Chr.8: 143698085-143698085 on Build GRCh38
rs961308481 Chr.8: 143698215-143698215 on Build GRCh38
rs531001290 Chr.8: 143697959-143697959 on Build GRCh38
rs564073602 Chr.8: 143697862-143697862 on Build GRCh38
rs548737355 Chr.8: 143697919-143697919 on Build GRCh38
rs370285451 Chr.8: 143698006-143698006 on Build GRCh38
rs889539075 Chr.8: 143698187-143698187 on Build GRCh38
rs140453971 Chr.8: 143698243-143698243 on Build GRCh38
rs575009049 Chr.8: 143698169-143698169 on Build GRCh38
rs575058639 Chr.8: 143698307-143698307 on Build GRCh38
rs568591409 Chr.8: 143698086-143698086 on Build GRCh38
rs973355229 Chr.8: 143698048-143698048 on Build GRCh38
rs1020795029 Chr.8: 143697937-143697937 on Build GRCh38
rs535052514 Chr.8: 143698064-143698064 on Build GRCh38
rs556216425 Chr.8: 143698079-143698079 on Build GRCh38
rs980237156 Chr.8: 143698264-143698264 on Build GRCh38
rs1024962693 Chr.8: 143698230-143698230 on Build GRCh38
rs994440690 Chr.8: 143698223-143698223 on Build GRCh38
rs34305381 Chr.8: 143697990-143697991 on Build GRCh38
rs570775427 Chr.8: 143698051-143698051 on Build GRCh38
rs557472460 Chr.8: 143698272-143698272 on Build GRCh38
rs770897074 Chr.8: 143698084-143698084 on Build GRCh38
rs1141722 Chr.8: 143698114-143698114 on Build GRCh38
rs528387125 Chr.8: 143697864-143697864 on Build GRCh38
rs1042674890 Chr.8: 143698126-143698126 on Build GRCh38
rs1054136188 Chr.8: 143697892-143697892 on Build GRCh38
rs547530830 Chr.8: 143698165-143698166 on Build GRCh38
rs561812882 Chr.8: 143697982-143697982 on Build GRCh38
rs950189925 Chr.8: 143698055-143698055 on Build GRCh38
rs10429285 Chr.8: 143697941-143697941 on Build GRCh38
rs572794268 Chr.8: 143698290-143698290 on Build GRCh38
rs773812517 Chr.8: 143698112-143698112 on Build GRCh38

View Full Product Details