Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134546448-134546954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR424
Entrez Gene ID
494336
Gene Name
microRNA 424
Gene Aliases
MIR322, MIRN424, hsa-mir-424, miRNA424, mir-424
UniGene
Gene Chromosome Location
Chr.X: 134546614-134546711 on Build GRCh38
Transcripts
NR_029946.1 NR_024607.1

Gene Symbol
MIR503HG
Entrez Gene ID
84848
Gene Name
MIR503 host gene
Gene Aliases
H19X, MIR503HG2
UniGene
Hs.416379
Gene Chromosome Location
Chr.X: 134543377-134546630 on Build GRCh38
Transcripts
NR_029946.1 NR_024607.1

SNP Detail

SNP ID Chromosome Location
rs776404815 Chr.X: 134546731-134546731 on Build GRCh38
rs768539946 Chr.X: 134546567-134546567 on Build GRCh38
rs763459662 Chr.X: 134546746-134546746 on Build GRCh38
rs992589885 Chr.X: 134546845-134546845 on Build GRCh38
rs1035831067 Chr.X: 134546865-134546865 on Build GRCh38
rs772803724 Chr.X: 134546596-134546596 on Build GRCh38
rs763446168 Chr.X: 134546781-134546781 on Build GRCh38
rs935998113 Chr.X: 134546911-134546911 on Build GRCh38
rs748070284 Chr.X: 134546711-134546711 on Build GRCh38
rs925950359 Chr.X: 134546530-134546530 on Build GRCh38
rs761637957 Chr.X: 134546866-134546866 on Build GRCh38
rs757309 Chr.X: 134546517-134546517 on Build GRCh38
rs760264608 Chr.X: 134546724-134546724 on Build GRCh38
rs745987032 Chr.X: 134546654-134546654 on Build GRCh38
rs1023404795 Chr.X: 134546719-134546719 on Build GRCh38
rs764389968 Chr.X: 134546749-134546749 on Build GRCh38
rs757452371 Chr.X: 134546641-134546641 on Build GRCh38
rs753245359 Chr.X: 134546610-134546610 on Build GRCh38
rs1044969556 Chr.X: 134546507-134546531 on Build GRCh38
rs917150206 Chr.X: 134546494-134546494 on Build GRCh38
rs761643060 Chr.X: 134546574-134546574 on Build GRCh38
rs764836839 Chr.X: 134546741-134546741 on Build GRCh38
rs888095819 Chr.X: 134546701-134546701 on Build GRCh38
rs774239229 Chr.X: 134546571-134546571 on Build GRCh38
rs1050933493 Chr.X: 134546554-134546554 on Build GRCh38
rs769840433 Chr.X: 134546670-134546670 on Build GRCh38
rs781139344 Chr.X: 134546585-134546590 on Build GRCh38
rs778134590 Chr.X: 134546635-134546635 on Build GRCh38
rs373943888 Chr.X: 134546604-134546604 on Build GRCh38
rs775869877 Chr.X: 134546643-134546643 on Build GRCh38
rs778877675 Chr.X: 134546710-134546710 on Build GRCh38
rs186029845 Chr.X: 134546897-134546897 on Build GRCh38
rs967229893 Chr.X: 134546884-134546884 on Build GRCh38
rs913230394 Chr.X: 134546508-134546508 on Build GRCh38
rs774385095 Chr.X: 134546846-134546846 on Build GRCh38
rs749246414 Chr.X: 134546564-134546564 on Build GRCh38
rs370636915 Chr.X: 134546638-134546638 on Build GRCh38
rs180713552 Chr.X: 134546896-134546896 on Build GRCh38
rs758758217 Chr.X: 134546628-134546628 on Build GRCh38
rs765816463 Chr.X: 134546605-134546605 on Build GRCh38
rs1009051285 Chr.X: 134546570-134546570 on Build GRCh38
rs368699680 Chr.X: 134546738-134546738 on Build GRCh38
rs993751623 Chr.X: 134546607-134546607 on Build GRCh38
rs749320241 Chr.X: 134546630-134546630 on Build GRCh38
rs746311873 Chr.X: 134546583-134546583 on Build GRCh38
rs751585871 Chr.X: 134546789-134546789 on Build GRCh38
rs767327919 Chr.X: 134546589-134546589 on Build GRCh38
rs766771674 Chr.X: 134546833-134546833 on Build GRCh38
rs757120140 Chr.X: 134546578-134546579 on Build GRCh38
rs768891044 Chr.X: 134546730-134546730 on Build GRCh38
rs901093510 Chr.X: 134546630-134546630 on Build GRCh38
rs376261811 Chr.X: 134546608-134546608 on Build GRCh38
rs753263457 Chr.X: 134546745-134546745 on Build GRCh38
rs371045221 Chr.X: 134546541-134546541 on Build GRCh38
rs771766225 Chr.X: 134546715-134546715 on Build GRCh38
rs781378501 Chr.X: 134546582-134546582 on Build GRCh38
rs374618265 Chr.X: 134546717-134546717 on Build GRCh38
rs759089735 Chr.X: 134546609-134546609 on Build GRCh38
rs945977906 Chr.X: 134546500-134546500 on Build GRCh38

View Full Product Details