Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39375312-39375864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
UniGene
Hs.369765
Gene Chromosome Location
Chr.8: 39314663-39417226 on Build GRCh38
Transcripts
NR_001448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036068 c.392G>A p.R131Q Chr.8: 39375839-39375839 on Build GRCh38
1624006 c.317G>C p.C106S Chr.8: 39375764-39375764 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891593740 Chr.8: 39375674-39375674 on Build GRCh38
rs1045605539 Chr.8: 39375639-39375639 on Build GRCh38
rs996275411 Chr.8: 39375722-39375722 on Build GRCh38
rs924481706 Chr.8: 39375739-39375739 on Build GRCh38
rs185243607 Chr.8: 39375839-39375839 on Build GRCh38
rs936041309 Chr.8: 39375626-39375626 on Build GRCh38
rs558425578 Chr.8: 39375630-39375630 on Build GRCh38
rs751771332 Chr.8: 39375590-39375590 on Build GRCh38
rs951786973 Chr.8: 39375338-39375338 on Build GRCh38
rs181749559 Chr.8: 39375440-39375440 on Build GRCh38
rs190468651 Chr.8: 39375691-39375691 on Build GRCh38
rs867629044 Chr.8: 39375732-39375732 on Build GRCh38
rs1054257563 Chr.8: 39375831-39375831 on Build GRCh38
rs767942931 Chr.8: 39375611-39375611 on Build GRCh38
rs766620432 Chr.8: 39375504-39375504 on Build GRCh38
rs577973366 Chr.8: 39375499-39375499 on Build GRCh38
rs185541867 Chr.8: 39375642-39375642 on Build GRCh38
rs190051603 Chr.8: 39375340-39375340 on Build GRCh38
rs181998274 Chr.8: 39375702-39375702 on Build GRCh38
rs1005890376 Chr.8: 39375822-39375822 on Build GRCh38
rs374503621 Chr.8: 39375606-39375606 on Build GRCh38
rs984268715 Chr.8: 39375372-39375372 on Build GRCh38
rs901755800 Chr.8: 39375713-39375713 on Build GRCh38
rs778152351 Chr.8: 39375781-39375781 on Build GRCh38
rs934502576 Chr.8: 39375797-39375797 on Build GRCh38
rs756622269 Chr.8: 39375753-39375753 on Build GRCh38
rs950134355 Chr.8: 39375526-39375526 on Build GRCh38
rs949999458 Chr.8: 39375433-39375433 on Build GRCh38
rs910032438 Chr.8: 39375418-39375418 on Build GRCh38
rs945952354 Chr.8: 39375687-39375687 on Build GRCh38

View Full Product Details