Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48101611-48101987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKINT1L
Entrez Gene ID
391037
Gene Name
Skint1 like (pseudogene)
Gene Aliases
BTN12, SKINT1, SKINTL, SKINTP
UniGene
Hs.585393
Gene Chromosome Location
Chr.1: 48101715-48182428 on Build GRCh38
Transcripts
NR_026749.2

SNP Detail

SNP ID Chromosome Location
rs578166557 Chr.1: 48101779-48101779 on Build GRCh38
rs146043916 Chr.1: 48101957-48101957 on Build GRCh38
rs1056000883 Chr.1: 48101666-48101666 on Build GRCh38
rs1029701725 Chr.1: 48101740-48101740 on Build GRCh38
rs138061816 Chr.1: 48101825-48101825 on Build GRCh38
rs991273687 Chr.1: 48101913-48101913 on Build GRCh38
rs17468958 Chr.1: 48101790-48101790 on Build GRCh38
rs534395245 Chr.1: 48101944-48101944 on Build GRCh38
rs1036297051 Chr.1: 48101706-48101706 on Build GRCh38
rs750631178 Chr.1: 48101729-48101729 on Build GRCh38
rs1006976352 Chr.1: 48101747-48101747 on Build GRCh38
rs1004576610 Chr.1: 48101910-48101910 on Build GRCh38
rs924766954 Chr.1: 48101758-48101758 on Build GRCh38
rs928293289 Chr.1: 48101904-48101904 on Build GRCh38
rs533097807 Chr.1: 48101773-48101773 on Build GRCh38
rs114055058 Chr.1: 48101903-48101903 on Build GRCh38
rs150354030 Chr.1: 48101698-48101698 on Build GRCh38
rs567387229 Chr.1: 48101932-48101932 on Build GRCh38
rs996528911 Chr.1: 48101948-48101948 on Build GRCh38
rs114986270 Chr.1: 48101658-48101658 on Build GRCh38
rs891173766 Chr.1: 48101743-48101743 on Build GRCh38
rs955081303 Chr.1: 48101718-48101718 on Build GRCh38
rs898193900 Chr.1: 48101945-48101945 on Build GRCh38
rs994154437 Chr.1: 48101731-48101731 on Build GRCh38
rs185707725 Chr.1: 48101772-48101772 on Build GRCh38
rs757012465 Chr.1: 48101799-48101799 on Build GRCh38
rs909012297 Chr.1: 48101929-48101929 on Build GRCh38
rs528563334 Chr.1: 48101672-48101672 on Build GRCh38
rs1020557660 Chr.1: 48101796-48101796 on Build GRCh38
rs142535987 Chr.1: 48101908-48101908 on Build GRCh38
rs1056709105 Chr.1: 48101907-48101907 on Build GRCh38
rs1044345945 Chr.1: 48101795-48101795 on Build GRCh38
rs956522039 Chr.1: 48101762-48101762 on Build GRCh38
rs887621315 Chr.1: 48101684-48101684 on Build GRCh38

View Full Product Details