Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202183220-202183764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRVP
Entrez Gene ID
148713
Gene Name
protein tyrosine phosphatase, receptor type V, pseudogene
Gene Aliases
ESP, OST-PTP, PTPRV
UniGene
Hs.523870
Gene Chromosome Location
Chr.1: 202168051-202189449 on Build GRCh38
Transcripts
NR_002930.2

SNP Detail

SNP ID Chromosome Location
rs766691081 Chr.1: 202183679-202183679 on Build GRCh38
rs72746673 Chr.1: 202183519-202183519 on Build GRCh38
rs916000721 Chr.1: 202183722-202183722 on Build GRCh38
rs868175479 Chr.1: 202183728-202183728 on Build GRCh38
rs899023757 Chr.1: 202183532-202183532 on Build GRCh38
rs1055929226 Chr.1: 202183376-202183384 on Build GRCh38
rs764948450 Chr.1: 202183366-202183366 on Build GRCh38
rs71635573 Chr.1: 202183497-202183497 on Build GRCh38
rs906463714 Chr.1: 202183445-202183445 on Build GRCh38
rs968045989 Chr.1: 202183643-202183643 on Build GRCh38
rs761391046 Chr.1: 202183272-202183272 on Build GRCh38
rs2993441 Chr.1: 202183731-202183731 on Build GRCh38
rs537107762 Chr.1: 202183705-202183705 on Build GRCh38
rs926096201 Chr.1: 202183667-202183667 on Build GRCh38
rs577418743 Chr.1: 202183651-202183651 on Build GRCh38
rs1019168474 Chr.1: 202183587-202183587 on Build GRCh38
rs957559933 Chr.1: 202183699-202183699 on Build GRCh38
rs1028730827 Chr.1: 202183535-202183535 on Build GRCh38
rs950041096 Chr.1: 202183386-202183386 on Build GRCh38
rs946196194 Chr.1: 202183292-202183292 on Build GRCh38
rs1046307711 Chr.1: 202183432-202183432 on Build GRCh38
rs765400358 Chr.1: 202183703-202183703 on Build GRCh38
rs915907098 Chr.1: 202183378-202183386 on Build GRCh38
rs546400118 Chr.1: 202183693-202183693 on Build GRCh38
rs1008518708 Chr.1: 202183584-202183584 on Build GRCh38
rs146012254 Chr.1: 202183365-202183365 on Build GRCh38
rs534839608 Chr.1: 202183526-202183526 on Build GRCh38
rs1004692449 Chr.1: 202183453-202183453 on Build GRCh38
rs987673999 Chr.1: 202183286-202183286 on Build GRCh38
rs538828884 Chr.1: 202183348-202183348 on Build GRCh38
rs751251453 Chr.1: 202183685-202183685 on Build GRCh38
rs28484948 Chr.1: 202183498-202183498 on Build GRCh38
rs185530736 Chr.1: 202183602-202183602 on Build GRCh38
rs754081895 Chr.1: 202183686-202183686 on Build GRCh38
rs912136321 Chr.1: 202183290-202183290 on Build GRCh38
rs72746675 Chr.1: 202183533-202183533 on Build GRCh38
rs953085814 Chr.1: 202183270-202183270 on Build GRCh38
rs978010281 Chr.1: 202183312-202183312 on Build GRCh38
rs144103092 Chr.1: 202183268-202183268 on Build GRCh38
rs977730263 Chr.1: 202183644-202183644 on Build GRCh38
rs936057477 Chr.1: 202183360-202183360 on Build GRCh38
rs780244919 Chr.1: 202183658-202183658 on Build GRCh38
rs569872726 Chr.1: 202183581-202183581 on Build GRCh38
rs557697456 Chr.1: 202183635-202183635 on Build GRCh38

View Full Product Details