Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21102819-21103382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCRP2
Entrez Gene ID
400892
Gene Name
breakpoint cluster region pseudogene 2
Gene Aliases
BCR-2, BCR2, BCRL2
UniGene
Hs.745077
Gene Chromosome Location
Chr.22: 21103016-21122286 on Build GRCh38
Transcripts
NR_037566.1

SNP Detail

SNP ID Chromosome Location
rs890593363 Chr.22: 21103314-21103314 on Build GRCh38
rs892685526 Chr.22: 21103199-21103199 on Build GRCh38
rs1044179433 Chr.22: 21102931-21102931 on Build GRCh38
rs1037765227 Chr.22: 21103284-21103284 on Build GRCh38
rs905462493 Chr.22: 21102908-21102908 on Build GRCh38
rs1016476652 Chr.22: 21103023-21103023 on Build GRCh38
rs771614262 Chr.22: 21103324-21103324 on Build GRCh38
rs556294027 Chr.22: 21103275-21103275 on Build GRCh38
rs568648825 Chr.22: 21102878-21102878 on Build GRCh38
rs1028402135 Chr.22: 21103277-21103277 on Build GRCh38
rs184556148 Chr.22: 21102927-21102927 on Build GRCh38
rs538294264 Chr.22: 21102858-21102858 on Build GRCh38
rs939668384 Chr.22: 21103026-21103026 on Build GRCh38
rs552243862 Chr.22: 21102861-21102861 on Build GRCh38
rs987470284 Chr.22: 21102868-21102868 on Build GRCh38
rs1048325067 Chr.22: 21102853-21102853 on Build GRCh38
rs979971181 Chr.22: 21103190-21103190 on Build GRCh38
rs1031431753 Chr.22: 21103098-21103098 on Build GRCh38
rs908238518 Chr.22: 21102862-21102862 on Build GRCh38
rs938441236 Chr.22: 21103222-21103222 on Build GRCh38
rs961253172 Chr.22: 21103349-21103349 on Build GRCh38
rs1045611572 Chr.22: 21102897-21102897 on Build GRCh38
rs914462483 Chr.22: 21103120-21103120 on Build GRCh38
rs1020011799 Chr.22: 21103244-21103244 on Build GRCh38
rs921985201 Chr.22: 21102842-21102842 on Build GRCh38
rs989139764 Chr.22: 21103117-21103117 on Build GRCh38
rs921122030 Chr.22: 21103015-21103015 on Build GRCh38
rs433457 Chr.22: 21103357-21103357 on Build GRCh38
rs947281563 Chr.22: 21103142-21103142 on Build GRCh38
rs575772815 Chr.22: 21103231-21103231 on Build GRCh38
rs940333743 Chr.22: 21103267-21103267 on Build GRCh38
rs566420615 Chr.22: 21102845-21102845 on Build GRCh38
rs547053322 Chr.22: 21102918-21102918 on Build GRCh38
rs908456347 Chr.22: 21103024-21103024 on Build GRCh38
rs927093594 Chr.22: 21103193-21103193 on Build GRCh38
rs905386768 Chr.22: 21103148-21103148 on Build GRCh38
rs539495478 Chr.22: 21103253-21103253 on Build GRCh38
rs1009537536 Chr.22: 21103201-21103202 on Build GRCh38
rs1056854951 Chr.22: 21103234-21103234 on Build GRCh38
rs188240558 Chr.22: 21102994-21102994 on Build GRCh38
rs914410240 Chr.22: 21102916-21102916 on Build GRCh38
rs180720161 Chr.22: 21102998-21102998 on Build GRCh38
rs983914866 Chr.22: 21103305-21103305 on Build GRCh38
rs570481279 Chr.22: 21103068-21103068 on Build GRCh38
rs375458418 Chr.22: 21103022-21103022 on Build GRCh38
rs545100332 Chr.22: 21103076-21103076 on Build GRCh38
rs189423621 Chr.22: 21103017-21103017 on Build GRCh38
rs1009413611 Chr.22: 21103327-21103327 on Build GRCh38
rs1053011415 Chr.22: 21102883-21102883 on Build GRCh38
rs992524149 Chr.22: 21103353-21103353 on Build GRCh38
rs1029144540 Chr.22: 21103308-21103308 on Build GRCh38
rs369605042 Chr.22: 21103030-21103030 on Build GRCh38
rs1056939235 Chr.22: 21102877-21102877 on Build GRCh38
rs1019703478 Chr.22: 21102995-21102995 on Build GRCh38
rs939657242 Chr.22: 21102876-21102876 on Build GRCh38
rs527613596 Chr.22: 21102893-21102893 on Build GRCh38
rs956709930 Chr.22: 21103109-21103109 on Build GRCh38
rs954814265 Chr.22: 21103088-21103088 on Build GRCh38
rs569782965 Chr.22: 21103334-21103334 on Build GRCh38
rs947165668 Chr.22: 21102930-21102930 on Build GRCh38
rs996335030 Chr.22: 21103074-21103074 on Build GRCh38
rs923944534 Chr.22: 21102871-21102871 on Build GRCh38
rs1009485224 Chr.22: 21102991-21102991 on Build GRCh38
rs899463539 Chr.22: 21103301-21103301 on Build GRCh38
rs373217305 Chr.22: 21103302-21103302 on Build GRCh38
rs797011403 Chr.22: 21103002-21103002 on Build GRCh38
rs1013760041 Chr.22: 21102881-21102881 on Build GRCh38
rs554316981 Chr.22: 21102986-21102986 on Build GRCh38
rs781758657 Chr.22: 21103157-21103157 on Build GRCh38
rs61744393 Chr.22: 21103096-21103096 on Build GRCh38
rs183569939 Chr.22: 21103003-21103003 on Build GRCh38

View Full Product Details