Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150000736-150001306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAET1K
Entrez Gene ID
646024
Gene Name
retinoic acid early transcript 1K pseudogene
Gene Aliases
-
UniGene
Hs.560537
Gene Chromosome Location
Chr.6: 149998019-150005144 on Build GRCh38
Transcripts
NR_024045.1

SNP Detail

SNP ID Chromosome Location
rs1028222141 Chr.6: 150000790-150000790 on Build GRCh38
rs371170312 Chr.6: 150000855-150000855 on Build GRCh38
rs554347667 Chr.6: 150000929-150000929 on Build GRCh38
rs796804657 Chr.6: 150000770-150000771 on Build GRCh38
rs149456419 Chr.6: 150001202-150001202 on Build GRCh38
rs900862170 Chr.6: 150000771-150000772 on Build GRCh38
rs761266427 Chr.6: 150000975-150000975 on Build GRCh38
rs989484806 Chr.6: 150001141-150001142 on Build GRCh38
rs142842862 Chr.6: 150000771-150000771 on Build GRCh38
rs543240667 Chr.6: 150001131-150001131 on Build GRCh38
rs538621223 Chr.6: 150001097-150001097 on Build GRCh38
rs748293377 Chr.6: 150000910-150000910 on Build GRCh38
rs572746261 Chr.6: 150001168-150001168 on Build GRCh38
rs757952188 Chr.6: 150000810-150000810 on Build GRCh38
rs996457646 Chr.6: 150000789-150000789 on Build GRCh38
rs1033433986 Chr.6: 150001121-150001121 on Build GRCh38
rs140862025 Chr.6: 150000803-150000803 on Build GRCh38
rs565563303 Chr.6: 150000959-150000959 on Build GRCh38
rs186986022 Chr.6: 150001071-150001071 on Build GRCh38
rs9766403 Chr.6: 150000779-150000779 on Build GRCh38
rs1017781730 Chr.6: 150000768-150000768 on Build GRCh38
rs528510174 Chr.6: 150001142-150001142 on Build GRCh38
rs34905964 Chr.6: 150000770-150000771 on Build GRCh38
rs138648792 Chr.6: 150000830-150000830 on Build GRCh38
rs187802343 Chr.6: 150000917-150000917 on Build GRCh38
rs9397541 Chr.6: 150001075-150001075 on Build GRCh38
rs373392705 Chr.6: 150001001-150001001 on Build GRCh38
rs561888205 Chr.6: 150001226-150001226 on Build GRCh38
rs562875799 Chr.6: 150000940-150000940 on Build GRCh38
rs139234402 Chr.6: 150000791-150000791 on Build GRCh38
rs71694514 Chr.6: 150000787-150000788 on Build GRCh38
rs771509887 Chr.6: 150000997-150000997 on Build GRCh38
rs563357588 Chr.6: 150001269-150001269 on Build GRCh38
rs141667997 Chr.6: 150000941-150000941 on Build GRCh38
rs368056586 Chr.6: 150000762-150000762 on Build GRCh38
rs996508468 Chr.6: 150001002-150001002 on Build GRCh38
rs397744426 Chr.6: 150000790-150000791 on Build GRCh38
rs961198436 Chr.6: 150000812-150000812 on Build GRCh38
rs888088257 Chr.6: 150001066-150001066 on Build GRCh38
rs116563274 Chr.6: 150001115-150001115 on Build GRCh38
rs369630228 Chr.6: 150000988-150000988 on Build GRCh38
rs574277546 Chr.6: 150001119-150001119 on Build GRCh38
rs900211707 Chr.6: 150000895-150000895 on Build GRCh38
rs558279746 Chr.6: 150001108-150001108 on Build GRCh38
rs140535837 Chr.6: 150000769-150000770 on Build GRCh38
rs980081257 Chr.6: 150000927-150000927 on Build GRCh38
rs373503833 Chr.6: 150000768-150000769 on Build GRCh38
rs557543545 Chr.6: 150000799-150000799 on Build GRCh38
rs369502300 Chr.6: 150001278-150001278 on Build GRCh38
rs115976795 Chr.6: 150001147-150001147 on Build GRCh38
rs888129754 Chr.6: 150001227-150001227 on Build GRCh38
rs117062721 Chr.6: 150000769-150000769 on Build GRCh38
rs569699656 Chr.6: 150001096-150001096 on Build GRCh38
rs144794258 Chr.6: 150000827-150000827 on Build GRCh38
rs753277064 Chr.6: 150001054-150001054 on Build GRCh38
rs556616245 Chr.6: 150001134-150001134 on Build GRCh38
rs112214945 Chr.6: 150001074-150001074 on Build GRCh38
rs533758935 Chr.6: 150000804-150000806 on Build GRCh38
rs542729383 Chr.6: 150000938-150000938 on Build GRCh38
rs374734662 Chr.6: 150000871-150000871 on Build GRCh38
rs35141392 Chr.6: 150001251-150001252 on Build GRCh38
rs182660557 Chr.6: 150001070-150001070 on Build GRCh38
rs11752135 Chr.6: 150001272-150001272 on Build GRCh38
rs372659039 Chr.6: 150000805-150000807 on Build GRCh38
rs527998892 Chr.6: 150001005-150001005 on Build GRCh38
rs957353452 Chr.6: 150001123-150001123 on Build GRCh38
rs567027287 Chr.6: 150001135-150001135 on Build GRCh38
rs1040802022 Chr.6: 150000998-150000998 on Build GRCh38
rs1015488912 Chr.6: 150001082-150001082 on Build GRCh38
rs765488538 Chr.6: 150001181-150001181 on Build GRCh38
rs565920331 Chr.6: 150001111-150001111 on Build GRCh38
rs9384023 Chr.6: 150001245-150001245 on Build GRCh38
rs941006465 Chr.6: 150001228-150001228 on Build GRCh38
rs1024417096 Chr.6: 150001105-150001105 on Build GRCh38
rs922392782 Chr.6: 150001183-150001183 on Build GRCh38
rs143846903 Chr.6: 150001204-150001204 on Build GRCh38
rs538286369 Chr.6: 150000806-150000806 on Build GRCh38
rs554308951 Chr.6: 150001118-150001118 on Build GRCh38
rs547962256 Chr.6: 150001042-150001042 on Build GRCh38
rs191575720 Chr.6: 150000931-150000931 on Build GRCh38

View Full Product Details