Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108464838-108465280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBPF6
Entrez Gene ID
653149
Gene Name
neuroblastoma breakpoint family member 6
Gene Aliases
-
UniGene
Hs.712226
Gene Chromosome Location
Chr.1: 108421496-108471903 on Build GRCh38
Transcripts
NM_001143987.1 NM_001143988.1 XM_005271144.3 XM_011542012.2 XM_011542014.2 XM_017002148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427604 c.1497T>A p.I499I Chr.1: 108465261-108465261 on Build GRCh38
4602322 c.1463G>A p.G488E Chr.1: 108465227-108465227 on Build GRCh38
6354071 c.1456T>G p.W486G Chr.1: 108465220-108465220 on Build GRCh38
6502573 c.1508A>T p.Q503L Chr.1: 108465272-108465272 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577081160 Chr.1: 108465027-108465027 on Build GRCh38
rs921601678 Chr.1: 108465171-108465172 on Build GRCh38
rs758588007 Chr.1: 108465196-108465196 on Build GRCh38
rs200044795 Chr.1: 108465133-108465133 on Build GRCh38
rs781645147 Chr.1: 108465245-108465245 on Build GRCh38
rs201232305 Chr.1: 108465200-108465200 on Build GRCh38
rs548531485 Chr.1: 108465259-108465259 on Build GRCh38
rs753000127 Chr.1: 108465195-108465195 on Build GRCh38
rs1020378610 Chr.1: 108465003-108465003 on Build GRCh38
rs202116448 Chr.1: 108465201-108465201 on Build GRCh38
rs2260439 Chr.1: 108465255-108465255 on Build GRCh38
rs202245430 Chr.1: 108465227-108465227 on Build GRCh38
rs944639014 Chr.1: 108465039-108465039 on Build GRCh38
rs531923017 Chr.1: 108465244-108465244 on Build GRCh38
rs796266881 Chr.1: 108464859-108464859 on Build GRCh38
rs966557207 Chr.1: 108465022-108465022 on Build GRCh38
rs990674127 Chr.1: 108464984-108464984 on Build GRCh38
rs974646703 Chr.1: 108465075-108465075 on Build GRCh38

View Full Product Details