Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80025508-80025680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX22
Entrez Gene ID
50945
Gene Name
T-box 22
Gene Aliases
ABERS, CLPA, CPX, TBXX, dJ795G23.1
UniGene
Hs.374253
Gene Chromosome Location
Chr.X: 80014730-80035828 on Build GRCh38
Transcripts
NM_001109878.1 NM_001109879.1 NM_001303475.1 NM_016954.2 XM_005262136.2 XM_006724657.3 XM_011530972.1

SNP Detail

SNP ID Chromosome Location
rs753047555 Chr.X: 80025652-80025652 on Build GRCh38
rs796291421 Chr.X: 80025565-80025565 on Build GRCh38
rs765680258 Chr.X: 80025632-80025632 on Build GRCh38
rs771150368 Chr.X: 80025584-80025584 on Build GRCh38
rs760626449 Chr.X: 80025559-80025559 on Build GRCh38
rs781287118 Chr.X: 80025593-80025594 on Build GRCh38
rs200060292 Chr.X: 80025598-80025598 on Build GRCh38
rs773515696 Chr.X: 80025553-80025553 on Build GRCh38
rs1005546228 Chr.X: 80025589-80025589 on Build GRCh38
rs760190777 Chr.X: 80025613-80025613 on Build GRCh38

View Full Product Details