Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69937791-69937953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MITF
Entrez Gene ID
4286
Gene Name
melanogenesis associated transcription factor
Gene Aliases
CMM8, MI, WS2, WS2A, bHLHe32
UniGene
Hs.166017
Gene Chromosome Location
Chr.3: 69739435-69968337 on Build GRCh38
Transcripts
NM_000248.3 NM_001184967.1 NM_001184968.1 NM_006722.2 NM_198158.2 NM_198159.2 NM_198177.2 NM_198178.2 XM_005264754.1 XM_005264755.3 XM_006713164.2 XM_011533722.2 XM_011533723.2 XM_011533725.1 XM_011533726.1 XM_017006444.1 XM_017006445.1 XM_017006446.1 XM_017006447.1 XM_017006448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753381 c.366C>G p.L122L Chr.3: 69937836-69937836 on Build GRCh38
3824605 c.403C>T p.R135W Chr.3: 69937873-69937873 on Build GRCh38
1753378 c.369C>G p.L123L Chr.3: 69937836-69937836 on Build GRCh38
6975230 c.133G>T p.A45S Chr.3: 69937921-69937921 on Build GRCh38
3824603 c.358C>T p.R120W Chr.3: 69937873-69937873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763327055 Chr.3: 69937817-69937817 on Build GRCh38
rs921777463 Chr.3: 69937871-69937871 on Build GRCh38
rs750608171 Chr.3: 69937874-69937874 on Build GRCh38
rs140663277 Chr.3: 69937833-69937833 on Build GRCh38
rs371031432 Chr.3: 69937882-69937882 on Build GRCh38
rs1040660099 Chr.3: 69937824-69937824 on Build GRCh38
rs201297175 Chr.3: 69937861-69937861 on Build GRCh38
rs764892628 Chr.3: 69937901-69937901 on Build GRCh38
rs758308546 Chr.3: 69937909-69937909 on Build GRCh38
rs994568417 Chr.3: 69937837-69937837 on Build GRCh38
rs752712909 Chr.3: 69937902-69937902 on Build GRCh38
rs373819498 Chr.3: 69937836-69937836 on Build GRCh38
rs776083519 Chr.3: 69937815-69937815 on Build GRCh38
rs200583343 Chr.3: 69937873-69937873 on Build GRCh38
rs777666331 Chr.3: 69937917-69937917 on Build GRCh38

View Full Product Details