Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10090269-10090429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Gene Symbol
FANCD2OS
Entrez Gene ID
115795
Gene Name
FANCD2 opposite strand
Gene Aliases
C3orf24
UniGene
Hs.190413
Gene Chromosome Location
Chr.3: 10081320-10108603 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207162 c.3714G>A p.M1238I Chr.3: 10090322-10090322 on Build GRCh38
5963290 c.3777+5G>C p.? Chr.3: 10090390-10090390 on Build GRCh38
4975872 c.3707G>T p.R1236L Chr.3: 10090315-10090315 on Build GRCh38
1417256 c.3738_3739insA p.I1249fs*2 Chr.3: 10090346-10090347 on Build GRCh38
4991808 c.3756C>A p.G1252G Chr.3: 10090364-10090364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757808149 Chr.3: 10090356-10090356 on Build GRCh38
rs771721585 Chr.3: 10090386-10090389 on Build GRCh38
rs568515612 Chr.3: 10090359-10090359 on Build GRCh38
rs750130327 Chr.3: 10090343-10090343 on Build GRCh38
rs949649696 Chr.3: 10090320-10090320 on Build GRCh38
rs368999017 Chr.3: 10090390-10090390 on Build GRCh38
rs781344292 Chr.3: 10090362-10090362 on Build GRCh38
rs769903320 Chr.3: 10090296-10090296 on Build GRCh38
rs774421283 Chr.3: 10090398-10090398 on Build GRCh38
rs779242727 Chr.3: 10090379-10090379 on Build GRCh38
rs761089162 Chr.3: 10090321-10090321 on Build GRCh38
rs764775864 Chr.3: 10090342-10090342 on Build GRCh38
rs374940277 Chr.3: 10090378-10090378 on Build GRCh38
rs1047237177 Chr.3: 10090393-10090393 on Build GRCh38
rs775831214 Chr.3: 10090319-10090319 on Build GRCh38
rs368684162 Chr.3: 10090376-10090376 on Build GRCh38
rs994778998 Chr.3: 10090380-10090380 on Build GRCh38
rs121917786 Chr.3: 10090315-10090315 on Build GRCh38
rs747942447 Chr.3: 10090366-10090366 on Build GRCh38
rs773540135 Chr.3: 10090297-10090297 on Build GRCh38
rs771078251 Chr.3: 10090314-10090314 on Build GRCh38
rs760054945 Chr.3: 10090316-10090316 on Build GRCh38
rs767806932 Chr.3: 10090318-10090318 on Build GRCh38
rs762915519 Chr.3: 10090300-10090300 on Build GRCh38
rs770863196 Chr.3: 10090385-10090385 on Build GRCh38
rs748559266 Chr.3: 10090292-10090292 on Build GRCh38

View Full Product Details