Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56166488-56166644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAG1
Entrez Gene ID
5324
Gene Name
PLAG1 zinc finger
Gene Aliases
PSA, SGPA, ZNF912
UniGene
Hs.14968
Gene Chromosome Location
Chr.8: 56160909-56211327 on Build GRCh38
Transcripts
NM_001114634.1 NM_001114635.1 NM_002655.2 XM_011517544.2 XM_017013576.1 XM_017013577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884718 c.1116A>T p.Q372H Chr.8: 56166630-56166630 on Build GRCh38
237976 c.1230C>T p.P410P Chr.8: 56166516-56166516 on Build GRCh38
3925424 c.1107T>C p.S369S Chr.8: 56166639-56166639 on Build GRCh38
95020 c.1213A>G p.I405V Chr.8: 56166533-56166533 on Build GRCh38
5597380 c.1258C>T p.Q420* Chr.8: 56166488-56166488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004492262 Chr.8: 56166554-56166554 on Build GRCh38
rs768984120 Chr.8: 56166528-56166528 on Build GRCh38
rs944845643 Chr.8: 56166519-56166519 on Build GRCh38
rs762239914 Chr.8: 56166537-56166537 on Build GRCh38
rs776708502 Chr.8: 56166533-56166533 on Build GRCh38
rs907471752 Chr.8: 56166553-56166553 on Build GRCh38
rs267601956 Chr.8: 56166590-56166590 on Build GRCh38
rs763407619 Chr.8: 56166602-56166602 on Build GRCh38
rs765459935 Chr.8: 56166584-56166584 on Build GRCh38
rs1015758225 Chr.8: 56166607-56166609 on Build GRCh38
rs141407863 Chr.8: 56166518-56166518 on Build GRCh38
rs376111654 Chr.8: 56166618-56166618 on Build GRCh38
rs138135087 Chr.8: 56166595-56166595 on Build GRCh38
rs745720729 Chr.8: 56166563-56166565 on Build GRCh38
rs375925227 Chr.8: 56166513-56166513 on Build GRCh38
rs773728791 Chr.8: 56166592-56166592 on Build GRCh38
rs767203369 Chr.8: 56166620-56166620 on Build GRCh38
rs751117942 Chr.8: 56166612-56166612 on Build GRCh38
rs766840293 Chr.8: 56166607-56166607 on Build GRCh38

View Full Product Details