Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36284375-36284536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH9
Entrez Gene ID
4627
Gene Name
myosin, heavy chain 9, non-muscle
Gene Aliases
BDPLT6, DFNA17, EPSTS, FTNS, MHA, NMHC-II-A, NMMHC-IIA, NMMHCA
UniGene
Hs.474751
Gene Chromosome Location
Chr.22: 36281277-36388067 on Build GRCh38
Transcripts
NM_002473.5 XM_011530197.2 XM_017028803.1 XM_017028804.1 XM_017028805.1 XM_017028806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7351951 c.5484-1G>A p.? Chr.22: 36284512-36284512 on Build GRCh38
7209686 c.5531T>G p.L1844R Chr.22: 36284464-36284464 on Build GRCh38
302709 c.5515C>T p.R1839W Chr.22: 36284480-36284480 on Build GRCh38
7164990 c.5564G>A p.R1855Q Chr.22: 36284431-36284431 on Build GRCh38
5471478 c.5561A>G p.E1854G Chr.22: 36284434-36284434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767694496 Chr.22: 36284466-36284466 on Build GRCh38
rs774510520 Chr.22: 36284463-36284463 on Build GRCh38
rs966415372 Chr.22: 36284506-36284506 on Build GRCh38
rs758034745 Chr.22: 36284423-36284423 on Build GRCh38
rs766271245 Chr.22: 36284487-36284487 on Build GRCh38
rs754771834 Chr.22: 36284505-36284505 on Build GRCh38
rs752580965 Chr.22: 36284516-36284516 on Build GRCh38
rs80338834 Chr.22: 36284474-36284474 on Build GRCh38
rs368994883 Chr.22: 36284450-36284450 on Build GRCh38
rs775990931 Chr.22: 36284436-36284436 on Build GRCh38
rs200409945 Chr.22: 36284424-36284424 on Build GRCh38
rs112094073 Chr.22: 36284401-36284401 on Build GRCh38
rs778668519 Chr.22: 36284395-36284395 on Build GRCh38
rs533453470 Chr.22: 36284490-36284490 on Build GRCh38
rs760681627 Chr.22: 36284479-36284479 on Build GRCh38
rs754825078 Chr.22: 36284394-36284394 on Build GRCh38
rs773325666 Chr.22: 36284472-36284472 on Build GRCh38
rs759956261 Chr.22: 36284420-36284420 on Build GRCh38
rs201174456 Chr.22: 36284453-36284453 on Build GRCh38
rs765102267 Chr.22: 36284514-36284514 on Build GRCh38
rs746568745 Chr.22: 36284431-36284431 on Build GRCh38
rs964706010 Chr.22: 36284475-36284475 on Build GRCh38
rs769044774 Chr.22: 36284457-36284457 on Build GRCh38
rs991435881 Chr.22: 36284421-36284421 on Build GRCh38
rs958590413 Chr.22: 36284416-36284416 on Build GRCh38
rs113947470 Chr.22: 36284400-36284400 on Build GRCh38
rs770321765 Chr.22: 36284433-36284433 on Build GRCh38
rs971112582 Chr.22: 36284504-36284504 on Build GRCh38

View Full Product Details