Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206126470-206126741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS1
Entrez Gene ID
4719
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
Gene Aliases
CI-75Kd, CI-75k, PRO1304
UniGene
Hs.471207
Gene Chromosome Location
Chr.2: 206121971-206159519 on Build GRCh38
Transcripts
NM_001199981.1 NM_001199982.1 NM_001199983.1 NM_001199984.1 NM_005006.6 XM_017004188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6723783 c.2092G>T p.D698Y Chr.2: 206126539-206126539 on Build GRCh38
6089971 c.2088G>T p.M696I Chr.2: 206126543-206126543 on Build GRCh38
6089970 c.2130G>T p.M710I Chr.2: 206126543-206126543 on Build GRCh38
6723782 c.2134G>T p.D712Y Chr.2: 206126539-206126539 on Build GRCh38
4154761 c.2006A>G p.N669S Chr.2: 206126723-206126723 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774782792 Chr.2: 206126609-206126609 on Build GRCh38
rs1034633382 Chr.2: 206126657-206126657 on Build GRCh38
rs985389260 Chr.2: 206126641-206126641 on Build GRCh38
rs759104039 Chr.2: 206126552-206126552 on Build GRCh38
rs760215826 Chr.2: 206126559-206126559 on Build GRCh38
rs768984620 Chr.2: 206126603-206126603 on Build GRCh38
rs751257853 Chr.2: 206126569-206126569 on Build GRCh38
rs773465401 Chr.2: 206126547-206126547 on Build GRCh38
rs377157717 Chr.2: 206126583-206126583 on Build GRCh38
rs374212468 Chr.2: 206126651-206126651 on Build GRCh38
rs960911568 Chr.2: 206126549-206126549 on Build GRCh38
rs540872475 Chr.2: 206126578-206126578 on Build GRCh38
rs377115146 Chr.2: 206126535-206126535 on Build GRCh38
rs765314993 Chr.2: 206126557-206126557 on Build GRCh38
rs563517641 Chr.2: 206126528-206126528 on Build GRCh38
rs750673640 Chr.2: 206126695-206126695 on Build GRCh38
rs757020109 Chr.2: 206126679-206126679 on Build GRCh38
rs760386524 Chr.2: 206126541-206126541 on Build GRCh38
rs148491633 Chr.2: 206126568-206126568 on Build GRCh38
rs1127600 Chr.2: 206126560-206126560 on Build GRCh38
rs139928027 Chr.2: 206126538-206126538 on Build GRCh38
rs564354783 Chr.2: 206126660-206126660 on Build GRCh38
rs780194304 Chr.2: 206126702-206126702 on Build GRCh38
rs751441546 Chr.2: 206126670-206126670 on Build GRCh38
rs567648334 Chr.2: 206126506-206126506 on Build GRCh38
rs749822261 Chr.2: 206126601-206126601 on Build GRCh38
rs992433859 Chr.2: 206126559-206126559 on Build GRCh38
rs781392058 Chr.2: 206126581-206126581 on Build GRCh38
rs753673133 Chr.2: 206126660-206126661 on Build GRCh38
rs753465234 Chr.2: 206126548-206126548 on Build GRCh38
rs762263731 Chr.2: 206126634-206126634 on Build GRCh38
rs372283660 Chr.2: 206126662-206126662 on Build GRCh38
rs770765893 Chr.2: 206126639-206126639 on Build GRCh38
rs971423942 Chr.2: 206126602-206126602 on Build GRCh38
rs565291585 Chr.2: 206126650-206126650 on Build GRCh38
rs763906959 Chr.2: 206126656-206126657 on Build GRCh38
rs370252506 Chr.2: 206126558-206126558 on Build GRCh38
rs745993097 Chr.2: 206126582-206126582 on Build GRCh38
rs909393619 Chr.2: 206126673-206126673 on Build GRCh38
rs757479318 Chr.2: 206126573-206126573 on Build GRCh38
rs375291720 Chr.2: 206126664-206126664 on Build GRCh38
rs775949937 Chr.2: 206126505-206126505 on Build GRCh38
rs756276722 Chr.2: 206126700-206126700 on Build GRCh38
rs769092536 Chr.2: 206126536-206126536 on Build GRCh38
rs55793630 Chr.2: 206126692-206126692 on Build GRCh38
rs772112878 Chr.2: 206126504-206126504 on Build GRCh38
rs749258377 Chr.2: 206126709-206126709 on Build GRCh38
rs868793978 Chr.2: 206126571-206126571 on Build GRCh38
rs969580950 Chr.2: 206126605-206126605 on Build GRCh38
rs142684807 Chr.2: 206126599-206126599 on Build GRCh38

View Full Product Details