Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28914443-28914688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABEP2
Entrez Gene ID
79874
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
FRA
UniGene
Hs.555978
Gene Chromosome Location
Chr.16: 28904421-28925211 on Build GRCh38
Transcripts
NM_024816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964904 c.581C>T p.T194M Chr.16: 28914549-28914549 on Build GRCh38
3110426 c.661G>A p.A221T Chr.16: 28914469-28914469 on Build GRCh38
6294588 c.543+10C>A p.? Chr.16: 28914662-28914662 on Build GRCh38
969586 c.633G>T p.E211D Chr.16: 28914497-28914497 on Build GRCh38
374213 c.651G>A p.G217G Chr.16: 28914479-28914479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752147216 Chr.16: 28914625-28914625 on Build GRCh38
rs905512475 Chr.16: 28914634-28914634 on Build GRCh38
rs765487241 Chr.16: 28914607-28914607 on Build GRCh38
rs766151402 Chr.16: 28914557-28914557 on Build GRCh38
rs753478169 Chr.16: 28914636-28914636 on Build GRCh38
rs913664073 Chr.16: 28914615-28914615 on Build GRCh38
rs780725821 Chr.16: 28914570-28914570 on Build GRCh38
rs1044091361 Chr.16: 28914627-28914627 on Build GRCh38
rs965610516 Chr.16: 28914512-28914512 on Build GRCh38
rs754503497 Chr.16: 28914517-28914517 on Build GRCh38
rs755628227 Chr.16: 28914632-28914632 on Build GRCh38
rs774661924 Chr.16: 28914521-28914521 on Build GRCh38
rs757611376 Chr.16: 28914470-28914470 on Build GRCh38
rs376906760 Chr.16: 28914603-28914603 on Build GRCh38
rs372536339 Chr.16: 28914549-28914549 on Build GRCh38
rs765092338 Chr.16: 28914477-28914477 on Build GRCh38
rs759481480 Chr.16: 28914619-28914619 on Build GRCh38
rs1018006621 Chr.16: 28914551-28914551 on Build GRCh38
rs776901280 Chr.16: 28914541-28914541 on Build GRCh38
rs746165504 Chr.16: 28914577-28914577 on Build GRCh38
rs769074403 Chr.16: 28914591-28914591 on Build GRCh38
rs561077923 Chr.16: 28914528-28914528 on Build GRCh38
rs756909210 Chr.16: 28914637-28914637 on Build GRCh38
rs751347906 Chr.16: 28914617-28914617 on Build GRCh38
rs776070926 Chr.16: 28914531-28914531 on Build GRCh38
rs528258382 Chr.16: 28914548-28914548 on Build GRCh38
rs769480150 Chr.16: 28914519-28914519 on Build GRCh38
rs571421473 Chr.16: 28914576-28914576 on Build GRCh38
rs758361119 Chr.16: 28914495-28914495 on Build GRCh38
rs546801470 Chr.16: 28914568-28914568 on Build GRCh38
rs754086429 Chr.16: 28914469-28914469 on Build GRCh38
rs773561353 Chr.16: 28914608-28914608 on Build GRCh38
rs763140583 Chr.16: 28914611-28914611 on Build GRCh38
rs752951563 Chr.16: 28914613-28914613 on Build GRCh38
rs754916909 Chr.16: 28914564-28914564 on Build GRCh38
rs891071141 Chr.16: 28914664-28914664 on Build GRCh38
rs781771546 Chr.16: 28914595-28914595 on Build GRCh38
rs776802979 Chr.16: 28914596-28914596 on Build GRCh38
rs200867018 Chr.16: 28914582-28914582 on Build GRCh38
rs751498703 Chr.16: 28914562-28914562 on Build GRCh38
rs375831600 Chr.16: 28914656-28914656 on Build GRCh38
rs750274682 Chr.16: 28914488-28914488 on Build GRCh38
rs764221819 Chr.16: 28914538-28914538 on Build GRCh38
rs1018846602 Chr.16: 28914501-28914501 on Build GRCh38
rs780435722 Chr.16: 28914650-28914650 on Build GRCh38
rs746435432 Chr.16: 28914513-28914513 on Build GRCh38
rs375415976 Chr.16: 28914529-28914529 on Build GRCh38
rs3924376 Chr.16: 28914657-28914657 on Build GRCh38
rs758271327 Chr.16: 28914556-28914556 on Build GRCh38
rs780052455 Chr.16: 28914496-28914496 on Build GRCh38
rs369333079 Chr.16: 28914609-28914609 on Build GRCh38
rs766170732 Chr.16: 28914616-28914616 on Build GRCh38
rs554753074 Chr.16: 28914662-28914662 on Build GRCh38
rs896988321 Chr.16: 28914515-28914515 on Build GRCh38
rs747355458 Chr.16: 28914651-28914651 on Build GRCh38
rs748729728 Chr.16: 28914658-28914658 on Build GRCh38
rs750182845 Chr.16: 28914554-28914554 on Build GRCh38
rs988903786 Chr.16: 28914599-28914599 on Build GRCh38
rs777333571 Chr.16: 28914635-28914635 on Build GRCh38
rs769149372 Chr.16: 28914653-28914653 on Build GRCh38
rs780620978 Chr.16: 28914518-28914518 on Build GRCh38
rs767469780 Chr.16: 28914623-28914623 on Build GRCh38
rs550518363 Chr.16: 28914583-28914583 on Build GRCh38

View Full Product Details