Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 16588635-16588908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDB2
Entrez Gene ID
9079
Gene Name
LIM domain binding 2
Gene Aliases
CLIM1, LDB-2, LDB1
UniGene
Hs.714330
Gene Chromosome Location
Chr.4: 16501534-16898809 on Build GRCh38
Transcripts
NM_001130834.2 NM_001290.4 NM_001304434.1 NM_001304435.1 XM_005248197.3 XM_005248201.2 XM_005248202.2 XM_006713975.3 XM_006713976.3 XM_006713977.3 XM_006713978.3 XM_006713979.3 XM_006713980.3 XM_006713981.3 XM_006713982.3 XM_006713983.3 XM_011513905.2 XM_017008812.1 XM_017008813.1 XM_017008814.1 XM_017008815.1 XM_017008816.1 XM_017008817.1 XM_017008818.1 XM_017008819.1 XM_017008820.1 XM_017008821.1 XM_017008822.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848394 c.425G>C p.R142T Chr.4: 16588816-16588816 on Build GRCh38
6700179 c.119A>G p.Q40R Chr.4: 16588750-16588750 on Build GRCh38
365813 c.496C>A p.R166R Chr.4: 16588745-16588745 on Build GRCh38
3674129 c.531T>C p.H177H Chr.4: 16588710-16588710 on Build GRCh38
6700185 c.82C>T p.L28F Chr.4: 16588787-16588787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747953774 Chr.4: 16588668-16588668 on Build GRCh38
rs778810128 Chr.4: 16588661-16588661 on Build GRCh38
rs575292293 Chr.4: 16588852-16588852 on Build GRCh38
rs774846224 Chr.4: 16588843-16588843 on Build GRCh38
rs377653994 Chr.4: 16588860-16588860 on Build GRCh38
rs766017618 Chr.4: 16588734-16588734 on Build GRCh38
rs370724459 Chr.4: 16588756-16588756 on Build GRCh38
rs982293354 Chr.4: 16588745-16588745 on Build GRCh38
rs771032741 Chr.4: 16588692-16588692 on Build GRCh38
rs771934255 Chr.4: 16588869-16588869 on Build GRCh38
rs758397394 Chr.4: 16588882-16588882 on Build GRCh38
rs1039760026 Chr.4: 16588847-16588847 on Build GRCh38
rs541432412 Chr.4: 16588665-16588666 on Build GRCh38
rs749377598 Chr.4: 16588824-16588824 on Build GRCh38
rs772829738 Chr.4: 16588668-16588668 on Build GRCh38
rs935153071 Chr.4: 16588716-16588716 on Build GRCh38
rs746746213 Chr.4: 16588782-16588782 on Build GRCh38
rs867687482 Chr.4: 16588822-16588822 on Build GRCh38
rs745957305 Chr.4: 16588823-16588823 on Build GRCh38
rs770479786 Chr.4: 16588711-16588711 on Build GRCh38
rs201913394 Chr.4: 16588686-16588686 on Build GRCh38
rs771836326 Chr.4: 16588670-16588670 on Build GRCh38
rs776080275 Chr.4: 16588721-16588721 on Build GRCh38
rs763230398 Chr.4: 16588729-16588729 on Build GRCh38
rs373557534 Chr.4: 16588829-16588829 on Build GRCh38
rs769013294 Chr.4: 16588832-16588832 on Build GRCh38
rs142034467 Chr.4: 16588744-16588744 on Build GRCh38
rs915317684 Chr.4: 16588865-16588865 on Build GRCh38
rs780153433 Chr.4: 16588875-16588875 on Build GRCh38
rs138524887 Chr.4: 16588732-16588732 on Build GRCh38
rs753404871 Chr.4: 16588740-16588740 on Build GRCh38
rs535184275 Chr.4: 16588681-16588681 on Build GRCh38
rs771249608 Chr.4: 16588785-16588785 on Build GRCh38
rs764768245 Chr.4: 16588666-16588666 on Build GRCh38
rs773341444 Chr.4: 16588672-16588672 on Build GRCh38
rs776172181 Chr.4: 16588825-16588825 on Build GRCh38
rs745752435 Chr.4: 16588757-16588757 on Build GRCh38
rs762071438 Chr.4: 16588858-16588858 on Build GRCh38
rs777669362 Chr.4: 16588771-16588771 on Build GRCh38
rs762660450 Chr.4: 16588848-16588849 on Build GRCh38
rs367619914 Chr.4: 16588667-16588668 on Build GRCh38
rs753424931 Chr.4: 16588866-16588866 on Build GRCh38
rs781548489 Chr.4: 16588788-16588788 on Build GRCh38
rs200875552 Chr.4: 16588731-16588731 on Build GRCh38
rs150687426 Chr.4: 16588770-16588770 on Build GRCh38
rs113993338 Chr.4: 16588761-16588761 on Build GRCh38
rs759681472 Chr.4: 16588710-16588710 on Build GRCh38
rs780496128 Chr.4: 16588701-16588701 on Build GRCh38

View Full Product Details