Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 87518193-87518438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL4
Entrez Gene ID
56062
Gene Name
kelch like family member 4
Gene Aliases
DKELCHL, KHL4
UniGene
Hs.49075
Gene Chromosome Location
Chr.X: 87517541-87670050 on Build GRCh38
Transcripts
NM_019117.4 NM_057162.2

SNP Detail

SNP ID Chromosome Location
rs758407325 Chr.X: 87518319-87518319 on Build GRCh38
rs747107232 Chr.X: 87518398-87518398 on Build GRCh38
rs755863623 Chr.X: 87518343-87518343 on Build GRCh38
rs748198647 Chr.X: 87518367-87518367 on Build GRCh38
rs750484826 Chr.X: 87518331-87518331 on Build GRCh38
rs763209456 Chr.X: 87518370-87518370 on Build GRCh38
rs768932351 Chr.X: 87518356-87518356 on Build GRCh38
rs777412756 Chr.X: 87518272-87518272 on Build GRCh38
rs749497717 Chr.X: 87518354-87518354 on Build GRCh38
rs186431341 Chr.X: 87518232-87518232 on Build GRCh38
rs752370580 Chr.X: 87518265-87518265 on Build GRCh38
rs756726092 Chr.X: 87518303-87518303 on Build GRCh38
rs766877636 Chr.X: 87518255-87518255 on Build GRCh38
rs781128337 Chr.X: 87518308-87518308 on Build GRCh38
rs766570678 Chr.X: 87518400-87518400 on Build GRCh38
rs774439055 Chr.X: 87518364-87518364 on Build GRCh38
rs144159864 Chr.X: 87518297-87518297 on Build GRCh38
rs745816672 Chr.X: 87518339-87518339 on Build GRCh38
rs779873296 Chr.X: 87518347-87518347 on Build GRCh38
rs780241301 Chr.X: 87518380-87518380 on Build GRCh38

View Full Product Details