Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57525422-57525663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBD6
Entrez Gene ID
114785
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
-
UniGene
Hs.524523
Gene Chromosome Location
Chr.12: 57520710-57530148 on Build GRCh38
Transcripts
NM_052897.3 XM_005268617.1 XM_005268618.1 XM_005268620.2 XM_006719217.1 XM_006719218.1 XM_011537851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575616 c.551delT p.F185fs*83 Chr.12: 57525519-57525519 on Build GRCh38
5802394 c.600delC p.R202fs*66 Chr.12: 57525568-57525568 on Build GRCh38
4450346 c.555delC p.P187fs*81 Chr.12: 57525523-57525523 on Build GRCh38
353810 c.681C>A p.Y227* Chr.12: 57525649-57525649 on Build GRCh38
5781614 c.601C>T p.P201S Chr.12: 57525569-57525569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376941207 Chr.12: 57525488-57525488 on Build GRCh38
rs761129930 Chr.12: 57525633-57525633 on Build GRCh38
rs907195350 Chr.12: 57525562-57525562 on Build GRCh38
rs765386670 Chr.12: 57525527-57525527 on Build GRCh38
rs368801753 Chr.12: 57525620-57525620 on Build GRCh38
rs771785025 Chr.12: 57525513-57525513 on Build GRCh38
rs773594377 Chr.12: 57525528-57525528 on Build GRCh38
rs568608586 Chr.12: 57525519-57525519 on Build GRCh38
rs778344227 Chr.12: 57525557-57525557 on Build GRCh38
rs374612145 Chr.12: 57525536-57525536 on Build GRCh38
rs535184098 Chr.12: 57525450-57525450 on Build GRCh38
rs910322923 Chr.12: 57525475-57525475 on Build GRCh38
rs781327833 Chr.12: 57525598-57525598 on Build GRCh38
rs756612577 Chr.12: 57525619-57525619 on Build GRCh38
rs1010453461 Chr.12: 57525544-57525544 on Build GRCh38
rs756752333 Chr.12: 57525490-57525490 on Build GRCh38
rs778572979 Chr.12: 57525493-57525493 on Build GRCh38
rs779725716 Chr.12: 57525572-57525572 on Build GRCh38
rs982498860 Chr.12: 57525632-57525632 on Build GRCh38
rs370675521 Chr.12: 57525479-57525479 on Build GRCh38
rs768077384 Chr.12: 57525548-57525548 on Build GRCh38
rs766660837 Chr.12: 57525540-57525540 on Build GRCh38
rs371650701 Chr.12: 57525523-57525523 on Build GRCh38
rs775314817 Chr.12: 57525516-57525516 on Build GRCh38
rs148047385 Chr.12: 57525552-57525552 on Build GRCh38
rs768306171 Chr.12: 57525587-57525587 on Build GRCh38
rs745493399 Chr.12: 57525497-57525497 on Build GRCh38
rs756805545 Chr.12: 57525554-57525554 on Build GRCh38
rs746924078 Chr.12: 57525518-57525518 on Build GRCh38
rs942203885 Chr.12: 57525638-57525638 on Build GRCh38
rs878918885 Chr.12: 57525455-57525455 on Build GRCh38
rs748050898 Chr.12: 57525604-57525604 on Build GRCh38
rs757832372 Chr.12: 57525641-57525641 on Build GRCh38
rs867867563 Chr.12: 57525630-57525630 on Build GRCh38
rs968961944 Chr.12: 57525522-57525522 on Build GRCh38
rs746890285 Chr.12: 57525573-57525573 on Build GRCh38
rs758000550 Chr.12: 57525570-57525570 on Build GRCh38
rs922347068 Chr.12: 57525600-57525600 on Build GRCh38
rs373733340 Chr.12: 57525483-57525483 on Build GRCh38
rs201489162 Chr.12: 57525547-57525547 on Build GRCh38
rs573105845 Chr.12: 57525626-57525626 on Build GRCh38
rs764731352 Chr.12: 57525635-57525635 on Build GRCh38
rs770028929 Chr.12: 57525611-57525611 on Build GRCh38
rs752041738 Chr.12: 57525457-57525457 on Build GRCh38
rs976412055 Chr.12: 57525582-57525582 on Build GRCh38
rs758738208 Chr.12: 57525448-57525448 on Build GRCh38
rs774428798 Chr.12: 57525622-57525622 on Build GRCh38
rs866560533 Chr.12: 57525612-57525612 on Build GRCh38
rs749888994 Chr.12: 57525636-57525636 on Build GRCh38
rs759983542 Chr.12: 57525624-57525624 on Build GRCh38
rs755450033 Chr.12: 57525458-57525458 on Build GRCh38
rs771245689 Chr.12: 57525621-57525621 on Build GRCh38
rs200229613 Chr.12: 57525613-57525613 on Build GRCh38

View Full Product Details