Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78995649-78995892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
UniGene
Hs.459035
Gene Chromosome Location
Chr.15: 78959947-79090873 on Build GRCh38
Transcripts
NM_001145648.1 NM_002891.4 NM_153815.2 XM_011521866.2 XM_011521867.2 XM_017022455.1 XM_017022456.1 XM_017022457.1 XM_017022458.1 XM_017022459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243400 c.3015-4C>T p.? Chr.15: 78995804-78995804 on Build GRCh38
6538351 c.3074T>C p.M1025T Chr.15: 78995741-78995741 on Build GRCh38
6077968 c.3060G>T p.E1020D Chr.15: 78995755-78995755 on Build GRCh38
4057186 c.3054G>A p.T1018T Chr.15: 78995761-78995761 on Build GRCh38
6763213 c.3021G>A p.L1007L Chr.15: 78995794-78995794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35937501 Chr.15: 78995843-78995843 on Build GRCh38
rs774094235 Chr.15: 78995818-78995818 on Build GRCh38
rs78449814 Chr.15: 78995843-78995843 on Build GRCh38
rs906106456 Chr.15: 78995709-78995709 on Build GRCh38
rs761957930 Chr.15: 78995749-78995749 on Build GRCh38
rs560005420 Chr.15: 78995826-78995826 on Build GRCh38
rs199713827 Chr.15: 78995762-78995762 on Build GRCh38
rs187075133 Chr.15: 78995700-78995700 on Build GRCh38
rs780605433 Chr.15: 78995761-78995761 on Build GRCh38
rs367800761 Chr.15: 78995746-78995746 on Build GRCh38
rs770570986 Chr.15: 78995797-78995797 on Build GRCh38
rs761987529 Chr.15: 78995819-78995819 on Build GRCh38
rs755030546 Chr.15: 78995863-78995863 on Build GRCh38
rs74786906 Chr.15: 78995842-78995842 on Build GRCh38
rs779617243 Chr.15: 78995874-78995874 on Build GRCh38
rs371581224 Chr.15: 78995815-78995815 on Build GRCh38
rs764358114 Chr.15: 78995715-78995715 on Build GRCh38
rs769687966 Chr.15: 78995813-78995813 on Build GRCh38
rs568366991 Chr.15: 78995864-78995864 on Build GRCh38
rs750530992 Chr.15: 78995823-78995823 on Build GRCh38
rs1056408689 Chr.15: 78995764-78995764 on Build GRCh38
rs6495359 Chr.15: 78995669-78995669 on Build GRCh38
rs767514389 Chr.15: 78995754-78995754 on Build GRCh38
rs34428440 Chr.15: 78995840-78995840 on Build GRCh38
rs756619146 Chr.15: 78995758-78995758 on Build GRCh38
rs386785659 Chr.15: 78995840-78995843 on Build GRCh38
rs937627811 Chr.15: 78995717-78995717 on Build GRCh38
rs192851872 Chr.15: 78995814-78995814 on Build GRCh38
rs950013411 Chr.15: 78995688-78995688 on Build GRCh38
rs543546791 Chr.15: 78995676-78995676 on Build GRCh38
rs202149543 Chr.15: 78995839-78995843 on Build GRCh38
rs769751685 Chr.15: 78995691-78995691 on Build GRCh38
rs577416088 Chr.15: 78995780-78995780 on Build GRCh38
rs371768177 Chr.15: 78995757-78995757 on Build GRCh38
rs767744886 Chr.15: 78995822-78995822 on Build GRCh38
rs768447442 Chr.15: 78995816-78995816 on Build GRCh38
rs775192361 Chr.15: 78995693-78995693 on Build GRCh38
rs1038496543 Chr.15: 78995718-78995718 on Build GRCh38
rs780893396 Chr.15: 78995851-78995851 on Build GRCh38
rs745353682 Chr.15: 78995812-78995812 on Build GRCh38
rs375192909 Chr.15: 78995839-78995839 on Build GRCh38
rs7173604 Chr.15: 78995850-78995850 on Build GRCh38
rs766311063 Chr.15: 78995827-78995827 on Build GRCh38
rs780617118 Chr.15: 78995809-78995809 on Build GRCh38

View Full Product Details