Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48970276-48970537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT10B
Entrez Gene ID
7480
Gene Name
Wnt family member 10B
Gene Aliases
SHFM6, WNT-12
UniGene
Hs.91985
Gene Chromosome Location
Chr.12: 48965340-48971858 on Build GRCh38
Transcripts
NM_003394.3 XM_011538724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761352 c.74G>A p.R25Q Chr.12: 48970456-48970456 on Build GRCh38
693729 c.46G>A p.G16S Chr.12: 48970484-48970484 on Build GRCh38
1232815 c.55T>C p.F19L Chr.12: 48970475-48970475 on Build GRCh38
1756779 c.35C>T p.S12L Chr.12: 48970495-48970495 on Build GRCh38
4953102 c.20C>A p.P7Q Chr.12: 48970510-48970510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772246409 Chr.12: 48970309-48970309 on Build GRCh38
rs189275380 Chr.12: 48970385-48970385 on Build GRCh38
rs376314279 Chr.12: 48970330-48970330 on Build GRCh38
rs1056953352 Chr.12: 48970343-48970343 on Build GRCh38
rs763644537 Chr.12: 48970446-48970446 on Build GRCh38
rs376351795 Chr.12: 48970509-48970509 on Build GRCh38
rs576014059 Chr.12: 48970390-48970390 on Build GRCh38
rs765724297 Chr.12: 48970351-48970351 on Build GRCh38
rs376064819 Chr.12: 48970362-48970362 on Build GRCh38
rs1000240555 Chr.12: 48970392-48970392 on Build GRCh38
rs986477630 Chr.12: 48970303-48970303 on Build GRCh38
rs763579407 Chr.12: 48970445-48970445 on Build GRCh38
rs993388545 Chr.12: 48970339-48970339 on Build GRCh38
rs977022015 Chr.12: 48970415-48970415 on Build GRCh38
rs941734373 Chr.12: 48970321-48970321 on Build GRCh38
rs1031682597 Chr.12: 48970404-48970404 on Build GRCh38
rs773382996 Chr.12: 48970312-48970312 on Build GRCh38
rs771121981 Chr.12: 48970514-48970514 on Build GRCh38
rs753716896 Chr.12: 48970483-48970483 on Build GRCh38
rs781070248 Chr.12: 48970370-48970370 on Build GRCh38
rs751121541 Chr.12: 48970447-48970447 on Build GRCh38
rs770305247 Chr.12: 48970422-48970422 on Build GRCh38
rs754912151 Chr.12: 48970484-48970484 on Build GRCh38
rs753550944 Chr.12: 48970325-48970326 on Build GRCh38
rs201214695 Chr.12: 48970387-48970387 on Build GRCh38
rs760085808 Chr.12: 48970344-48970344 on Build GRCh38
rs756054074 Chr.12: 48970383-48970383 on Build GRCh38
rs775822112 Chr.12: 48970440-48970440 on Build GRCh38
rs373412829 Chr.12: 48970494-48970494 on Build GRCh38
rs750199538 Chr.12: 48970467-48970467 on Build GRCh38
rs774472799 Chr.12: 48970307-48970307 on Build GRCh38
rs771208070 Chr.12: 48970406-48970406 on Build GRCh38
rs755821230 Chr.12: 48970476-48970476 on Build GRCh38
rs777844172 Chr.12: 48970398-48970398 on Build GRCh38
rs767121886 Chr.12: 48970457-48970457 on Build GRCh38
rs891760346 Chr.12: 48970400-48970400 on Build GRCh38
rs145655440 Chr.12: 48970316-48970316 on Build GRCh38
rs71459091 Chr.12: 48970396-48970397 on Build GRCh38
rs780487216 Chr.12: 48970417-48970417 on Build GRCh38
rs374560972 Chr.12: 48970486-48970486 on Build GRCh38
rs1037380551 Chr.12: 48970501-48970501 on Build GRCh38
rs766670692 Chr.12: 48970325-48970325 on Build GRCh38
rs367700694 Chr.12: 48970349-48970349 on Build GRCh38
rs761556530 Chr.12: 48970456-48970456 on Build GRCh38
rs371837779 Chr.12: 48970380-48970380 on Build GRCh38
rs747166206 Chr.12: 48970399-48970399 on Build GRCh38
rs184390483 Chr.12: 48970367-48970367 on Build GRCh38
rs1037328549 Chr.12: 48970328-48970328 on Build GRCh38
rs763732402 Chr.12: 48970363-48970363 on Build GRCh38
rs561483849 Chr.12: 48970477-48970477 on Build GRCh38
rs756913212 Chr.12: 48970377-48970385 on Build GRCh38
rs371482821 Chr.12: 48970359-48970359 on Build GRCh38
rs772090711 Chr.12: 48970393-48970393 on Build GRCh38
rs191833243 Chr.12: 48970409-48970409 on Build GRCh38

View Full Product Details