Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20369651-20369876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEP1
Entrez Gene ID
7011
Gene Name
telomerase associated protein 1
Gene Aliases
TLP1, TP1, TROVE1, VAULT2, p240
UniGene
Hs.508835
Gene Chromosome Location
Chr.14: 20365667-20413540 on Build GRCh38
Transcripts
NM_001319035.1 NM_007110.4 XM_005268027.4 XM_011537110.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5493134 c.7423+10A>G p.? Chr.14: 20369664-20369664 on Build GRCh38
3955866 c.7383T>C p.S2461S Chr.14: 20369714-20369714 on Build GRCh38
5647274 c.7397C>T p.S2466L Chr.14: 20369700-20369700 on Build GRCh38
6385255 c.7320G>T p.R2440S Chr.14: 20369777-20369777 on Build GRCh38
1228919 c.7382G>A p.S2461N Chr.14: 20369715-20369715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139512373 Chr.14: 20369777-20369777 on Build GRCh38
rs61030625 Chr.14: 20369835-20369835 on Build GRCh38
rs757777095 Chr.14: 20369688-20369688 on Build GRCh38
rs769409744 Chr.14: 20369831-20369831 on Build GRCh38
rs745374474 Chr.14: 20369687-20369687 on Build GRCh38
rs370853183 Chr.14: 20369741-20369741 on Build GRCh38
rs768184396 Chr.14: 20369778-20369778 on Build GRCh38
rs756908200 Chr.14: 20369673-20369673 on Build GRCh38
rs764520385 Chr.14: 20369784-20369784 on Build GRCh38
rs187374984 Chr.14: 20369699-20369699 on Build GRCh38
rs781166168 Chr.14: 20369819-20369819 on Build GRCh38
rs770044035 Chr.14: 20369726-20369726 on Build GRCh38
rs138309066 Chr.14: 20369770-20369770 on Build GRCh38
rs201140993 Chr.14: 20369774-20369774 on Build GRCh38
rs768609519 Chr.14: 20369700-20369700 on Build GRCh38
rs775243587 Chr.14: 20369695-20369695 on Build GRCh38
rs952545740 Chr.14: 20369822-20369822 on Build GRCh38
rs576018081 Chr.14: 20369678-20369678 on Build GRCh38
rs754889407 Chr.14: 20369818-20369818 on Build GRCh38
rs199748092 Chr.14: 20369763-20369763 on Build GRCh38
rs1027823499 Chr.14: 20369791-20369791 on Build GRCh38
rs201001220 Chr.14: 20369804-20369804 on Build GRCh38
rs750995222 Chr.14: 20369820-20369820 on Build GRCh38
rs749984247 Chr.14: 20369785-20369785 on Build GRCh38
rs777332859 Chr.14: 20369815-20369816 on Build GRCh38
rs911254374 Chr.14: 20369844-20369845 on Build GRCh38
rs747944020 Chr.14: 20369718-20369718 on Build GRCh38
rs747971587 Chr.14: 20369825-20369825 on Build GRCh38
rs947681920 Chr.14: 20369813-20369813 on Build GRCh38
rs761123055 Chr.14: 20369750-20369751 on Build GRCh38
rs532372748 Chr.14: 20369782-20369782 on Build GRCh38
rs779496925 Chr.14: 20369796-20369796 on Build GRCh38
rs758090740 Chr.14: 20369790-20369790 on Build GRCh38
rs1008883667 Chr.14: 20369710-20369710 on Build GRCh38
rs769080917 Chr.14: 20369809-20369810 on Build GRCh38
rs79167882 Chr.14: 20369695-20369695 on Build GRCh38
rs776378524 Chr.14: 20369702-20369702 on Build GRCh38

View Full Product Details