Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56420752-56420981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMELESS
Entrez Gene ID
8914
Gene Name
timeless circadian clock
Gene Aliases
TIM, TIM1, hTIM
UniGene
Hs.118631
Gene Chromosome Location
Chr.12: 56416373-56449416 on Build GRCh38
Transcripts
NM_003920.3 XM_011538939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1664565 c.3028C>G p.L1010V Chr.12: 56420975-56420975 on Build GRCh38
32250 c.3022C>G p.Q1008E Chr.12: 56420981-56420981 on Build GRCh38
5935976 c.3089C>T p.A1030V Chr.12: 56420833-56420833 on Build GRCh38
5070075 c.3083G>A p.R1028Q Chr.12: 56420839-56420839 on Build GRCh38
1705813 c.3082C>T p.R1028* Chr.12: 56420840-56420840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2638290 Chr.12: 56420822-56420822 on Build GRCh38
rs750201232 Chr.12: 56420785-56420785 on Build GRCh38
rs747981910 Chr.12: 56420938-56420938 on Build GRCh38
rs538000533 Chr.12: 56420895-56420895 on Build GRCh38
rs767511357 Chr.12: 56420865-56420865 on Build GRCh38
rs774316510 Chr.12: 56420925-56420925 on Build GRCh38
rs890101108 Chr.12: 56420893-56420893 on Build GRCh38
rs142128551 Chr.12: 56420877-56420877 on Build GRCh38
rs115228821 Chr.12: 56420880-56420880 on Build GRCh38
rs550004313 Chr.12: 56420825-56420825 on Build GRCh38
rs143413940 Chr.12: 56420868-56420868 on Build GRCh38
rs1040669995 Chr.12: 56420907-56420907 on Build GRCh38
rs763159822 Chr.12: 56420839-56420839 on Build GRCh38
rs812279 Chr.12: 56420786-56420786 on Build GRCh38
rs753391222 Chr.12: 56420794-56420794 on Build GRCh38
rs201585405 Chr.12: 56420857-56420857 on Build GRCh38
rs66491720 Chr.12: 56420779-56420779 on Build GRCh38
rs770141147 Chr.12: 56420890-56420890 on Build GRCh38
rs778398187 Chr.12: 56420812-56420812 on Build GRCh38
rs754438242 Chr.12: 56420810-56420810 on Build GRCh38
rs761715291 Chr.12: 56420781-56420781 on Build GRCh38
rs775624962 Chr.12: 56420836-56420836 on Build GRCh38
rs373156287 Chr.12: 56420934-56420934 on Build GRCh38
rs749439160 Chr.12: 56420911-56420911 on Build GRCh38
rs768881751 Chr.12: 56420916-56420916 on Build GRCh38
rs369088775 Chr.12: 56420931-56420931 on Build GRCh38
rs61376834 Chr.12: 56420872-56420872 on Build GRCh38
rs767388816 Chr.12: 56420783-56420783 on Build GRCh38
rs1036564218 Chr.12: 56420957-56420957 on Build GRCh38
rs761687091 Chr.12: 56420863-56420863 on Build GRCh38
rs765553995 Chr.12: 56420790-56420790 on Build GRCh38
rs150692151 Chr.12: 56420824-56420824 on Build GRCh38
rs201008145 Chr.12: 56420888-56420888 on Build GRCh38
rs144783032 Chr.12: 56420958-56420958 on Build GRCh38
rs770017871 Chr.12: 56420833-56420833 on Build GRCh38
rs781623652 Chr.12: 56420881-56420881 on Build GRCh38
rs202075297 Chr.12: 56420874-56420874 on Build GRCh38
rs200059760 Chr.12: 56420891-56420891 on Build GRCh38
rs774386629 Chr.12: 56420853-56420853 on Build GRCh38
rs2291739 Chr.12: 56420869-56420869 on Build GRCh38
rs372449809 Chr.12: 56420840-56420840 on Build GRCh38
rs773273529 Chr.12: 56420951-56420951 on Build GRCh38
rs201494969 Chr.12: 56420873-56420873 on Build GRCh38
rs746185610 Chr.12: 56420827-56420827 on Build GRCh38

View Full Product Details