Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57927672-57927911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF418
Entrez Gene ID
147686
Gene Name
zinc finger protein 418
Gene Aliases
-
UniGene
Hs.660728
Gene Chromosome Location
Chr.19: 57921882-57935404 on Build GRCh38
Transcripts
NM_001317027.1 NM_001317028.1 NM_001317029.1 NM_001317030.1 NM_133460.2 XM_017026304.1 XM_017026305.1 XM_017026306.1 XM_017026307.1 XM_017026308.1 XM_017026309.1 XM_017026310.1 XM_017026311.1 XM_017026312.1 XM_017026313.1 XM_017026314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1002547 c.416G>A p.S139N Chr.19: 57927765-57927765 on Build GRCh38
7399099 c.350A>G p.N117S Chr.19: 57927831-57927831 on Build GRCh38
3540582 c.281T>G p.I94S Chr.19: 57927900-57927900 on Build GRCh38
1002548 c.270C>A p.I90I Chr.19: 57927911-57927911 on Build GRCh38
4847902 c.476C>G p.S159C Chr.19: 57927705-57927705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760309022 Chr.19: 57927703-57927703 on Build GRCh38
rs375536083 Chr.19: 57927856-57927856 on Build GRCh38
rs148118518 Chr.19: 57927722-57927722 on Build GRCh38
rs761062015 Chr.19: 57927818-57927820 on Build GRCh38
rs1013901365 Chr.19: 57927871-57927871 on Build GRCh38
rs183050569 Chr.19: 57927705-57927705 on Build GRCh38
rs996446270 Chr.19: 57927867-57927878 on Build GRCh38
rs543987866 Chr.19: 57927804-57927804 on Build GRCh38
rs1022802872 Chr.19: 57927875-57927875 on Build GRCh38
rs28673938 Chr.19: 57927834-57927834 on Build GRCh38
rs749388698 Chr.19: 57927799-57927799 on Build GRCh38
rs150200636 Chr.19: 57927839-57927839 on Build GRCh38
rs79628444 Chr.19: 57927791-57927791 on Build GRCh38
rs775109234 Chr.19: 57927885-57927885 on Build GRCh38
rs773068256 Chr.19: 57927706-57927706 on Build GRCh38
rs936747681 Chr.19: 57927742-57927742 on Build GRCh38
rs748988325 Chr.19: 57927887-57927887 on Build GRCh38
rs776331808 Chr.19: 57927808-57927808 on Build GRCh38
rs770740807 Chr.19: 57927807-57927807 on Build GRCh38
rs769459630 Chr.19: 57927813-57927813 on Build GRCh38
rs376164701 Chr.19: 57927774-57927774 on Build GRCh38
rs1047217332 Chr.19: 57927837-57927837 on Build GRCh38
rs764588787 Chr.19: 57927751-57927751 on Build GRCh38
rs1001389363 Chr.19: 57927825-57927825 on Build GRCh38
rs1006141787 Chr.19: 57927854-57927854 on Build GRCh38
rs187390651 Chr.19: 57927725-57927725 on Build GRCh38
rs780424443 Chr.19: 57927838-57927838 on Build GRCh38
rs756342594 Chr.19: 57927833-57927833 on Build GRCh38
rs752235176 Chr.19: 57927726-57927726 on Build GRCh38
rs752072494 Chr.19: 57927773-57927773 on Build GRCh38
rs368413523 Chr.19: 57927792-57927792 on Build GRCh38
rs770701342 Chr.19: 57927701-57927701 on Build GRCh38
rs750623957 Chr.19: 57927783-57927783 on Build GRCh38
rs776336777 Chr.19: 57927704-57927704 on Build GRCh38
rs200431632 Chr.19: 57927827-57927827 on Build GRCh38
rs762444816 Chr.19: 57927817-57927817 on Build GRCh38
rs192674158 Chr.19: 57927879-57927879 on Build GRCh38
rs536484268 Chr.19: 57927824-57927824 on Build GRCh38
rs745665479 Chr.19: 57927869-57927869 on Build GRCh38
rs763652097 Chr.19: 57927821-57927821 on Build GRCh38
rs766769727 Chr.19: 57927833-57927840 on Build GRCh38
rs780991544 Chr.19: 57927859-57927859 on Build GRCh38
rs754258761 Chr.19: 57927873-57927874 on Build GRCh38
rs772071932 Chr.19: 57927802-57927802 on Build GRCh38
rs370547229 Chr.19: 57927780-57927780 on Build GRCh38
rs141826842 Chr.19: 57927803-57927803 on Build GRCh38
rs746935341 Chr.19: 57927849-57927849 on Build GRCh38
rs971290442 Chr.19: 57927889-57927889 on Build GRCh38
rs750772996 Chr.19: 57927831-57927831 on Build GRCh38
rs774925182 Chr.19: 57927816-57927816 on Build GRCh38
rs762265528 Chr.19: 57927826-57927826 on Build GRCh38
rs756318899 Chr.19: 57927786-57927786 on Build GRCh38
rs571896908 Chr.19: 57927723-57927723 on Build GRCh38
rs371111601 Chr.19: 57927743-57927743 on Build GRCh38

View Full Product Details