Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98585536-98585761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPOX
Entrez Gene ID
1371
Gene Name
coproporphyrinogen oxidase
Gene Aliases
CPO, CPX, HCP
UniGene
Hs.476982
Gene Chromosome Location
Chr.3: 98568448-98593684 on Build GRCh38
Transcripts
NM_000097.5 XM_005247125.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426010 c.1075A>G p.R359G Chr.3: 98585538-98585538 on Build GRCh38
1049241 c.1068C>G p.S356R Chr.3: 98585545-98585545 on Build GRCh38
6988695 c.1047G>A p.E349E Chr.3: 98585566-98585566 on Build GRCh38
732203 c.996G>T p.R332R Chr.3: 98585617-98585617 on Build GRCh38
1049242 c.995G>A p.R332Q Chr.3: 98585618-98585618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756432783 Chr.3: 98585621-98585621 on Build GRCh38
rs367947247 Chr.3: 98585670-98585670 on Build GRCh38
rs757811105 Chr.3: 98585605-98585605 on Build GRCh38
rs753601837 Chr.3: 98585568-98585568 on Build GRCh38
rs753600083 Chr.3: 98585673-98585673 on Build GRCh38
rs587777271 Chr.3: 98585633-98585633 on Build GRCh38
rs771079616 Chr.3: 98585710-98585710 on Build GRCh38
rs766384918 Chr.3: 98585560-98585560 on Build GRCh38
rs551789535 Chr.3: 98585676-98585676 on Build GRCh38
rs750242759 Chr.3: 98585697-98585698 on Build GRCh38
rs752340562 Chr.3: 98585578-98585578 on Build GRCh38
rs200327383 Chr.3: 98585674-98585674 on Build GRCh38
rs775145231 Chr.3: 98585677-98585677 on Build GRCh38
rs763715428 Chr.3: 98585689-98585689 on Build GRCh38
rs768991166 Chr.3: 98585634-98585634 on Build GRCh38
rs762633981 Chr.3: 98585688-98585688 on Build GRCh38
rs763662517 Chr.3: 98585600-98585600 on Build GRCh38
rs749579807 Chr.3: 98585630-98585630 on Build GRCh38
rs559774273 Chr.3: 98585664-98585664 on Build GRCh38
rs751091246 Chr.3: 98585705-98585705 on Build GRCh38
rs774609973 Chr.3: 98585647-98585647 on Build GRCh38
rs113274863 Chr.3: 98585590-98585590 on Build GRCh38
rs575595009 Chr.3: 98585620-98585620 on Build GRCh38
rs768795093 Chr.3: 98585557-98585557 on Build GRCh38
rs1002586414 Chr.3: 98585698-98585698 on Build GRCh38
rs781627991 Chr.3: 98585618-98585618 on Build GRCh38
rs121917873 Chr.3: 98585631-98585631 on Build GRCh38
rs368775479 Chr.3: 98585575-98585575 on Build GRCh38
rs376372510 Chr.3: 98585558-98585558 on Build GRCh38
rs759340330 Chr.3: 98585573-98585573 on Build GRCh38
rs201071538 Chr.3: 98585585-98585585 on Build GRCh38
rs1729995 Chr.3: 98585623-98585623 on Build GRCh38
rs11921054 Chr.3: 98585559-98585559 on Build GRCh38
rs989262487 Chr.3: 98585723-98585723 on Build GRCh38
rs121917866 Chr.3: 98585622-98585622 on Build GRCh38
rs751038412 Chr.3: 98585604-98585604 on Build GRCh38

View Full Product Details