Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32713166-32713439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
UniGene
Hs.444851
Gene Chromosome Location
Chr.3: 32685145-32773875 on Build GRCh38
Transcripts
NM_001256741.1 NM_001256742.1 NM_015442.2 NR_046352.1 XM_006713084.3 XM_006713085.3 XM_011533566.2 XM_017006109.1 XM_017006110.1 XM_017006111.1 XM_017006112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2984105 c.558A>T p.K186N Chr.3: 32713354-32713354 on Build GRCh38
6468841 c.433G>A p.E145K Chr.3: 32713229-32713229 on Build GRCh38
5974731 c.549C>T p.N183N Chr.3: 32713345-32713345 on Build GRCh38
4323604 c.728A>G p.N243S Chr.3: 32713344-32713344 on Build GRCh38
4398476 c.443C>T p.A148V Chr.3: 32713239-32713239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375124764 Chr.3: 32713390-32713390 on Build GRCh38
rs755141852 Chr.3: 32713270-32713270 on Build GRCh38
rs777183400 Chr.3: 32713391-32713391 on Build GRCh38
rs139334381 Chr.3: 32713274-32713274 on Build GRCh38
rs200222971 Chr.3: 32713219-32713219 on Build GRCh38
rs764577018 Chr.3: 32713230-32713230 on Build GRCh38
rs865993607 Chr.3: 32713281-32713281 on Build GRCh38
rs777093785 Chr.3: 32713237-32713237 on Build GRCh38
rs370558352 Chr.3: 32713258-32713258 on Build GRCh38
rs775884650 Chr.3: 32713211-32713212 on Build GRCh38
rs771688957 Chr.3: 32713211-32713211 on Build GRCh38
rs760348130 Chr.3: 32713228-32713228 on Build GRCh38
rs758497569 Chr.3: 32713267-32713267 on Build GRCh38
rs757203542 Chr.3: 32713304-32713304 on Build GRCh38
rs371469964 Chr.3: 32713268-32713268 on Build GRCh38
rs745374209 Chr.3: 32713208-32713208 on Build GRCh38
rs376161971 Chr.3: 32713271-32713271 on Build GRCh38
rs771625796 Chr.3: 32713347-32713347 on Build GRCh38
rs979494356 Chr.3: 32713229-32713229 on Build GRCh38
rs374004782 Chr.3: 32713259-32713259 on Build GRCh38
rs942748922 Chr.3: 32713225-32713225 on Build GRCh38
rs371042727 Chr.3: 32713386-32713386 on Build GRCh38
rs201398883 Chr.3: 32713379-32713379 on Build GRCh38
rs745726837 Chr.3: 32713345-32713345 on Build GRCh38
rs142210935 Chr.3: 32713240-32713240 on Build GRCh38
rs935369397 Chr.3: 32713299-32713299 on Build GRCh38
rs762223721 Chr.3: 32713401-32713401 on Build GRCh38
rs775092727 Chr.3: 32713356-32713356 on Build GRCh38
rs778909170 Chr.3: 32713340-32713340 on Build GRCh38
rs377611501 Chr.3: 32713276-32713276 on Build GRCh38
rs765730485 Chr.3: 32713402-32713402 on Build GRCh38
rs367958489 Chr.3: 32713407-32713407 on Build GRCh38

View Full Product Details