Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3783919-3784188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC47
Entrez Gene ID
57470
Gene Name
leucine rich repeat containing 47
Gene Aliases
-
UniGene
Hs.268488
Gene Chromosome Location
Chr.1: 3780220-3796504 on Build GRCh38
Transcripts
NM_020710.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5416838 c.1224G>T p.K408N Chr.1: 3784082-3784082 on Build GRCh38
4699116 c.1302C>T p.G434G Chr.1: 3784004-3784004 on Build GRCh38
5428082 c.1261G>T p.E421* Chr.1: 3784045-3784045 on Build GRCh38
6703916 c.1261G>A p.E421K Chr.1: 3784045-3784045 on Build GRCh38
2081679 c.1305G>C p.L435L Chr.1: 3784001-3784001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768169128 Chr.1: 3784041-3784041 on Build GRCh38
rs778414118 Chr.1: 3784131-3784131 on Build GRCh38
rs779882344 Chr.1: 3784020-3784020 on Build GRCh38
rs759594928 Chr.1: 3783989-3783989 on Build GRCh38
rs755365589 Chr.1: 3783974-3783974 on Build GRCh38
rs75360301 Chr.1: 3784060-3784060 on Build GRCh38
rs778807067 Chr.1: 3784045-3784045 on Build GRCh38
rs763952177 Chr.1: 3783943-3783943 on Build GRCh38
rs1035080284 Chr.1: 3784064-3784064 on Build GRCh38
rs779235885 Chr.1: 3784020-3784021 on Build GRCh38
rs138017878 Chr.1: 3783977-3783977 on Build GRCh38
rs200587617 Chr.1: 3784116-3784116 on Build GRCh38
rs758699833 Chr.1: 3783961-3783961 on Build GRCh38
rs376822471 Chr.1: 3783969-3783969 on Build GRCh38
rs768456710 Chr.1: 3783981-3783981 on Build GRCh38
rs754723598 Chr.1: 3784125-3784125 on Build GRCh38
rs201743263 Chr.1: 3784097-3784097 on Build GRCh38
rs372376497 Chr.1: 3784152-3784152 on Build GRCh38
rs374820243 Chr.1: 3784022-3784022 on Build GRCh38
rs752966528 Chr.1: 3784022-3784024 on Build GRCh38
rs981575660 Chr.1: 3783959-3783959 on Build GRCh38
rs376296720 Chr.1: 3784161-3784161 on Build GRCh38
rs893042756 Chr.1: 3784055-3784055 on Build GRCh38
rs372871019 Chr.1: 3783947-3783947 on Build GRCh38
rs202197521 Chr.1: 3784079-3784079 on Build GRCh38
rs370914502 Chr.1: 3783987-3783987 on Build GRCh38
rs757851284 Chr.1: 3784008-3784008 on Build GRCh38
rs368549162 Chr.1: 3784162-3784162 on Build GRCh38
rs1022817009 Chr.1: 3784083-3784083 on Build GRCh38
rs145640191 Chr.1: 3784007-3784007 on Build GRCh38
rs199585760 Chr.1: 3784134-3784134 on Build GRCh38
rs891131454 Chr.1: 3784035-3784035 on Build GRCh38
rs763254452 Chr.1: 3784004-3784004 on Build GRCh38
rs758151441 Chr.1: 3783965-3783965 on Build GRCh38
rs765454732 Chr.1: 3783991-3783991 on Build GRCh38
rs773071926 Chr.1: 3784065-3784065 on Build GRCh38
rs532513715 Chr.1: 3783958-3783958 on Build GRCh38
rs369025107 Chr.1: 3784046-3784046 on Build GRCh38
rs553798562 Chr.1: 3784067-3784067 on Build GRCh38
rs921454384 Chr.1: 3784167-3784167 on Build GRCh38
rs762028198 Chr.1: 3784085-3784085 on Build GRCh38
rs750920435 Chr.1: 3784109-3784109 on Build GRCh38
rs368444121 Chr.1: 3784117-3784117 on Build GRCh38
rs1015882724 Chr.1: 3784126-3784126 on Build GRCh38
rs371330730 Chr.1: 3783971-3783971 on Build GRCh38
rs763278653 Chr.1: 3784075-3784075 on Build GRCh38
rs755230175 Chr.1: 3783956-3783956 on Build GRCh38
rs765438214 Chr.1: 3783960-3783960 on Build GRCh38
rs138073750 Chr.1: 3784015-3784015 on Build GRCh38
rs747505620 Chr.1: 3783964-3783964 on Build GRCh38
rs775529048 Chr.1: 3784074-3784074 on Build GRCh38
rs751509865 Chr.1: 3784021-3784021 on Build GRCh38
rs781761723 Chr.1: 3784013-3784013 on Build GRCh38
rs761856310 Chr.1: 3783986-3783986 on Build GRCh38
rs771784324 Chr.1: 3784138-3784138 on Build GRCh38
rs766327720 Chr.1: 3784016-3784016 on Build GRCh38
rs777713622 Chr.1: 3784147-3784147 on Build GRCh38
rs181171007 Chr.1: 3783983-3783983 on Build GRCh38
rs753890734 Chr.1: 3783950-3783950 on Build GRCh38
rs373264626 Chr.1: 3783954-3783954 on Build GRCh38
rs768143516 Chr.1: 3784096-3784096 on Build GRCh38
rs375328574 Chr.1: 3784080-3784080 on Build GRCh38
rs774278398 Chr.1: 3783945-3783945 on Build GRCh38
rs754940236 Chr.1: 3783955-3783955 on Build GRCh38
rs369354364 Chr.1: 3784114-3784114 on Build GRCh38
rs752880681 Chr.1: 3783998-3783998 on Build GRCh38
rs766432491 Chr.1: 3783949-3783949 on Build GRCh38
rs745368866 Chr.1: 3783970-3783970 on Build GRCh38
rs74764187 Chr.1: 3784054-3784054 on Build GRCh38
rs905664720 Chr.1: 3784089-3784089 on Build GRCh38
rs372947494 Chr.1: 3784066-3784066 on Build GRCh38
rs775225327 Chr.1: 3783976-3783976 on Build GRCh38

View Full Product Details