Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73950426-73950673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGUOK
Entrez Gene ID
1716
Gene Name
deoxyguanosine kinase
Gene Aliases
MTDPS3, dGK
UniGene
Hs.469022
Gene Chromosome Location
Chr.2: 73926826-73958961 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643226 c.393G>T p.K131N Chr.2: 73950600-73950600 on Build GRCh38
3785319 c.463C>G p.L155V Chr.2: 73950604-73950604 on Build GRCh38
1690707 c.479C>T p.S160F Chr.2: 73950620-73950620 on Build GRCh38
4816639 c.481C>G p.L161V Chr.2: 73950622-73950622 on Build GRCh38
3840017 c.521C>G p.S174C Chr.2: 73950662-73950662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769253642 Chr.2: 73950617-73950617 on Build GRCh38
rs1006073607 Chr.2: 73950515-73950515 on Build GRCh38
rs772413307 Chr.2: 73950627-73950627 on Build GRCh38
rs946023730 Chr.2: 73950511-73950511 on Build GRCh38
rs914554593 Chr.2: 73950510-73950513 on Build GRCh38
rs745460442 Chr.2: 73950575-73950575 on Build GRCh38
rs774844548 Chr.2: 73950466-73950466 on Build GRCh38
rs751438357 Chr.2: 73950561-73950561 on Build GRCh38
rs770950831 Chr.2: 73950635-73950635 on Build GRCh38
rs748579483 Chr.2: 73950625-73950625 on Build GRCh38
rs1017344472 Chr.2: 73950519-73950519 on Build GRCh38
rs760888346 Chr.2: 73950634-73950634 on Build GRCh38
rs774810856 Chr.2: 73950620-73950620 on Build GRCh38
rs757112492 Chr.2: 73950562-73950562 on Build GRCh38
rs763819454 Chr.2: 73950546-73950546 on Build GRCh38
rs536746349 Chr.2: 73950574-73950574 on Build GRCh38
rs1026255073 Chr.2: 73950479-73950479 on Build GRCh38
rs139533561 Chr.2: 73950591-73950591 on Build GRCh38
rs995755469 Chr.2: 73950587-73950587 on Build GRCh38
rs140339626 Chr.2: 73950510-73950511 on Build GRCh38
rs148915942 Chr.2: 73950633-73950633 on Build GRCh38
rs990135536 Chr.2: 73950509-73950510 on Build GRCh38
rs528587600 Chr.2: 73950523-73950523 on Build GRCh38
rs369870836 Chr.2: 73950607-73950607 on Build GRCh38
rs546873527 Chr.2: 73950645-73950645 on Build GRCh38
rs758317393 Chr.2: 73950539-73950539 on Build GRCh38
rs938889819 Chr.2: 73950524-73950524 on Build GRCh38
rs951847350 Chr.2: 73950512-73950512 on Build GRCh38
rs975743023 Chr.2: 73950600-73950600 on Build GRCh38
rs561192133 Chr.2: 73950517-73950517 on Build GRCh38
rs144181978 Chr.2: 73950603-73950603 on Build GRCh38
rs776727459 Chr.2: 73950642-73950642 on Build GRCh38
rs74874677 Chr.2: 73950650-73950650 on Build GRCh38

View Full Product Details