Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186449304-186449549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724919
Entrez Gene ID
102724919
Gene Name
uncharacterized LOC102724919
Gene Aliases
-
UniGene
Hs.576493
Gene Chromosome Location
Chr.1: 186435161-186470291 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 XM_011509603.2

Gene Symbol
PDC
Entrez Gene ID
5132
Gene Name
phosducin
Gene Aliases
MEKA, PHD, PhLOP, PhLP
UniGene
Hs.654381
Gene Chromosome Location
Chr.1: 186443566-186461108 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 XM_011509603.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6742227 c.43G>A p.G15R Chr.1: 186449417-186449417 on Build GRCh38
4550413 c.4G>A p.E2K Chr.1: 186449456-186449456 on Build GRCh38
5844838 c.27G>T p.L9F Chr.1: 186449433-186449433 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751186174 Chr.1: 186449428-186449428 on Build GRCh38
rs757859950 Chr.1: 186449360-186449364 on Build GRCh38
rs755022016 Chr.1: 186449496-186449496 on Build GRCh38
rs922474332 Chr.1: 186449476-186449476 on Build GRCh38
rs779985738 Chr.1: 186449502-186449504 on Build GRCh38
rs369506577 Chr.1: 186449427-186449427 on Build GRCh38
rs2774532 Chr.1: 186449457-186449457 on Build GRCh38
rs775540696 Chr.1: 186449405-186449405 on Build GRCh38
rs780515635 Chr.1: 186449351-186449351 on Build GRCh38
rs2737150 Chr.1: 186449355-186449355 on Build GRCh38
rs756049792 Chr.1: 186449477-186449485 on Build GRCh38
rs945872579 Chr.1: 186449483-186449483 on Build GRCh38
rs762404613 Chr.1: 186449406-186449406 on Build GRCh38
rs754321407 Chr.1: 186449473-186449473 on Build GRCh38
rs563557888 Chr.1: 186449399-186449399 on Build GRCh38
rs2774531 Chr.1: 186449454-186449454 on Build GRCh38
rs770633904 Chr.1: 186449394-186449394 on Build GRCh38
rs761463580 Chr.1: 186449456-186449456 on Build GRCh38
rs377291422 Chr.1: 186449382-186449382 on Build GRCh38
rs749409578 Chr.1: 186449352-186449352 on Build GRCh38
rs545283554 Chr.1: 186449376-186449376 on Build GRCh38
rs12407957 Chr.1: 186449468-186449468 on Build GRCh38
rs769904390 Chr.1: 186449402-186449402 on Build GRCh38
rs377352034 Chr.1: 186449466-186449466 on Build GRCh38
rs768740990 Chr.1: 186449370-186449370 on Build GRCh38
rs549035553 Chr.1: 186449396-186449396 on Build GRCh38

View Full Product Details