Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62781807-62782062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF6L
Entrez Gene ID
10629
Gene Name
TATA-box binding protein associated factor 6 like
Gene Aliases
PAF65A
UniGene
Hs.714400
Gene Chromosome Location
Chr.11: 62771303-62787342 on Build GRCh38
Transcripts
NM_006473.3 XM_005273714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622227 c.560C>T p.S187F Chr.11: 62781922-62781922 on Build GRCh38
1355461 c.570G>A p.G190G Chr.11: 62781932-62781932 on Build GRCh38
1267382 c.550C>T p.Q184* Chr.11: 62781912-62781912 on Build GRCh38
929814 c.564G>T p.K188N Chr.11: 62781926-62781926 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767333499 Chr.11: 62781985-62781985 on Build GRCh38
rs767208601 Chr.11: 62781857-62781857 on Build GRCh38
rs892618951 Chr.11: 62781939-62781939 on Build GRCh38
rs376643499 Chr.11: 62781869-62781869 on Build GRCh38
rs181124445 Chr.11: 62781994-62781994 on Build GRCh38
rs947925940 Chr.11: 62781836-62781836 on Build GRCh38
rs776824631 Chr.11: 62781934-62781934 on Build GRCh38
rs752589970 Chr.11: 62781997-62781997 on Build GRCh38
rs754830816 Chr.11: 62781904-62781904 on Build GRCh38
rs761537085 Chr.11: 62781852-62781852 on Build GRCh38
rs906486021 Chr.11: 62781873-62781873 on Build GRCh38
rs773912140 Chr.11: 62781977-62781977 on Build GRCh38
rs748109811 Chr.11: 62781916-62781916 on Build GRCh38
rs772348407 Chr.11: 62781917-62781917 on Build GRCh38
rs1036267922 Chr.11: 62781872-62781872 on Build GRCh38
rs761399823 Chr.11: 62781983-62781983 on Build GRCh38
rs777959397 Chr.11: 62781926-62781926 on Build GRCh38
rs1010264188 Chr.11: 62781963-62781963 on Build GRCh38
rs555697514 Chr.11: 62781900-62781900 on Build GRCh38
rs901521485 Chr.11: 62781891-62781891 on Build GRCh38
rs754955720 Chr.11: 62781998-62781998 on Build GRCh38
rs1050173074 Chr.11: 62781909-62781909 on Build GRCh38
rs535425195 Chr.11: 62781847-62781847 on Build GRCh38
rs770145677 Chr.11: 62781955-62781955 on Build GRCh38
rs770295621 Chr.11: 62781987-62781987 on Build GRCh38
rs985648742 Chr.11: 62782037-62782037 on Build GRCh38
rs1002817841 Chr.11: 62781919-62781919 on Build GRCh38
rs780187358 Chr.11: 62781886-62781886 on Build GRCh38
rs993611114 Chr.11: 62781884-62781884 on Build GRCh38
rs145421020 Chr.11: 62781928-62781928 on Build GRCh38
rs888243462 Chr.11: 62781835-62781835 on Build GRCh38
rs377230261 Chr.11: 62781993-62781993 on Build GRCh38
rs746327823 Chr.11: 62781954-62781957 on Build GRCh38
rs779072560 Chr.11: 62781911-62781911 on Build GRCh38
rs760114599 Chr.11: 62781938-62781938 on Build GRCh38
rs778712506 Chr.11: 62782020-62782020 on Build GRCh38
rs763694808 Chr.11: 62781844-62781844 on Build GRCh38
rs185181799 Chr.11: 62782041-62782041 on Build GRCh38
rs575563368 Chr.11: 62781929-62781929 on Build GRCh38

View Full Product Details