Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14450245-14450500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKN1
Entrez Gene ID
5585
Gene Name
protein kinase N1
Gene Aliases
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
UniGene
Hs.466044
Gene Chromosome Location
Chr.19: 14433354-14471867 on Build GRCh38
Transcripts
NM_002741.3 NM_213560.1 XM_011528128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420389 c.641G>A p.R214H Chr.19: 14450348-14450348 on Build GRCh38
4074811 c.640C>T p.R214C Chr.19: 14450347-14450347 on Build GRCh38
6420388 c.659G>A p.R220H Chr.19: 14450348-14450348 on Build GRCh38
4714812 c.664G>A p.E222K Chr.19: 14450353-14450353 on Build GRCh38
7384138 c.727C>T p.R243C Chr.19: 14450416-14450416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773954469 Chr.19: 14450375-14450375 on Build GRCh38
rs372284261 Chr.19: 14450388-14450388 on Build GRCh38
rs752747644 Chr.19: 14450467-14450468 on Build GRCh38
rs201738052 Chr.19: 14450374-14450374 on Build GRCh38
rs367813415 Chr.19: 14450429-14450429 on Build GRCh38
rs200219286 Chr.19: 14450400-14450400 on Build GRCh38
rs535215114 Chr.19: 14450441-14450441 on Build GRCh38
rs748971281 Chr.19: 14450456-14450456 on Build GRCh38
rs764910777 Chr.19: 14450414-14450415 on Build GRCh38
rs779650993 Chr.19: 14450452-14450452 on Build GRCh38
rs568766672 Chr.19: 14450461-14450461 on Build GRCh38
rs536109196 Chr.19: 14450471-14450471 on Build GRCh38
rs758764518 Chr.19: 14450328-14450328 on Build GRCh38
rs1045385298 Chr.19: 14450450-14450450 on Build GRCh38
rs759428474 Chr.19: 14450298-14450299 on Build GRCh38
rs752963905 Chr.19: 14450440-14450440 on Build GRCh38
rs780300900 Chr.19: 14450287-14450287 on Build GRCh38
rs768512052 Chr.19: 14450371-14450371 on Build GRCh38
rs748701383 Chr.19: 14450320-14450320 on Build GRCh38
rs769398142 Chr.19: 14450352-14450352 on Build GRCh38
rs755099537 Chr.19: 14450434-14450434 on Build GRCh38
rs990696500 Chr.19: 14450432-14450432 on Build GRCh38
rs924607765 Chr.19: 14450383-14450383 on Build GRCh38
rs780871877 Chr.19: 14450334-14450334 on Build GRCh38
rs747513157 Chr.19: 14450446-14450446 on Build GRCh38
rs371342455 Chr.19: 14450460-14450460 on Build GRCh38
rs767592620 Chr.19: 14450277-14450277 on Build GRCh38
rs760786724 Chr.19: 14450403-14450403 on Build GRCh38
rs779131109 Chr.19: 14450307-14450307 on Build GRCh38
rs547358062 Chr.19: 14450445-14450445 on Build GRCh38
rs746213444 Chr.19: 14450263-14450263 on Build GRCh38
rs755741840 Chr.19: 14450447-14450447 on Build GRCh38
rs765639952 Chr.19: 14450438-14450438 on Build GRCh38
rs755435768 Chr.19: 14450301-14450301 on Build GRCh38
rs767390024 Chr.19: 14450395-14450395 on Build GRCh38
rs748882175 Chr.19: 14450361-14450361 on Build GRCh38
rs762581193 Chr.19: 14450399-14450399 on Build GRCh38
rs761601123 Chr.19: 14450276-14450276 on Build GRCh38
rs775350737 Chr.19: 14450360-14450360 on Build GRCh38
rs768150367 Chr.19: 14450459-14450459 on Build GRCh38
rs376296280 Chr.19: 14450472-14450472 on Build GRCh38
rs562462584 Chr.19: 14450274-14450274 on Build GRCh38
rs745421577 Chr.19: 14450335-14450335 on Build GRCh38
rs950477235 Chr.19: 14450364-14450364 on Build GRCh38
rs750466566 Chr.19: 14450279-14450279 on Build GRCh38
rs766445271 Chr.19: 14450405-14450405 on Build GRCh38
rs770373043 Chr.19: 14450348-14450348 on Build GRCh38
rs754149651 Chr.19: 14450412-14450412 on Build GRCh38
rs753973751 Chr.19: 14450300-14450300 on Build GRCh38
rs978722232 Chr.19: 14450321-14450321 on Build GRCh38

View Full Product Details