Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 17068337-17068562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REPS2
Entrez Gene ID
9185
Gene Name
RALBP1 associated Eps domain containing 2
Gene Aliases
POB1
UniGene
Hs.186810
Gene Chromosome Location
Chr.X: 16946691-17194274 on Build GRCh38
Transcripts
NM_001080975.1 NM_004726.2 XM_005274625.3 XM_005274626.3 XM_011545603.2 XM_011545604.2 XM_011545605.2 XM_011545606.2 XM_011545607.2 XM_011545609.2 XM_017029955.1 XM_017029956.1 XM_017029957.1 XM_017029958.1

SNP Detail

SNP ID Chromosome Location
rs769349139 Chr.X: 17068377-17068377 on Build GRCh38
rs920597803 Chr.X: 17068466-17068466 on Build GRCh38
rs377668547 Chr.X: 17068503-17068503 on Build GRCh38
rs766042969 Chr.X: 17068408-17068408 on Build GRCh38
rs758995290 Chr.X: 17068522-17068522 on Build GRCh38
rs762879456 Chr.X: 17068386-17068386 on Build GRCh38
rs956535345 Chr.X: 17068420-17068420 on Build GRCh38
rs773072057 Chr.X: 17068385-17068385 on Build GRCh38
rs975916785 Chr.X: 17068423-17068423 on Build GRCh38
rs181282465 Chr.X: 17068425-17068425 on Build GRCh38
rs750922644 Chr.X: 17068506-17068506 on Build GRCh38
rs868556060 Chr.X: 17068441-17068441 on Build GRCh38
rs757355242 Chr.X: 17068481-17068481 on Build GRCh38
rs751349371 Chr.X: 17068422-17068422 on Build GRCh38
rs764375554 Chr.X: 17068433-17068433 on Build GRCh38
rs184329917 Chr.X: 17068445-17068445 on Build GRCh38

View Full Product Details