Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 66355436-66355709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE4B
Entrez Gene ID
5142
Gene Name
phosphodiesterase 4B
Gene Aliases
DPDE4, PDEIVB
UniGene
Hs.198072
Gene Chromosome Location
Chr.1: 65792510-66374579 on Build GRCh38
Transcripts
NM_001037339.2 NM_001037340.2 NM_001037341.1 NM_001297440.1 NM_001297441.1 NM_001297442.1 NM_002600.3 XM_005270924.3 XM_005270925.2 XM_006710680.3 XM_011541566.1 XM_017001445.1 XM_017001446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627157 c.325+7T>C p.? Chr.1: 66355627-66355627 on Build GRCh38
6540512 c.252G>T p.R84R Chr.1: 66355547-66355547 on Build GRCh38
357698 c.287G>A p.G96E Chr.1: 66355582-66355582 on Build GRCh38
215657 c.755C>T p.S252L Chr.1: 66355579-66355579 on Build GRCh38
3705993 c.795C>A p.S265R Chr.1: 66355574-66355574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771492592 Chr.1: 66355491-66355491 on Build GRCh38
rs75814276 Chr.1: 66355477-66355477 on Build GRCh38
rs201804146 Chr.1: 66355626-66355626 on Build GRCh38
rs199554373 Chr.1: 66355576-66355576 on Build GRCh38
rs944398383 Chr.1: 66355505-66355505 on Build GRCh38
rs758874556 Chr.1: 66355569-66355569 on Build GRCh38
rs765651623 Chr.1: 66355545-66355545 on Build GRCh38
rs776447800 Chr.1: 66355665-66355665 on Build GRCh38
rs750880967 Chr.1: 66355492-66355492 on Build GRCh38
rs772112354 Chr.1: 66355634-66355634 on Build GRCh38
rs376229903 Chr.1: 66355548-66355548 on Build GRCh38
rs751719874 Chr.1: 66355583-66355583 on Build GRCh38
rs935950947 Chr.1: 66355678-66355678 on Build GRCh38
rs757527325 Chr.1: 66355597-66355597 on Build GRCh38
rs760014855 Chr.1: 66355519-66355519 on Build GRCh38
rs531284366 Chr.1: 66355673-66355673 on Build GRCh38
rs759209285 Chr.1: 66355669-66355669 on Build GRCh38
rs761759889 Chr.1: 66355488-66355488 on Build GRCh38
rs145203756 Chr.1: 66355551-66355551 on Build GRCh38
rs765702770 Chr.1: 66355670-66355670 on Build GRCh38
rs770528897 Chr.1: 66355652-66355652 on Build GRCh38
rs528931858 Chr.1: 66355587-66355587 on Build GRCh38
rs371297521 Chr.1: 66355609-66355609 on Build GRCh38
rs768408591 Chr.1: 66355479-66355479 on Build GRCh38
rs983059398 Chr.1: 66355478-66355478 on Build GRCh38
rs112142177 Chr.1: 66355486-66355486 on Build GRCh38
rs766188915 Chr.1: 66355510-66355510 on Build GRCh38
rs774300682 Chr.1: 66355487-66355487 on Build GRCh38
rs778710865 Chr.1: 66355619-66355619 on Build GRCh38
rs754502755 Chr.1: 66355637-66355638 on Build GRCh38
rs528278981 Chr.1: 66355512-66355512 on Build GRCh38
rs568504072 Chr.1: 66355635-66355635 on Build GRCh38
rs564355022 Chr.1: 66355492-66355492 on Build GRCh38
rs367582250 Chr.1: 66355495-66355495 on Build GRCh38
rs760276042 Chr.1: 66355651-66355651 on Build GRCh38

View Full Product Details