Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107778062-107778317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A3
Entrez Gene ID
1811
Gene Name
solute carrier family 26 member 3
Gene Aliases
CLD, DRA
UniGene
Hs.1650
Gene Chromosome Location
Chr.7: 107765467-107803233 on Build GRCh38
Transcripts
NM_000111.2 XM_011515867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6108142 c.1506G>T p.R502S Chr.7: 107778183-107778183 on Build GRCh38
5871506 c.1454G>A p.G485E Chr.7: 107778235-107778235 on Build GRCh38
6340339 c.1408-4A>T p.? Chr.7: 107778285-107778285 on Build GRCh38
3877059 c.1409T>G p.L470* Chr.7: 107778280-107778280 on Build GRCh38
744201 c.1483C>A p.Q495K Chr.7: 107778206-107778206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946125707 Chr.7: 107778108-107778108 on Build GRCh38
rs771896738 Chr.7: 107778143-107778143 on Build GRCh38
rs530484848 Chr.7: 107778283-107778284 on Build GRCh38
rs1007579231 Chr.7: 107778111-107778111 on Build GRCh38
rs376815427 Chr.7: 107778130-107778130 on Build GRCh38
rs745802192 Chr.7: 107778139-107778139 on Build GRCh38
rs150724234 Chr.7: 107778194-107778194 on Build GRCh38
rs761057828 Chr.7: 107778258-107778258 on Build GRCh38
rs778970535 Chr.7: 107778135-107778135 on Build GRCh38
rs200404042 Chr.7: 107778237-107778237 on Build GRCh38
rs1042298430 Chr.7: 107778117-107778117 on Build GRCh38
rs1050658838 Chr.7: 107778167-107778167 on Build GRCh38
rs199870537 Chr.7: 107778177-107778177 on Build GRCh38
rs386833455 Chr.7: 107778282-107778282 on Build GRCh38
rs753961089 Chr.7: 107778128-107778128 on Build GRCh38
rs746758186 Chr.7: 107778174-107778174 on Build GRCh38
rs993595530 Chr.7: 107778173-107778173 on Build GRCh38
rs779802136 Chr.7: 107778157-107778157 on Build GRCh38
rs386833457 Chr.7: 107778202-107778202 on Build GRCh38
rs753130798 Chr.7: 107778248-107778248 on Build GRCh38
rs774987077 Chr.7: 107778227-107778227 on Build GRCh38
rs753994994 Chr.7: 107778267-107778267 on Build GRCh38
rs764393881 Chr.7: 107778260-107778260 on Build GRCh38
rs889348672 Chr.7: 107778102-107778102 on Build GRCh38
rs768327121 Chr.7: 107778176-107778176 on Build GRCh38
rs757427764 Chr.7: 107778270-107778270 on Build GRCh38
rs1040032743 Chr.7: 107778251-107778251 on Build GRCh38
rs111750004 Chr.7: 107778180-107778180 on Build GRCh38
rs760086563 Chr.7: 107778236-107778236 on Build GRCh38
rs778828932 Chr.7: 107778283-107778283 on Build GRCh38
rs926125963 Chr.7: 107778124-107778124 on Build GRCh38
rs534566329 Chr.7: 107778097-107778097 on Build GRCh38
rs910313442 Chr.7: 107778217-107778217 on Build GRCh38
rs375977220 Chr.7: 107778191-107778191 on Build GRCh38

View Full Product Details