Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22602466-22602717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
-
UniGene
Hs.743508
Gene Chromosome Location
Chr.8: 22599601-22604150 on Build GRCh38
Transcripts
NM_001013842.2 NM_001198827.1 NM_173686.2 XM_017014122.1

Gene Symbol
LOC107986876
Entrez Gene ID
107986876
Gene Name
proline-rich proteoglycan 2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 22602708-22605328 on Build GRCh38
Transcripts
NM_001013842.2 NM_001198827.1 NM_173686.2 XM_017014122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
106690 c.890C>T p.S297F Chr.8: 22602547-22602547 on Build GRCh38
3647306 c.970G>A p.D324N Chr.8: 22602627-22602627 on Build GRCh38
1755668 c.933G>A p.R311R Chr.8: 22602590-22602590 on Build GRCh38
4841983 c.883G>A p.D295N Chr.8: 22602540-22602540 on Build GRCh38
6616735 c.932G>A p.R311Q Chr.8: 22602589-22602589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115118237 Chr.8: 22602502-22602502 on Build GRCh38
rs759338230 Chr.8: 22602618-22602618 on Build GRCh38
rs773229768 Chr.8: 22602512-22602512 on Build GRCh38
rs373709022 Chr.8: 22602538-22602538 on Build GRCh38
rs746191263 Chr.8: 22602575-22602575 on Build GRCh38
rs760952227 Chr.8: 22602653-22602653 on Build GRCh38
rs755134177 Chr.8: 22602529-22602529 on Build GRCh38
rs117289616 Chr.8: 22602588-22602588 on Build GRCh38
rs201757790 Chr.8: 22602578-22602578 on Build GRCh38
rs766267595 Chr.8: 22602541-22602541 on Build GRCh38
rs761082356 Chr.8: 22602561-22602562 on Build GRCh38
rs1051772764 Chr.8: 22602536-22602536 on Build GRCh38
rs906421101 Chr.8: 22602665-22602665 on Build GRCh38
rs774846062 Chr.8: 22602485-22602485 on Build GRCh38
rs751412565 Chr.8: 22602526-22602526 on Build GRCh38
rs1033499008 Chr.8: 22602687-22602687 on Build GRCh38
rs757800724 Chr.8: 22602567-22602567 on Build GRCh38
rs767222187 Chr.8: 22602642-22602642 on Build GRCh38
rs760643230 Chr.8: 22602498-22602498 on Build GRCh38
rs764127205 Chr.8: 22602675-22602675 on Build GRCh38
rs750895678 Chr.8: 22602694-22602694 on Build GRCh38
rs1014224949 Chr.8: 22602652-22602652 on Build GRCh38
rs768399676 Chr.8: 22602490-22602490 on Build GRCh38
rs140762609 Chr.8: 22602547-22602547 on Build GRCh38
rs773735096 Chr.8: 22602560-22602561 on Build GRCh38
rs752901421 Chr.8: 22602645-22602645 on Build GRCh38
rs1042320596 Chr.8: 22602638-22602638 on Build GRCh38
rs754429346 Chr.8: 22602676-22602676 on Build GRCh38
rs776165778 Chr.8: 22602492-22602492 on Build GRCh38
rs763044281 Chr.8: 22602514-22602514 on Build GRCh38
rs767732421 Chr.8: 22602537-22602537 on Build GRCh38
rs772927630 Chr.8: 22602585-22602585 on Build GRCh38
rs772616111 Chr.8: 22602544-22602544 on Build GRCh38
rs1010822156 Chr.8: 22602590-22602590 on Build GRCh38
rs34268501 Chr.8: 22602691-22602691 on Build GRCh38
rs766086269 Chr.8: 22602515-22602515 on Build GRCh38
rs942287245 Chr.8: 22602494-22602494 on Build GRCh38
rs929323811 Chr.8: 22602535-22602535 on Build GRCh38
rs774177566 Chr.8: 22602606-22602606 on Build GRCh38
rs764840983 Chr.8: 22602506-22602506 on Build GRCh38
rs779121265 Chr.8: 22602572-22602572 on Build GRCh38
rs752894404 Chr.8: 22602560-22602560 on Build GRCh38
rs201285147 Chr.8: 22602580-22602580 on Build GRCh38
rs756127332 Chr.8: 22602562-22602562 on Build GRCh38
rs369243879 Chr.8: 22602684-22602684 on Build GRCh38
rs754339042 Chr.8: 22602563-22602563 on Build GRCh38
rs770951331 Chr.8: 22602589-22602589 on Build GRCh38
rs182703906 Chr.8: 22602486-22602486 on Build GRCh38
rs758729214 Chr.8: 22602695-22602695 on Build GRCh38
rs747819385 Chr.8: 22602579-22602579 on Build GRCh38

View Full Product Details