Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39315832-39316089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9
Entrez Gene ID
735
Gene Name
complement component 9
Gene Aliases
ARMD15, C9D
UniGene
Hs.654443
Gene Chromosome Location
Chr.5: 39284275-39364553 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1068079 c.652G>A p.E218K Chr.5: 39315993-39315993 on Build GRCh38
1068078 c.702T>G p.N234K Chr.5: 39315943-39315943 on Build GRCh38
737770 c.734C>A p.P245H Chr.5: 39315911-39315911 on Build GRCh38
2689716 c.714T>C p.A238A Chr.5: 39315931-39315931 on Build GRCh38
2689712 c.812G>A p.R271Q Chr.5: 39315833-39315833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756910234 Chr.5: 39315956-39315956 on Build GRCh38
rs775577722 Chr.5: 39315861-39315861 on Build GRCh38
rs768789173 Chr.5: 39315989-39315989 on Build GRCh38
rs779905943 Chr.5: 39315981-39315981 on Build GRCh38
rs751239379 Chr.5: 39315930-39315930 on Build GRCh38
rs759488738 Chr.5: 39315944-39315944 on Build GRCh38
rs774253048 Chr.5: 39315924-39315924 on Build GRCh38
rs773607296 Chr.5: 39315882-39315882 on Build GRCh38
rs748222991 Chr.5: 39316056-39316056 on Build GRCh38
rs369467784 Chr.5: 39315869-39315869 on Build GRCh38
rs763744652 Chr.5: 39316015-39316015 on Build GRCh38
rs779818000 Chr.5: 39316034-39316034 on Build GRCh38
rs139000312 Chr.5: 39316021-39316021 on Build GRCh38
rs780870258 Chr.5: 39315960-39315960 on Build GRCh38
rs967752696 Chr.5: 39315881-39315881 on Build GRCh38
rs778884265 Chr.5: 39316051-39316051 on Build GRCh38
rs768274343 Chr.5: 39316060-39316060 on Build GRCh38
rs114751055 Chr.5: 39316009-39316009 on Build GRCh38
rs763685618 Chr.5: 39315911-39315911 on Build GRCh38
rs188115836 Chr.5: 39315862-39315862 on Build GRCh38
rs758235529 Chr.5: 39315889-39315889 on Build GRCh38
rs773484295 Chr.5: 39315998-39315998 on Build GRCh38
rs141625765 Chr.5: 39315994-39315994 on Build GRCh38
rs750144844 Chr.5: 39315885-39315885 on Build GRCh38
rs761402841 Chr.5: 39316020-39316020 on Build GRCh38
rs756049525 Chr.5: 39315977-39315977 on Build GRCh38
rs760535084 Chr.5: 39316037-39316037 on Build GRCh38
rs200777803 Chr.5: 39315913-39315913 on Build GRCh38
rs1046874570 Chr.5: 39315854-39315854 on Build GRCh38
rs760118971 Chr.5: 39316011-39316011 on Build GRCh38
rs752596584 Chr.5: 39316015-39316021 on Build GRCh38
rs751030525 Chr.5: 39316017-39316017 on Build GRCh38
rs757923990 Chr.5: 39315910-39315910 on Build GRCh38
rs755957609 Chr.5: 39316023-39316023 on Build GRCh38
rs199670039 Chr.5: 39316053-39316053 on Build GRCh38
rs1039789207 Chr.5: 39315962-39315962 on Build GRCh38
rs111478418 Chr.5: 39316030-39316030 on Build GRCh38
rs752260198 Chr.5: 39315892-39315892 on Build GRCh38
rs903055186 Chr.5: 39316003-39316003 on Build GRCh38
rs753810956 Chr.5: 39316047-39316047 on Build GRCh38
rs763866944 Chr.5: 39316039-39316041 on Build GRCh38
rs761034354 Chr.5: 39316007-39316007 on Build GRCh38
rs745635536 Chr.5: 39315969-39315969 on Build GRCh38
rs749205892 Chr.5: 39315987-39315987 on Build GRCh38
rs956317953 Chr.5: 39316002-39316002 on Build GRCh38
rs774610867 Chr.5: 39315871-39315871 on Build GRCh38
rs754889772 Chr.5: 39316049-39316049 on Build GRCh38
rs148389506 Chr.5: 39315993-39315993 on Build GRCh38
rs561709823 Chr.5: 39315872-39315872 on Build GRCh38
rs767488401 Chr.5: 39315947-39315948 on Build GRCh38

View Full Product Details