Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163494603-163494876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMA16
Entrez Gene ID
55319
Gene Name
translation machinery associated 16 homolog
Gene Aliases
C4orf43
UniGene
Hs.267446
Gene Chromosome Location
Chr.4: 163494521-163520539 on Build GRCh38
Transcripts
NM_018352.2

SNP Detail

SNP ID Chromosome Location
rs569380737 Chr.4: 163494760-163494760 on Build GRCh38
rs534363323 Chr.4: 163494809-163494809 on Build GRCh38
rs897686523 Chr.4: 163494697-163494697 on Build GRCh38
rs1055027803 Chr.4: 163494687-163494687 on Build GRCh38
rs760713807 Chr.4: 163494764-163494764 on Build GRCh38
rs766028314 Chr.4: 163494781-163494781 on Build GRCh38
rs1046080120 Chr.4: 163494696-163494696 on Build GRCh38
rs756527864 Chr.4: 163494822-163494822 on Build GRCh38
rs957777005 Chr.4: 163494816-163494816 on Build GRCh38
rs1054675784 Chr.4: 163494639-163494639 on Build GRCh38
rs370848427 Chr.4: 163494841-163494841 on Build GRCh38
rs376743723 Chr.4: 163494773-163494773 on Build GRCh38
rs778106745 Chr.4: 163494823-163494823 on Build GRCh38
rs772126543 Chr.4: 163494833-163494833 on Build GRCh38
rs554897304 Chr.4: 163494768-163494768 on Build GRCh38
rs1019014328 Chr.4: 163494729-163494729 on Build GRCh38
rs2276978 Chr.4: 163494640-163494640 on Build GRCh38
rs542207759 Chr.4: 163494710-163494710 on Build GRCh38
rs562084890 Chr.4: 163494712-163494712 on Build GRCh38
rs116596859 Chr.4: 163494856-163494856 on Build GRCh38
rs906311980 Chr.4: 163494665-163494665 on Build GRCh38
rs577357782 Chr.4: 163494631-163494631 on Build GRCh38
rs750518099 Chr.4: 163494801-163494801 on Build GRCh38
rs368992524 Chr.4: 163494767-163494767 on Build GRCh38
rs561867134 Chr.4: 163494719-163494719 on Build GRCh38
rs949359885 Chr.4: 163494693-163494693 on Build GRCh38
rs777983802 Chr.4: 163494757-163494757 on Build GRCh38
rs2276977 Chr.4: 163494778-163494778 on Build GRCh38
rs139765399 Chr.4: 163494721-163494721 on Build GRCh38
rs762690647 Chr.4: 163494780-163494780 on Build GRCh38
rs761832538 Chr.4: 163494844-163494844 on Build GRCh38
rs562949029 Chr.4: 163494675-163494675 on Build GRCh38
rs531876344 Chr.4: 163494688-163494688 on Build GRCh38
rs372448573 Chr.4: 163494770-163494770 on Build GRCh38
rs978179912 Chr.4: 163494648-163494648 on Build GRCh38
rs748485743 Chr.4: 163494752-163494752 on Build GRCh38
rs898626297 Chr.4: 163494713-163494713 on Build GRCh38
rs373737076 Chr.4: 163494817-163494817 on Build GRCh38
rs770075428 Chr.4: 163494755-163494755 on Build GRCh38
rs1004628068 Chr.4: 163494709-163494709 on Build GRCh38
rs554442301 Chr.4: 163494829-163494829 on Build GRCh38
rs912584213 Chr.4: 163494628-163494628 on Build GRCh38
rs527544425 Chr.4: 163494714-163494714 on Build GRCh38
rs766604686 Chr.4: 163494840-163494840 on Build GRCh38
rs1008471360 Chr.4: 163494727-163494727 on Build GRCh38
rs549601851 Chr.4: 163494750-163494750 on Build GRCh38
rs978107881 Chr.4: 163494802-163494802 on Build GRCh38
rs936261821 Chr.4: 163494630-163494630 on Build GRCh38
rs947702363 Chr.4: 163494826-163494826 on Build GRCh38
rs547677760 Chr.4: 163494716-163494716 on Build GRCh38
rs751012296 Chr.4: 163494794-163494794 on Build GRCh38
rs754464406 Chr.4: 163494812-163494812 on Build GRCh38
rs769064686 Chr.4: 163494698-163494698 on Build GRCh38
rs376731479 Chr.4: 163494825-163494825 on Build GRCh38
rs925105587 Chr.4: 163494655-163494655 on Build GRCh38
rs753189020 Chr.4: 163494818-163494818 on Build GRCh38
rs996099761 Chr.4: 163494715-163494715 on Build GRCh38
rs533130321 Chr.4: 163494741-163494741 on Build GRCh38
rs577495164 Chr.4: 163494831-163494831 on Build GRCh38
rs771060924 Chr.4: 163494762-163494762 on Build GRCh38

View Full Product Details