Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110492012-110492253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS29
Entrez Gene ID
51699
Gene Name
VPS29, retromer complex component
Gene Aliases
DC15, DC7, PEP11
UniGene
Hs.600114
Gene Chromosome Location
Chr.12: 110491523-110502140 on Build GRCh38
Transcripts
NM_001282150.1 NM_001282151.1 NM_016226.4 NM_057180.2 NR_104099.1 NR_104100.1 NR_104101.1 NR_104102.1 XM_006719459.3 XM_006719460.3 XM_011538484.2 XM_017019457.1 XM_017019458.1 XM_017019459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219492 c.486C>G p.T162T Chr.12: 110492068-110492068 on Build GRCh38
4785483 c.444-1G>T p.? Chr.12: 110492123-110492123 on Build GRCh38
3398341 c.464T>C p.V155A Chr.12: 110492102-110492102 on Build GRCh38
6219491 c.498C>G p.T166T Chr.12: 110492068-110492068 on Build GRCh38
692157 c.523G>A p.E175K Chr.12: 110492031-110492031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989966459 Chr.12: 110492208-110492208 on Build GRCh38
rs751662191 Chr.12: 110492137-110492137 on Build GRCh38
rs921100075 Chr.12: 110492226-110492226 on Build GRCh38
rs780159203 Chr.12: 110492126-110492126 on Build GRCh38
rs763788076 Chr.12: 110492068-110492068 on Build GRCh38
rs758521601 Chr.12: 110492109-110492109 on Build GRCh38
rs199805827 Chr.12: 110492154-110492154 on Build GRCh38
rs753354260 Chr.12: 110492084-110492084 on Build GRCh38
rs998992051 Chr.12: 110492044-110492044 on Build GRCh38
rs562446591 Chr.12: 110492199-110492199 on Build GRCh38
rs1027615810 Chr.12: 110492115-110492115 on Build GRCh38
rs193184475 Chr.12: 110492173-110492173 on Build GRCh38
rs983074811 Chr.12: 110492191-110492191 on Build GRCh38
rs750727644 Chr.12: 110492080-110492080 on Build GRCh38
rs367915953 Chr.12: 110492043-110492043 on Build GRCh38
rs755013089 Chr.12: 110492147-110492147 on Build GRCh38
rs563713658 Chr.12: 110492150-110492150 on Build GRCh38
rs1031394888 Chr.12: 110492102-110492102 on Build GRCh38
rs755946036 Chr.12: 110492051-110492051 on Build GRCh38

View Full Product Details