Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47857629-47857856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VDR
Entrez Gene ID
7421
Gene Name
vitamin D (1,25- dihydroxyvitamin D3) receptor
Gene Aliases
NR1I1, PPP1R163
UniGene
Hs.524368
Gene Chromosome Location
Chr.12: 47841537-47905031 on Build GRCh38
Transcripts
NM_000376.2 NM_001017535.1 NM_001017536.1 XM_006719587.3 XM_011538720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671079 c.311G>T p.R104L Chr.12: 47857655-47857655 on Build GRCh38
4831384 c.318G>A p.M106I Chr.12: 47857648-47857648 on Build GRCh38
4831385 c.222G>A p.M74I Chr.12: 47857648-47857648 on Build GRCh38
6819772 c.333G>A p.K111K Chr.12: 47857633-47857633 on Build GRCh38
6819773 c.237G>A p.K79K Chr.12: 47857633-47857633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529556629 Chr.12: 47857796-47857796 on Build GRCh38
rs772197934 Chr.12: 47857678-47857678 on Build GRCh38
rs11168267 Chr.12: 47857759-47857759 on Build GRCh38
rs576216536 Chr.12: 47857690-47857690 on Build GRCh38
rs571587967 Chr.12: 47857823-47857823 on Build GRCh38
rs563947382 Chr.12: 47857795-47857795 on Build GRCh38
rs752684532 Chr.12: 47857701-47857701 on Build GRCh38
rs905516413 Chr.12: 47857824-47857824 on Build GRCh38
rs543332604 Chr.12: 47857807-47857807 on Build GRCh38
rs199705103 Chr.12: 47857656-47857656 on Build GRCh38
rs1037285266 Chr.12: 47857803-47857803 on Build GRCh38
rs555067950 Chr.12: 47857696-47857696 on Build GRCh38
rs778027901 Chr.12: 47857821-47857821 on Build GRCh38
rs921245742 Chr.12: 47857756-47857756 on Build GRCh38
rs1014568757 Chr.12: 47857811-47857811 on Build GRCh38
rs557402101 Chr.12: 47857754-47857754 on Build GRCh38
rs551270527 Chr.12: 47857693-47857693 on Build GRCh38
rs140896907 Chr.12: 47857651-47857651 on Build GRCh38
rs879593461 Chr.12: 47857704-47857704 on Build GRCh38
rs974776402 Chr.12: 47857762-47857762 on Build GRCh38
rs780886928 Chr.12: 47857667-47857667 on Build GRCh38
rs989150640 Chr.12: 47857832-47857832 on Build GRCh38
rs543608950 Chr.12: 47857761-47857761 on Build GRCh38
rs1028900059 Chr.12: 47857776-47857776 on Build GRCh38
rs1036246052 Chr.12: 47857732-47857732 on Build GRCh38
rs11168266 Chr.12: 47857750-47857750 on Build GRCh38
rs749763626 Chr.12: 47857655-47857655 on Build GRCh38
rs773064084 Chr.12: 47857692-47857693 on Build GRCh38
rs540639036 Chr.12: 47857753-47857753 on Build GRCh38
rs772581961 Chr.12: 47857806-47857806 on Build GRCh38
rs762850512 Chr.12: 47857739-47857739 on Build GRCh38
rs1003025930 Chr.12: 47857721-47857721 on Build GRCh38
rs373023178 Chr.12: 47857817-47857817 on Build GRCh38

View Full Product Details