Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129433811-129434036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBD4
Entrez Gene ID
8930
Gene Name
methyl-CpG binding domain 4, DNA glycosylase
Gene Aliases
MED1
UniGene
Hs.35947
Gene Chromosome Location
Chr.3: 129430944-129440179 on Build GRCh38
Transcripts
NM_001276270.1 NM_001276271.1 NM_001276272.1 NM_001276273.1 NM_003925.2 XM_011513268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587201 c.1324C>T p.R442W Chr.3: 129433937-129433937 on Build GRCh38
3587200 c.1404G>A p.R468R Chr.3: 129433857-129433857 on Build GRCh38
4113320 c.1318C>A p.P440T Chr.3: 129433943-129433943 on Build GRCh38
479397 c.1377T>C p.L459L Chr.3: 129433884-129433884 on Build GRCh38
3396517 c.1311A>G p.K437K Chr.3: 129433950-129433950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200137916 Chr.3: 129433966-129433966 on Build GRCh38
rs749993260 Chr.3: 129433995-129433996 on Build GRCh38
rs758935058 Chr.3: 129433977-129433977 on Build GRCh38
rs760432119 Chr.3: 129433975-129433975 on Build GRCh38
rs767140302 Chr.3: 129433875-129433875 on Build GRCh38
rs764777920 Chr.3: 129433987-129433990 on Build GRCh38
rs781435274 Chr.3: 129433927-129433927 on Build GRCh38
rs373218090 Chr.3: 129433969-129433969 on Build GRCh38
rs778144837 Chr.3: 129433914-129433914 on Build GRCh38
rs747394347 Chr.3: 129433919-129433919 on Build GRCh38
rs200995882 Chr.3: 129433936-129433936 on Build GRCh38
rs763910366 Chr.3: 129433869-129433869 on Build GRCh38
rs766007056 Chr.3: 129433881-129433881 on Build GRCh38
rs527715494 Chr.3: 129433871-129433871 on Build GRCh38
rs373984872 Chr.3: 129433893-129433893 on Build GRCh38
rs78782061 Chr.3: 129433861-129433861 on Build GRCh38
rs765916875 Chr.3: 129433976-129433976 on Build GRCh38
rs768532890 Chr.3: 129433965-129433965 on Build GRCh38
rs1054351182 Chr.3: 129433899-129433899 on Build GRCh38
rs757499279 Chr.3: 129433922-129433922 on Build GRCh38
rs376663356 Chr.3: 129433962-129433962 on Build GRCh38
rs761551915 Chr.3: 129433967-129433967 on Build GRCh38
rs895290906 Chr.3: 129433971-129433971 on Build GRCh38
rs762115445 Chr.3: 129433877-129433877 on Build GRCh38
rs948070065 Chr.3: 129433908-129433908 on Build GRCh38
rs772947874 Chr.3: 129433972-129433972 on Build GRCh38
rs535692976 Chr.3: 129433944-129433944 on Build GRCh38
rs776455996 Chr.3: 129433892-129433893 on Build GRCh38
rs935633452 Chr.3: 129433963-129433963 on Build GRCh38
rs757615567 Chr.3: 129433998-129433998 on Build GRCh38
rs746052367 Chr.3: 129433999-129433999 on Build GRCh38
rs749184158 Chr.3: 129433964-129433964 on Build GRCh38
rs761512828 Chr.3: 129433872-129433872 on Build GRCh38
rs551945879 Chr.3: 129433858-129433858 on Build GRCh38
rs769817977 Chr.3: 129433937-129433937 on Build GRCh38
rs893067080 Chr.3: 129433904-129433904 on Build GRCh38
rs201776693 Chr.3: 129433878-129433878 on Build GRCh38
rs769691633 Chr.3: 129433856-129433856 on Build GRCh38
rs761511435 Chr.3: 129433950-129433950 on Build GRCh38
rs377548810 Chr.3: 129433997-129433997 on Build GRCh38

View Full Product Details