Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201468354-201468615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLDA3
Entrez Gene ID
23612
Gene Name
pleckstrin homology like domain family A member 3
Gene Aliases
TIH1
UniGene
Hs.153944
Gene Chromosome Location
Chr.1: 201464284-201469171 on Build GRCh38
Transcripts
NM_012396.4 NR_073080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6745425 c.373C>A p.L125I Chr.1: 201468414-201468414 on Build GRCh38
237959 c.258C>A p.R86R Chr.1: 201468529-201468529 on Build GRCh38
6745424 c.220A>G p.T74A Chr.1: 201468567-201468567 on Build GRCh38
6745426 c.355A>G p.S119G Chr.1: 201468432-201468432 on Build GRCh38
1295687 c.202G>T p.G68W Chr.1: 201468585-201468585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148133646 Chr.1: 201468532-201468532 on Build GRCh38
rs773631516 Chr.1: 201468478-201468478 on Build GRCh38
rs1021246786 Chr.1: 201468463-201468463 on Build GRCh38
rs201117567 Chr.1: 201468412-201468412 on Build GRCh38
rs773865804 Chr.1: 201468464-201468464 on Build GRCh38
rs779197994 Chr.1: 201468430-201468430 on Build GRCh38
rs983582090 Chr.1: 201468431-201468431 on Build GRCh38
rs973217882 Chr.1: 201468511-201468511 on Build GRCh38
rs777599476 Chr.1: 201468592-201468600 on Build GRCh38
rs754246908 Chr.1: 201468489-201468489 on Build GRCh38
rs535862454 Chr.1: 201468582-201468582 on Build GRCh38
rs374733283 Chr.1: 201468544-201468544 on Build GRCh38
rs752650825 Chr.1: 201468436-201468436 on Build GRCh38
rs775166638 Chr.1: 201468375-201468375 on Build GRCh38
rs757849606 Chr.1: 201468417-201468417 on Build GRCh38
rs771495514 Chr.1: 201468580-201468580 on Build GRCh38
rs768232818 Chr.1: 201468451-201468451 on Build GRCh38
rs745790757 Chr.1: 201468439-201468439 on Build GRCh38
rs779812055 Chr.1: 201468441-201468441 on Build GRCh38
rs762388991 Chr.1: 201468396-201468396 on Build GRCh38
rs866347793 Chr.1: 201468551-201468551 on Build GRCh38
rs149004059 Chr.1: 201468429-201468429 on Build GRCh38
rs749008469 Chr.1: 201468444-201468444 on Build GRCh38
rs977054068 Chr.1: 201468585-201468585 on Build GRCh38
rs752855773 Chr.1: 201468509-201468509 on Build GRCh38
rs570267782 Chr.1: 201468380-201468380 on Build GRCh38
rs760907600 Chr.1: 201468415-201468415 on Build GRCh38
rs756041459 Chr.1: 201468529-201468529 on Build GRCh38
rs371562100 Chr.1: 201468496-201468496 on Build GRCh38
rs903637253 Chr.1: 201468588-201468588 on Build GRCh38
rs780954365 Chr.1: 201468438-201468438 on Build GRCh38
rs750821365 Chr.1: 201468414-201468414 on Build GRCh38
rs777146617 Chr.1: 201468587-201468587 on Build GRCh38
rs759941523 Chr.1: 201468594-201468594 on Build GRCh38
rs146948067 Chr.1: 201468574-201468574 on Build GRCh38
rs754158710 Chr.1: 201468427-201468427 on Build GRCh38
rs762576508 Chr.1: 201468377-201468377 on Build GRCh38
rs115283578 Chr.1: 201468440-201468440 on Build GRCh38
rs768573971 Chr.1: 201468521-201468521 on Build GRCh38
rs772663791 Chr.1: 201468475-201468475 on Build GRCh38
rs766904243 Chr.1: 201468487-201468487 on Build GRCh38
rs927301457 Chr.1: 201468470-201468470 on Build GRCh38
rs764215820 Chr.1: 201468522-201468522 on Build GRCh38
rs368012786 Chr.1: 201468469-201468469 on Build GRCh38
rs747623193 Chr.1: 201468381-201468382 on Build GRCh38
rs761222419 Chr.1: 201468484-201468484 on Build GRCh38
rs773847325 Chr.1: 201468392-201468392 on Build GRCh38
rs766818604 Chr.1: 201468419-201468419 on Build GRCh38
rs772761687 Chr.1: 201468572-201468572 on Build GRCh38
rs765668286 Chr.1: 201468597-201468597 on Build GRCh38
rs778387683 Chr.1: 201468549-201468549 on Build GRCh38
rs757243816 Chr.1: 201468437-201468437 on Build GRCh38
rs980157855 Chr.1: 201468459-201468459 on Build GRCh38
rs758452055 Chr.1: 201468516-201468516 on Build GRCh38
rs747803789 Chr.1: 201468550-201468550 on Build GRCh38
rs767852163 Chr.1: 201468405-201468405 on Build GRCh38
rs771531245 Chr.1: 201468556-201468556 on Build GRCh38

View Full Product Details