Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34983330-34983589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG3
Entrez Gene ID
3980
Gene Name
DNA ligase 3
Gene Aliases
LIG2
UniGene
Hs.100299
Gene Chromosome Location
Chr.17: 34980498-35009169 on Build GRCh38
Transcripts
NM_002311.4 NM_013975.3 XM_005257970.3 XM_006721896.3 XM_017024624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7230782 c.502G>C p.E168Q Chr.17: 34983507-34983507 on Build GRCh38
4623524 c.83G>A p.G28D Chr.17: 34983349-34983349 on Build GRCh38
6586959 c.517G>T p.E173* Chr.17: 34983522-34983522 on Build GRCh38
5924079 c.116C>T p.P39L Chr.17: 34983382-34983382 on Build GRCh38
4623523 c.344G>A p.G115D Chr.17: 34983349-34983349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763661439 Chr.17: 34983547-34983547 on Build GRCh38
rs61749869 Chr.17: 34983476-34983476 on Build GRCh38
rs761006623 Chr.17: 34983555-34983555 on Build GRCh38
rs377402837 Chr.17: 34983551-34983551 on Build GRCh38
rs756221253 Chr.17: 34983465-34983465 on Build GRCh38
rs752439656 Chr.17: 34983445-34983445 on Build GRCh38
rs1008833198 Chr.17: 34983378-34983378 on Build GRCh38
rs769477703 Chr.17: 34983532-34983532 on Build GRCh38
rs969144771 Chr.17: 34983485-34983485 on Build GRCh38
rs1021764612 Chr.17: 34983387-34983387 on Build GRCh38
rs939939370 Chr.17: 34983482-34983482 on Build GRCh38
rs772165438 Chr.17: 34983487-34983488 on Build GRCh38
rs775715005 Chr.17: 34983353-34983353 on Build GRCh38
rs577965471 Chr.17: 34983394-34983394 on Build GRCh38
rs777687871 Chr.17: 34983486-34983486 on Build GRCh38
rs750964359 Chr.17: 34983559-34983559 on Build GRCh38
rs771748154 Chr.17: 34983480-34983480 on Build GRCh38
rs745989900 Chr.17: 34983466-34983467 on Build GRCh38
rs558617611 Chr.17: 34983368-34983368 on Build GRCh38
rs770933343 Chr.17: 34983508-34983508 on Build GRCh38
rs776731503 Chr.17: 34983522-34983522 on Build GRCh38
rs537076655 Chr.17: 34983457-34983457 on Build GRCh38
rs774993954 Chr.17: 34983533-34983533 on Build GRCh38
rs146850795 Chr.17: 34983450-34983450 on Build GRCh38
rs746783203 Chr.17: 34983495-34983495 on Build GRCh38
rs772050625 Chr.17: 34983451-34983451 on Build GRCh38
rs764933003 Chr.17: 34983431-34983431 on Build GRCh38
rs991826335 Chr.17: 34983468-34983468 on Build GRCh38
rs770559358 Chr.17: 34983382-34983382 on Build GRCh38
rs368166360 Chr.17: 34983456-34983456 on Build GRCh38
rs145715449 Chr.17: 34983477-34983477 on Build GRCh38
rs979187016 Chr.17: 34983419-34983419 on Build GRCh38
rs762110291 Chr.17: 34983555-34983555 on Build GRCh38
rs542768230 Chr.17: 34983557-34983557 on Build GRCh38
rs762734586 Chr.17: 34983545-34983545 on Build GRCh38
rs781675755 Chr.17: 34983459-34983459 on Build GRCh38
rs370744567 Chr.17: 34983464-34983464 on Build GRCh38
rs775892244 Chr.17: 34983545-34983546 on Build GRCh38
rs573822077 Chr.17: 34983528-34983528 on Build GRCh38
rs149835429 Chr.17: 34983366-34983366 on Build GRCh38
rs747853708 Chr.17: 34983358-34983358 on Build GRCh38
rs759130376 Chr.17: 34983407-34983407 on Build GRCh38
rs767975337 Chr.17: 34983556-34983556 on Build GRCh38
rs892033365 Chr.17: 34983379-34983379 on Build GRCh38
rs760720520 Chr.17: 34983369-34983369 on Build GRCh38
rs968825832 Chr.17: 34983403-34983403 on Build GRCh38

View Full Product Details